Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24192195-24192701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TM9SF1
Entrez Gene ID
10548
Gene Name
transmembrane 9 superfamily member 1
Gene Aliases
HMP70, MP70
UniGene
Hs.91586
Gene Chromosome Location
Chr.14: 24189143-24195736 on Build GRCh38
Transcripts
NM_001014842.2 NM_001289006.1 NM_006405.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3401257 c.981G>A p.M327I Chr.14: 24192343-24192343 on Build GRCh38
4620684 c.1078T>C p.S360P Chr.14: 24192246-24192246 on Build GRCh38
347852 c.967G>A p.G323S Chr.14: 24192648-24192648 on Build GRCh38
5158918 c.936C>T p.G312G Chr.14: 24192679-24192679 on Build GRCh38
1369320 c.936C>T p.G312G Chr.14: 24192679-24192679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759292419 Chr.14: 24192249-24192249 on Build GRCh38
rs974642441 Chr.14: 24192428-24192428 on Build GRCh38
rs200901309 Chr.14: 24192399-24192399 on Build GRCh38
rs748251764 Chr.14: 24192328-24192328 on Build GRCh38
rs375072432 Chr.14: 24192451-24192451 on Build GRCh38
rs1055912110 Chr.14: 24192508-24192508 on Build GRCh38
rs1029530474 Chr.14: 24192434-24192434 on Build GRCh38
rs2281704 Chr.14: 24192500-24192500 on Build GRCh38
rs752298150 Chr.14: 24192251-24192251 on Build GRCh38
rs750924668 Chr.14: 24192390-24192390 on Build GRCh38
rs779446510 Chr.14: 24192293-24192293 on Build GRCh38
rs774077277 Chr.14: 24192235-24192235 on Build GRCh38
rs45543831 Chr.14: 24192531-24192531 on Build GRCh38
rs751961668 Chr.14: 24192406-24192406 on Build GRCh38
rs747164062 Chr.14: 24192317-24192317 on Build GRCh38
rs200912771 Chr.14: 24192372-24192372 on Build GRCh38
rs757773223 Chr.14: 24192291-24192291 on Build GRCh38
rs890939607 Chr.14: 24192436-24192436 on Build GRCh38
rs375173543 Chr.14: 24192314-24192314 on Build GRCh38
rs968077836 Chr.14: 24192453-24192453 on Build GRCh38
rs748504968 Chr.14: 24192377-24192377 on Build GRCh38
rs745738523 Chr.14: 24192645-24192645 on Build GRCh38
rs2234108 Chr.14: 24192349-24192349 on Build GRCh38
rs748222066 Chr.14: 24192621-24192621 on Build GRCh38
rs772793483 Chr.14: 24192570-24192570 on Build GRCh38
rs2234109 Chr.14: 24192323-24192323 on Build GRCh38
rs774319231 Chr.14: 24192336-24192336 on Build GRCh38
rs765685488 Chr.14: 24192389-24192389 on Build GRCh38
rs368677641 Chr.14: 24192547-24192547 on Build GRCh38
rs760208261 Chr.14: 24192266-24192266 on Build GRCh38
rs781442540 Chr.14: 24192599-24192599 on Build GRCh38
rs2234107 Chr.14: 24192404-24192404 on Build GRCh38
rs754380741 Chr.14: 24192281-24192281 on Build GRCh38
rs780459055 Chr.14: 24192315-24192315 on Build GRCh38
rs3215724 Chr.14: 24192428-24192428 on Build GRCh38
rs762743284 Chr.14: 24192217-24192217 on Build GRCh38
rs66471337 Chr.14: 24192483-24192483 on Build GRCh38
rs746790303 Chr.14: 24192670-24192670 on Build GRCh38
rs1018169562 Chr.14: 24192445-24192445 on Build GRCh38
rs775320385 Chr.14: 24192664-24192664 on Build GRCh38
rs768797845 Chr.14: 24192322-24192322 on Build GRCh38
rs902447506 Chr.14: 24192571-24192571 on Build GRCh38
rs992182681 Chr.14: 24192506-24192506 on Build GRCh38
rs111435072 Chr.14: 24192433-24192433 on Build GRCh38
rs149991411 Chr.14: 24192679-24192679 on Build GRCh38
rs749650518 Chr.14: 24192250-24192250 on Build GRCh38
rs779358284 Chr.14: 24192345-24192345 on Build GRCh38
rs746330999 Chr.14: 24192371-24192371 on Build GRCh38
rs79141239 Chr.14: 24192637-24192637 on Build GRCh38
rs935425519 Chr.14: 24192286-24192286 on Build GRCh38
rs769859678 Chr.14: 24192635-24192635 on Build GRCh38
rs893192682 Chr.14: 24192512-24192512 on Build GRCh38
rs150862560 Chr.14: 24192311-24192311 on Build GRCh38
rs905551434 Chr.14: 24192355-24192355 on Build GRCh38
rs367910573 Chr.14: 24192366-24192366 on Build GRCh38
rs540575479 Chr.14: 24192618-24192618 on Build GRCh38
rs534280794 Chr.14: 24192417-24192417 on Build GRCh38
rs763443997 Chr.14: 24192277-24192277 on Build GRCh38
rs371868078 Chr.14: 24192607-24192607 on Build GRCh38
rs777923081 Chr.14: 24192224-24192224 on Build GRCh38
rs918878564 Chr.14: 24192432-24192432 on Build GRCh38
rs574423137 Chr.14: 24192498-24192498 on Build GRCh38
rs370479199 Chr.14: 24192472-24192472 on Build GRCh38
rs776299811 Chr.14: 24192678-24192678 on Build GRCh38
rs376919085 Chr.14: 24192365-24192365 on Build GRCh38
rs368386700 Chr.14: 24192330-24192330 on Build GRCh38
rs753570509 Chr.14: 24192519-24192519 on Build GRCh38
rs1006825058 Chr.14: 24192444-24192444 on Build GRCh38
rs759347812 Chr.14: 24192343-24192343 on Build GRCh38
rs770885852 Chr.14: 24192230-24192230 on Build GRCh38
rs768244427 Chr.14: 24192672-24192672 on Build GRCh38
rs771866179 Chr.14: 24192657-24192657 on Build GRCh38
rs930176315 Chr.14: 24192456-24192456 on Build GRCh38
rs750867563 Chr.14: 24192310-24192310 on Build GRCh38
rs755220140 Chr.14: 24192598-24192598 on Build GRCh38
rs557307298 Chr.14: 24192535-24192535 on Build GRCh38
rs370975656 Chr.14: 24192216-24192216 on Build GRCh38

View Full Product Details