Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35539217-35539716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RALGAPA1
Entrez Gene ID
253959
Gene Name
Ral GTPase activating protein catalytic alpha subunit 1
Gene Aliases
GARNL1, GRIPE, RalGAPalpha1, TULIP1, p240
UniGene
Hs.113150
Gene Chromosome Location
Chr.14: 35538352-35809304 on Build GRCh38
Transcripts
NM_001283043.1 NM_014990.1 NM_194301.2 XM_005267491.3 XM_005267492.3 XM_005267493.3 XM_005267498.4 XM_006720098.2 XM_006720099.2 XM_006720100.3 XM_006720101.2 XM_006720102.3 XM_006720103.2 XM_017021141.1 XM_017021142.1 XM_017021144.1 XM_017021145.1 XM_017021146.1 XM_017021147.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
955510 c.6202C>T p.R2068C Chr.14: 35539592-35539592 on Build GRCh38
4502788 c.6223C>T p.R2075C Chr.14: 35539571-35539571 on Build GRCh38
4913414 c.6194G>A p.S2065N Chr.14: 35539600-35539600 on Build GRCh38
5462381 c.6201C>T p.S2067S Chr.14: 35539593-35539593 on Build GRCh38
955511 c.6188G>A p.R2063Q Chr.14: 35539606-35539606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745391407 Chr.14: 35539492-35539492 on Build GRCh38
rs927524565 Chr.14: 35539274-35539274 on Build GRCh38
rs753202872 Chr.14: 35539649-35539649 on Build GRCh38
rs546179294 Chr.14: 35539343-35539343 on Build GRCh38
rs763164034 Chr.14: 35539635-35539635 on Build GRCh38
rs937503657 Chr.14: 35539306-35539306 on Build GRCh38
rs982084031 Chr.14: 35539263-35539263 on Build GRCh38
rs756904690 Chr.14: 35539574-35539574 on Build GRCh38
rs933274713 Chr.14: 35539483-35539489 on Build GRCh38
rs914283496 Chr.14: 35539339-35539340 on Build GRCh38
rs771683356 Chr.14: 35539666-35539678 on Build GRCh38
rs771099748 Chr.14: 35539639-35539639 on Build GRCh38
rs989867039 Chr.14: 35539325-35539325 on Build GRCh38
rs777325079 Chr.14: 35539571-35539571 on Build GRCh38
rs769817636 Chr.14: 35539616-35539616 on Build GRCh38
rs945733400 Chr.14: 35539342-35539343 on Build GRCh38
rs759942457 Chr.14: 35539647-35539647 on Build GRCh38
rs866964134 Chr.14: 35539389-35539389 on Build GRCh38
rs773282922 Chr.14: 35539622-35539622 on Build GRCh38
rs200916773 Chr.14: 35539426-35539429 on Build GRCh38
rs34483694 Chr.14: 35539350-35539350 on Build GRCh38
rs201783643 Chr.14: 35539667-35539667 on Build GRCh38
rs763519205 Chr.14: 35539662-35539662 on Build GRCh38
rs56329672 Chr.14: 35539688-35539688 on Build GRCh38
rs766740429 Chr.14: 35539496-35539496 on Build GRCh38
rs200066443 Chr.14: 35539591-35539591 on Build GRCh38
rs746958271 Chr.14: 35539614-35539614 on Build GRCh38
rs763219505 Chr.14: 35539491-35539491 on Build GRCh38
rs778883344 Chr.14: 35539583-35539583 on Build GRCh38
rs867547466 Chr.14: 35539472-35539472 on Build GRCh38
rs529797226 Chr.14: 35539687-35539687 on Build GRCh38
rs761187454 Chr.14: 35539654-35539654 on Build GRCh38
rs767836197 Chr.14: 35539648-35539648 on Build GRCh38
rs760065187 Chr.14: 35539502-35539502 on Build GRCh38
rs183245677 Chr.14: 35539497-35539497 on Build GRCh38
rs374827448 Chr.14: 35539520-35539520 on Build GRCh38
rs201977932 Chr.14: 35539570-35539570 on Build GRCh38
rs994241564 Chr.14: 35539601-35539601 on Build GRCh38
rs779899564 Chr.14: 35539597-35539597 on Build GRCh38
rs771973738 Chr.14: 35539592-35539592 on Build GRCh38
rs146562217 Chr.14: 35539573-35539573 on Build GRCh38

View Full Product Details