Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 59605266-59605790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RTN1
Entrez Gene ID
6252
Gene Name
reticulon 1
Gene Aliases
NSP
UniGene
Hs.368626
Gene Chromosome Location
Chr.14: 59595976-59871288 on Build GRCh38
Transcripts
NM_021136.2 NM_206852.2 XM_017021578.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4859412 c.368G>T p.R123M Chr.14: 59605408-59605408 on Build GRCh38
4534237 c.2065G>A p.E689K Chr.14: 59605415-59605415 on Build GRCh38
3706332 c.307C>A p.Q103K Chr.14: 59605469-59605469 on Build GRCh38
353864 c.2027C>T p.T676M Chr.14: 59605453-59605453 on Build GRCh38
4875589 c.339C>A p.F113L Chr.14: 59605437-59605437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976378659 Chr.14: 59605483-59605483 on Build GRCh38
rs917061075 Chr.14: 59605731-59605731 on Build GRCh38
rs777103897 Chr.14: 59605505-59605505 on Build GRCh38
rs934480680 Chr.14: 59605313-59605313 on Build GRCh38
rs190881786 Chr.14: 59605587-59605587 on Build GRCh38
rs11559112 Chr.14: 59605469-59605469 on Build GRCh38
rs375257603 Chr.14: 59605538-59605538 on Build GRCh38
rs540920811 Chr.14: 59605666-59605666 on Build GRCh38
rs754671597 Chr.14: 59605391-59605391 on Build GRCh38
rs868177518 Chr.14: 59605415-59605415 on Build GRCh38
rs201164896 Chr.14: 59605453-59605453 on Build GRCh38
rs1029772934 Chr.14: 59605586-59605586 on Build GRCh38
rs762014526 Chr.14: 59605333-59605333 on Build GRCh38
rs1950786 Chr.14: 59605305-59605305 on Build GRCh38
rs755932541 Chr.14: 59605306-59605306 on Build GRCh38
rs763292959 Chr.14: 59605361-59605361 on Build GRCh38
rs756934888 Chr.14: 59605464-59605464 on Build GRCh38
rs762876394 Chr.14: 59605552-59605552 on Build GRCh38
rs993088021 Chr.14: 59605680-59605680 on Build GRCh38
rs1023242561 Chr.14: 59605359-59605359 on Build GRCh38
rs948428137 Chr.14: 59605417-59605417 on Build GRCh38
rs751376066 Chr.14: 59605364-59605364 on Build GRCh38
rs745355021 Chr.14: 59605599-59605599 on Build GRCh38
rs759358005 Chr.14: 59605558-59605558 on Build GRCh38
rs777785849 Chr.14: 59605447-59605447 on Build GRCh38
rs375289920 Chr.14: 59605352-59605352 on Build GRCh38
rs747418333 Chr.14: 59605489-59605489 on Build GRCh38
rs138910153 Chr.14: 59605470-59605470 on Build GRCh38
rs769295785 Chr.14: 59605322-59605322 on Build GRCh38
rs948459271 Chr.14: 59605757-59605757 on Build GRCh38
rs564777046 Chr.14: 59605622-59605622 on Build GRCh38
rs371456922 Chr.14: 59605529-59605529 on Build GRCh38
rs766507615 Chr.14: 59605363-59605363 on Build GRCh38
rs969011632 Chr.14: 59605742-59605742 on Build GRCh38
rs544984508 Chr.14: 59605603-59605603 on Build GRCh38
rs868015145 Chr.14: 59605321-59605321 on Build GRCh38
rs752594754 Chr.14: 59605437-59605437 on Build GRCh38
rs769099249 Chr.14: 59605504-59605504 on Build GRCh38
rs765447478 Chr.14: 59605345-59605345 on Build GRCh38
rs756018897 Chr.14: 59605445-59605445 on Build GRCh38
rs767465823 Chr.14: 59605433-59605433 on Build GRCh38
rs1022253896 Chr.14: 59605604-59605604 on Build GRCh38
rs770408388 Chr.14: 59605537-59605537 on Build GRCh38
rs559054340 Chr.14: 59605763-59605763 on Build GRCh38
rs777226273 Chr.14: 59605331-59605331 on Build GRCh38
rs115046853 Chr.14: 59605665-59605665 on Build GRCh38
rs922263049 Chr.14: 59605526-59605526 on Build GRCh38
rs1950785 Chr.14: 59605673-59605673 on Build GRCh38
rs746601453 Chr.14: 59605339-59605340 on Build GRCh38
rs941168094 Chr.14: 59605574-59605574 on Build GRCh38
rs148862799 Chr.14: 59605452-59605452 on Build GRCh38
rs745594757 Chr.14: 59605761-59605761 on Build GRCh38
rs895536672 Chr.14: 59605386-59605386 on Build GRCh38

View Full Product Details