Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 67392237-67392737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPHN
Entrez Gene ID
10243
Gene Name
gephyrin
Gene Aliases
GEPH, GPH, GPHRYN, HKPX1, MOCODC
UniGene
Hs.208765
Gene Chromosome Location
Chr.14: 66507407-67735831 on Build GRCh38
Transcripts
XM_017020925.1 NM_016445.2 XM_011536644.2

Gene Symbol
PLEK2
Entrez Gene ID
26499
Gene Name
pleckstrin 2
Gene Aliases
-
UniGene
Hs.170473
Gene Chromosome Location
Chr.14: 67386979-67412200 on Build GRCh38
Transcripts
XM_017020925.1 NM_016445.2 XM_011536644.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4051935 c.716T>C p.L239P Chr.14: 67392381-67392381 on Build GRCh38
147799 c.650C>G p.S217C Chr.14: 67392681-67392681 on Build GRCh38
4647776 c.733A>G p.S245G Chr.14: 67392364-67392364 on Build GRCh38
5909386 c.734G>T p.S245I Chr.14: 67392363-67392363 on Build GRCh38
957166 c.705A>C p.E235D Chr.14: 67392392-67392392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs58627213 Chr.14: 67392683-67392683 on Build GRCh38
rs751845436 Chr.14: 67392436-67392436 on Build GRCh38
rs572561274 Chr.14: 67392435-67392435 on Build GRCh38
rs751794322 Chr.14: 67392335-67392335 on Build GRCh38
rs371552019 Chr.14: 67392621-67392621 on Build GRCh38
rs752820733 Chr.14: 67392680-67392680 on Build GRCh38
rs936971763 Chr.14: 67392439-67392439 on Build GRCh38
rs769319967 Chr.14: 67392637-67392638 on Build GRCh38
rs559778257 Chr.14: 67392664-67392664 on Build GRCh38
rs770670232 Chr.14: 67392372-67392372 on Build GRCh38
rs1032074520 Chr.14: 67392381-67392381 on Build GRCh38
rs762317692 Chr.14: 67392312-67392312 on Build GRCh38
rs1024598219 Chr.14: 67392296-67392296 on Build GRCh38
rs57113671 Chr.14: 67392717-67392717 on Build GRCh38
rs778742378 Chr.14: 67392709-67392709 on Build GRCh38
rs757053915 Chr.14: 67392477-67392477 on Build GRCh38
rs745461677 Chr.14: 67392387-67392387 on Build GRCh38
rs748045095 Chr.14: 67392350-67392350 on Build GRCh38
rs375283064 Chr.14: 67392432-67392432 on Build GRCh38
rs773755656 Chr.14: 67392429-67392429 on Build GRCh38
rs753776042 Chr.14: 67392700-67392700 on Build GRCh38
rs746500054 Chr.14: 67392636-67392636 on Build GRCh38
rs990039910 Chr.14: 67392483-67392483 on Build GRCh38
rs191982550 Chr.14: 67392578-67392578 on Build GRCh38
rs200259677 Chr.14: 67392357-67392357 on Build GRCh38
rs568439876 Chr.14: 67392350-67392350 on Build GRCh38
rs762289758 Chr.14: 67392421-67392421 on Build GRCh38
rs72719128 Chr.14: 67392487-67392487 on Build GRCh38
rs777534402 Chr.14: 67392639-67392640 on Build GRCh38
rs763496008 Chr.14: 67392667-67392667 on Build GRCh38
rs745577625 Chr.14: 67392628-67392628 on Build GRCh38
rs750806435 Chr.14: 67392319-67392319 on Build GRCh38
rs773888135 Chr.14: 67392283-67392283 on Build GRCh38
rs937393642 Chr.14: 67392538-67392538 on Build GRCh38
rs759738400 Chr.14: 67392445-67392445 on Build GRCh38
rs775064941 Chr.14: 67392401-67392401 on Build GRCh38
rs920677425 Chr.14: 67392523-67392523 on Build GRCh38
rs140572250 Chr.14: 67392679-67392679 on Build GRCh38
rs780962024 Chr.14: 67392608-67392608 on Build GRCh38
rs199743283 Chr.14: 67392674-67392674 on Build GRCh38
rs112316908 Chr.14: 67392568-67392568 on Build GRCh38
rs141642889 Chr.14: 67392394-67392394 on Build GRCh38
rs777476738 Chr.14: 67392356-67392356 on Build GRCh38
rs142133041 Chr.14: 67392497-67392498 on Build GRCh38
rs770547635 Chr.14: 67392278-67392278 on Build GRCh38
rs778556585 Chr.14: 67392380-67392380 on Build GRCh38
rs368604605 Chr.14: 67392456-67392456 on Build GRCh38
rs781276214 Chr.14: 67392340-67392340 on Build GRCh38
rs987353698 Chr.14: 67392481-67392481 on Build GRCh38
rs576642961 Chr.14: 67392501-67392501 on Build GRCh38
rs147214173 Chr.14: 67392425-67392425 on Build GRCh38
rs558187891 Chr.14: 67392488-67392488 on Build GRCh38
rs777715655 Chr.14: 67392469-67392469 on Build GRCh38
rs753723204 Chr.14: 67392476-67392476 on Build GRCh38
rs369611787 Chr.14: 67392292-67392292 on Build GRCh38
rs767551191 Chr.14: 67392450-67392450 on Build GRCh38
rs755638854 Chr.14: 67392658-67392658 on Build GRCh38
rs998918323 Chr.14: 67392665-67392665 on Build GRCh38
rs1036405416 Chr.14: 67392591-67392591 on Build GRCh38
rs757179307 Chr.14: 67392701-67392701 on Build GRCh38
rs148402806 Chr.14: 67392333-67392333 on Build GRCh38
rs916716992 Chr.14: 67392716-67392716 on Build GRCh38
rs777013541 Chr.14: 67392301-67392301 on Build GRCh38
rs771746544 Chr.14: 67392629-67392629 on Build GRCh38
rs765591207 Chr.14: 67392315-67392315 on Build GRCh38
rs756044497 Chr.14: 67392460-67392460 on Build GRCh38
rs758745359 Chr.14: 67392328-67392328 on Build GRCh38
rs755036157 Chr.14: 67392337-67392337 on Build GRCh38
rs999188777 Chr.14: 67392619-67392619 on Build GRCh38
rs771427999 Chr.14: 67392669-67392669 on Build GRCh38
rs748720607 Chr.14: 67392643-67392644 on Build GRCh38
rs768013481 Chr.14: 67392640-67392640 on Build GRCh38
rs1046481162 Chr.14: 67392551-67392551 on Build GRCh38
rs139783690 Chr.14: 67392681-67392681 on Build GRCh38
rs748099671 Chr.14: 67392342-67392342 on Build GRCh38
rs1050232896 Chr.14: 67392622-67392622 on Build GRCh38
rs767000603 Chr.14: 67392293-67392293 on Build GRCh38
rs1018707884 Chr.14: 67392657-67392657 on Build GRCh38
rs779481931 Chr.14: 67392633-67392633 on Build GRCh38
rs543744287 Chr.14: 67392550-67392550 on Build GRCh38
rs541568477 Chr.14: 67392583-67392583 on Build GRCh38
rs778746448 Chr.14: 67392530-67392530 on Build GRCh38

View Full Product Details