Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74669869-74670366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AREL1
Entrez Gene ID
9870
Gene Name
apoptosis resistant E3 ubiquitin protein ligase 1
Gene Aliases
FIEL1, KIAA0317
UniGene
Hs.497417
Gene Chromosome Location
Chr.14: 74661252-74713118 on Build GRCh38
Transcripts
NM_001039479.1 XM_005268244.3 XM_006720344.2 XM_011537415.2 XM_011537416.2 XM_011537417.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1740067 c.1787A>C p.K596T Chr.14: 74669948-74669948 on Build GRCh38
4529619 c.1628G>A p.R543H Chr.14: 74670107-74670107 on Build GRCh38
470237 c.1742C>T p.T581I Chr.14: 74669993-74669993 on Build GRCh38
1678131 c.1700C>A p.S567Y Chr.14: 74670035-74670035 on Build GRCh38
6032787 c.1665G>A p.A555A Chr.14: 74670070-74670070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374246822 Chr.14: 74669898-74669898 on Build GRCh38
rs541797898 Chr.14: 74669934-74669934 on Build GRCh38
rs763622846 Chr.14: 74670150-74670150 on Build GRCh38
rs778707370 Chr.14: 74669990-74669990 on Build GRCh38
rs761997282 Chr.14: 74670056-74670056 on Build GRCh38
rs367584983 Chr.14: 74670013-74670013 on Build GRCh38
rs766865552 Chr.14: 74670061-74670061 on Build GRCh38
rs780978467 Chr.14: 74670071-74670071 on Build GRCh38
rs561716711 Chr.14: 74670060-74670060 on Build GRCh38
rs865971029 Chr.14: 74669979-74669979 on Build GRCh38
rs915385431 Chr.14: 74670192-74670192 on Build GRCh38
rs764170036 Chr.14: 74669954-74669954 on Build GRCh38
rs973044805 Chr.14: 74670266-74670266 on Build GRCh38
rs568845170 Chr.14: 74670246-74670246 on Build GRCh38
rs559637466 Chr.14: 74669907-74669907 on Build GRCh38
rs879063704 Chr.14: 74669904-74669904 on Build GRCh38
rs745831533 Chr.14: 74670091-74670091 on Build GRCh38
rs7493409 Chr.14: 74670335-74670335 on Build GRCh38
rs1042644769 Chr.14: 74670238-74670238 on Build GRCh38
rs760029346 Chr.14: 74670140-74670140 on Build GRCh38
rs779854887 Chr.14: 74670094-74670094 on Build GRCh38
rs368788804 Chr.14: 74670253-74670253 on Build GRCh38
rs571127129 Chr.14: 74670202-74670202 on Build GRCh38
rs537968123 Chr.14: 74670261-74670261 on Build GRCh38
rs762255128 Chr.14: 74669969-74669969 on Build GRCh38
rs748168958 Chr.14: 74670107-74670107 on Build GRCh38
rs199831228 Chr.14: 74670065-74670065 on Build GRCh38
rs17101845 Chr.14: 74670070-74670070 on Build GRCh38
rs774670383 Chr.14: 74670106-74670106 on Build GRCh38
rs775630852 Chr.14: 74670028-74670028 on Build GRCh38
rs201916897 Chr.14: 74670118-74670118 on Build GRCh38
rs2287399 Chr.14: 74670297-74670297 on Build GRCh38
rs934287270 Chr.14: 74669914-74669914 on Build GRCh38
rs755838503 Chr.14: 74670092-74670092 on Build GRCh38
rs767871926 Chr.14: 74669970-74669970 on Build GRCh38
rs767056134 Chr.14: 74670176-74670176 on Build GRCh38
rs376144017 Chr.14: 74670005-74670005 on Build GRCh38
rs929099306 Chr.14: 74670304-74670304 on Build GRCh38
rs760221707 Chr.14: 74670088-74670088 on Build GRCh38
rs374986772 Chr.14: 74670044-74670044 on Build GRCh38
rs746926987 Chr.14: 74670010-74670010 on Build GRCh38
rs550388590 Chr.14: 74670245-74670245 on Build GRCh38
rs768842320 Chr.14: 74670103-74670103 on Build GRCh38
rs751294093 Chr.14: 74670158-74670158 on Build GRCh38
rs374539009 Chr.14: 74669912-74669912 on Build GRCh38
rs781239421 Chr.14: 74670014-74670014 on Build GRCh38
rs752036105 Chr.14: 74669960-74669960 on Build GRCh38
rs757926059 Chr.14: 74670000-74670000 on Build GRCh38
rs372197341 Chr.14: 74670167-74670167 on Build GRCh38
rs774314378 Chr.14: 74670053-74670053 on Build GRCh38
rs538040717 Chr.14: 74669978-74669978 on Build GRCh38
rs762048025 Chr.14: 74670116-74670116 on Build GRCh38
rs772255519 Chr.14: 74670108-74670108 on Build GRCh38
rs759754971 Chr.14: 74669927-74669927 on Build GRCh38
rs773838767 Chr.14: 74670057-74670057 on Build GRCh38
rs371561437 Chr.14: 74670035-74670035 on Build GRCh38
rs775755649 Chr.14: 74669941-74669941 on Build GRCh38
rs952643381 Chr.14: 74670012-74670012 on Build GRCh38
rs966789957 Chr.14: 74670189-74670189 on Build GRCh38
rs750319173 Chr.14: 74670178-74670178 on Build GRCh38
rs777159161 Chr.14: 74670126-74670126 on Build GRCh38
rs370956665 Chr.14: 74669975-74669975 on Build GRCh38
rs752460182 Chr.14: 74669991-74669991 on Build GRCh38
rs77043880 Chr.14: 74670102-74670102 on Build GRCh38
rs776323663 Chr.14: 74669918-74669918 on Build GRCh38
rs763006454 Chr.14: 74669945-74669945 on Build GRCh38
rs766814268 Chr.14: 74670119-74670119 on Build GRCh38
rs76119092 Chr.14: 74670220-74670220 on Build GRCh38
rs745645128 Chr.14: 74670018-74670018 on Build GRCh38
rs372272809 Chr.14: 74670063-74670063 on Build GRCh38
rs773537772 Chr.14: 74670113-74670113 on Build GRCh38
rs775375955 Chr.14: 74670043-74670044 on Build GRCh38
rs769634579 Chr.14: 74670023-74670023 on Build GRCh38
rs922741619 Chr.14: 74669913-74669913 on Build GRCh38
rs796597875 Chr.14: 74670259-74670259 on Build GRCh38
rs768514896 Chr.14: 74669921-74669921 on Build GRCh38
rs763612853 Chr.14: 74670066-74670066 on Build GRCh38
rs955724947 Chr.14: 74670237-74670237 on Build GRCh38
rs1047929140 Chr.14: 74670308-74670308 on Build GRCh38

View Full Product Details