Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75960831-75961330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGFB3
Entrez Gene ID
7043
Gene Name
transforming growth factor beta 3
Gene Aliases
ARVD, ARVD1, LDS5, RNHF, TGF-beta3
UniGene
Hs.592317
Gene Chromosome Location
Chr.14: 75958097-75982022 on Build GRCh38
Transcripts
NM_003239.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4052519 c.1080+1G>A p.? Chr.14: 75960922-75960922 on Build GRCh38
4148258 c.1029C>A p.F343L Chr.14: 75960974-75960974 on Build GRCh38
5754630 c.974G>A p.R325Q Chr.14: 75961029-75961029 on Build GRCh38
6800733 c.1069A>G p.T357A Chr.14: 75960934-75960934 on Build GRCh38
1300839 c.979G>C p.D327H Chr.14: 75961024-75961024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922664831 Chr.14: 75961282-75961282 on Build GRCh38
rs908909961 Chr.14: 75961257-75961257 on Build GRCh38
rs543141551 Chr.14: 75961177-75961177 on Build GRCh38
rs748604413 Chr.14: 75960995-75960995 on Build GRCh38
rs1002241368 Chr.14: 75960865-75960865 on Build GRCh38
rs767224149 Chr.14: 75960909-75960909 on Build GRCh38
rs763527552 Chr.14: 75961043-75961043 on Build GRCh38
rs199714510 Chr.14: 75961095-75961095 on Build GRCh38
rs755947277 Chr.14: 75961090-75961090 on Build GRCh38
rs992984509 Chr.14: 75961167-75961167 on Build GRCh38
rs750299821 Chr.14: 75960924-75960924 on Build GRCh38
rs775311850 Chr.14: 75961034-75961034 on Build GRCh38
rs953148823 Chr.14: 75961245-75961245 on Build GRCh38
rs755894337 Chr.14: 75960948-75960948 on Build GRCh38
rs958019016 Chr.14: 75960897-75960897 on Build GRCh38
rs574227787 Chr.14: 75961096-75961096 on Build GRCh38
rs777862669 Chr.14: 75961308-75961308 on Build GRCh38
rs760903557 Chr.14: 75961064-75961066 on Build GRCh38
rs747427366 Chr.14: 75961024-75961024 on Build GRCh38
rs757163957 Chr.14: 75960881-75960881 on Build GRCh38
rs984374830 Chr.14: 75961220-75961220 on Build GRCh38
rs749485028 Chr.14: 75960949-75960949 on Build GRCh38
rs561869503 Chr.14: 75961194-75961194 on Build GRCh38
rs778990969 Chr.14: 75960983-75960983 on Build GRCh38
rs773846624 Chr.14: 75961048-75961048 on Build GRCh38
rs985871303 Chr.14: 75961249-75961249 on Build GRCh38
rs1036406592 Chr.14: 75961292-75961292 on Build GRCh38
rs781315120 Chr.14: 75960870-75960870 on Build GRCh38
rs934089067 Chr.14: 75961287-75961287 on Build GRCh38
rs762643638 Chr.14: 75961038-75961038 on Build GRCh38
rs750641855 Chr.14: 75960889-75960889 on Build GRCh38
rs948927134 Chr.14: 75961188-75961188 on Build GRCh38
rs564249022 Chr.14: 75961274-75961274 on Build GRCh38
rs767548724 Chr.14: 75961077-75961077 on Build GRCh38
rs761225134 Chr.14: 75961050-75961050 on Build GRCh38
rs1021920873 Chr.14: 75961002-75961002 on Build GRCh38
rs755084678 Chr.14: 75960963-75960963 on Build GRCh38
rs756264441 Chr.14: 75961045-75961045 on Build GRCh38
rs374988205 Chr.14: 75961224-75961224 on Build GRCh38
rs1020220506 Chr.14: 75961079-75961079 on Build GRCh38
rs143794337 Chr.14: 75961263-75961263 on Build GRCh38
rs772698647 Chr.14: 75961017-75961017 on Build GRCh38
rs558147236 Chr.14: 75961149-75961149 on Build GRCh38
rs752931875 Chr.14: 75961107-75961107 on Build GRCh38
rs572107401 Chr.14: 75961230-75961230 on Build GRCh38
rs977821614 Chr.14: 75961046-75961046 on Build GRCh38
rs773574487 Chr.14: 75961021-75961021 on Build GRCh38
rs201637508 Chr.14: 75961080-75961080 on Build GRCh38
rs771391164 Chr.14: 75961029-75961029 on Build GRCh38

View Full Product Details