Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 90263176-90263693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMC1
Entrez Gene ID
5700
Gene Name
proteasome 26S subunit, ATPase 1
Gene Aliases
P26S4, S4, p56
UniGene
Hs.356654
Gene Chromosome Location
Chr.14: 90256550-90272625 on Build GRCh38
Transcripts
NM_002802.2 XM_017021470.1 XM_017021471.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
958604 c.155-1G>A p.? Chr.14: 90263317-90263317 on Build GRCh38
1707692 c.277G>A p.E93K Chr.14: 90263440-90263440 on Build GRCh38
958605 c.194T>C p.L65S Chr.14: 90263357-90263357 on Build GRCh38
958606 c.229G>T p.E77* Chr.14: 90263392-90263392 on Build GRCh38
5716620 c.176G>A p.R59Q Chr.14: 90263339-90263339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963396778 Chr.14: 90263236-90263236 on Build GRCh38
rs935268267 Chr.14: 90263340-90263340 on Build GRCh38
rs750878841 Chr.14: 90263456-90263456 on Build GRCh38
rs751611393 Chr.14: 90263640-90263640 on Build GRCh38
rs185710849 Chr.14: 90263648-90263648 on Build GRCh38
rs4904663 Chr.14: 90263221-90263221 on Build GRCh38
rs766407157 Chr.14: 90263476-90263476 on Build GRCh38
rs563614835 Chr.14: 90263204-90263204 on Build GRCh38
rs746803374 Chr.14: 90263491-90263491 on Build GRCh38
rs773481271 Chr.14: 90263373-90263374 on Build GRCh38
rs528687469 Chr.14: 90263603-90263603 on Build GRCh38
rs763210784 Chr.14: 90263460-90263460 on Build GRCh38
rs973171188 Chr.14: 90263253-90263253 on Build GRCh38
rs774892342 Chr.14: 90263415-90263415 on Build GRCh38
rs745882963 Chr.14: 90263628-90263628 on Build GRCh38
rs1006943370 Chr.14: 90263574-90263574 on Build GRCh38
rs8015510 Chr.14: 90263640-90263640 on Build GRCh38
rs780995246 Chr.14: 90263623-90263623 on Build GRCh38
rs776990424 Chr.14: 90263278-90263284 on Build GRCh38
rs953178659 Chr.14: 90263272-90263272 on Build GRCh38
rs745382083 Chr.14: 90263370-90263370 on Build GRCh38
rs770805704 Chr.14: 90263338-90263338 on Build GRCh38
rs575189917 Chr.14: 90263533-90263533 on Build GRCh38
rs539482269 Chr.14: 90263448-90263448 on Build GRCh38
rs777595073 Chr.14: 90263335-90263335 on Build GRCh38
rs1018717903 Chr.14: 90263615-90263615 on Build GRCh38
rs1019591825 Chr.14: 90263271-90263271 on Build GRCh38
rs1008664266 Chr.14: 90263610-90263610 on Build GRCh38
rs1004520168 Chr.14: 90263226-90263226 on Build GRCh38
rs950642158 Chr.14: 90263436-90263436 on Build GRCh38
rs552798879 Chr.14: 90263459-90263459 on Build GRCh38
rs193920988 Chr.14: 90263450-90263450 on Build GRCh38
rs762749848 Chr.14: 90263388-90263388 on Build GRCh38
rs543431665 Chr.14: 90263537-90263537 on Build GRCh38
rs202028024 Chr.14: 90263647-90263647 on Build GRCh38
rs780262492 Chr.14: 90263267-90263267 on Build GRCh38
rs139285884 Chr.14: 90263481-90263481 on Build GRCh38
rs776582107 Chr.14: 90263339-90263339 on Build GRCh38
rs143172121 Chr.14: 90263394-90263394 on Build GRCh38
rs775133437 Chr.14: 90263387-90263387 on Build GRCh38
rs770134951 Chr.14: 90263300-90263301 on Build GRCh38
rs371091515 Chr.14: 90263406-90263406 on Build GRCh38
rs372025051 Chr.14: 90263303-90263303 on Build GRCh38
rs988488094 Chr.14: 90263375-90263375 on Build GRCh38
rs375594894 Chr.14: 90263616-90263616 on Build GRCh38
rs1003751682 Chr.14: 90263212-90263212 on Build GRCh38
rs76961711 Chr.14: 90263460-90263460 on Build GRCh38
rs1043073859 Chr.14: 90263527-90263527 on Build GRCh38
rs747218766 Chr.14: 90263553-90263553 on Build GRCh38
rs8022531 Chr.14: 90263587-90263587 on Build GRCh38
rs768450266 Chr.14: 90263653-90263653 on Build GRCh38
rs1029200972 Chr.14: 90263259-90263259 on Build GRCh38
rs568973877 Chr.14: 90263642-90263642 on Build GRCh38
rs566229848 Chr.14: 90263228-90263228 on Build GRCh38
rs576783805 Chr.14: 90263424-90263424 on Build GRCh38
rs74080702 Chr.14: 90263557-90263557 on Build GRCh38
rs1017632991 Chr.14: 90263572-90263572 on Build GRCh38
rs953145787 Chr.14: 90263596-90263596 on Build GRCh38
rs532665547 Chr.14: 90263218-90263218 on Build GRCh38
rs559932478 Chr.14: 90263595-90263595 on Build GRCh38
rs984541511 Chr.14: 90263442-90263442 on Build GRCh38
rs189839793 Chr.14: 90263262-90263262 on Build GRCh38
rs889845099 Chr.14: 90263569-90263569 on Build GRCh38
rs532417172 Chr.14: 90263576-90263576 on Build GRCh38
rs147908366 Chr.14: 90263471-90263471 on Build GRCh38
rs111737451 Chr.14: 90263220-90263220 on Build GRCh38
rs367719810 Chr.14: 90263291-90263291 on Build GRCh38
rs199839288 Chr.14: 90263357-90263357 on Build GRCh38
rs144593599 Chr.14: 90263337-90263337 on Build GRCh38
rs762095267 Chr.14: 90263300-90263305 on Build GRCh38
rs866336714 Chr.14: 90263235-90263235 on Build GRCh38
rs537026822 Chr.14: 90263383-90263383 on Build GRCh38
rs887332979 Chr.14: 90263546-90263546 on Build GRCh38
rs866431547 Chr.14: 90263545-90263545 on Build GRCh38
rs369205963 Chr.14: 90263488-90263488 on Build GRCh38
rs756578248 Chr.14: 90263457-90263457 on Build GRCh38
rs898830539 Chr.14: 90263584-90263584 on Build GRCh38

View Full Product Details