Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93237458-93237958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD7
Entrez Gene ID
55727
Gene Name
BTB domain containing 7
Gene Aliases
FUP1
UniGene
Hs.525549
Gene Chromosome Location
Chr.14: 93237550-93333092 on Build GRCh38
Transcripts
NM_001002860.3 NM_001289133.1

SNP Detail

SNP ID Chromosome Location
rs1001633451 Chr.14: 93237904-93237905 on Build GRCh38
rs978998792 Chr.14: 93237749-93237749 on Build GRCh38
rs944802667 Chr.14: 93237784-93237784 on Build GRCh38
rs913406576 Chr.14: 93237712-93237712 on Build GRCh38
rs1011969169 Chr.14: 93237795-93237795 on Build GRCh38
rs775841212 Chr.14: 93237696-93237696 on Build GRCh38
rs546273342 Chr.14: 93237651-93237651 on Build GRCh38
rs1034544704 Chr.14: 93237655-93237655 on Build GRCh38
rs906174249 Chr.14: 93237813-93237813 on Build GRCh38
rs749445890 Chr.14: 93237811-93237811 on Build GRCh38
rs45486899 Chr.14: 93237892-93237892 on Build GRCh38
rs979738574 Chr.14: 93237635-93237635 on Build GRCh38
rs974798221 Chr.14: 93237927-93237927 on Build GRCh38
rs974724892 Chr.14: 93237583-93237583 on Build GRCh38
rs993978104 Chr.14: 93237872-93237872 on Build GRCh38
rs964800562 Chr.14: 93237911-93237911 on Build GRCh38
rs781425896 Chr.14: 93237737-93237737 on Build GRCh38
rs545675825 Chr.14: 93237614-93237614 on Build GRCh38
rs762253502 Chr.14: 93237683-93237683 on Build GRCh38
rs1026995911 Chr.14: 93237829-93237829 on Build GRCh38
rs528501634 Chr.14: 93237689-93237689 on Build GRCh38
rs1050429913 Chr.14: 93237770-93237770 on Build GRCh38
rs574764379 Chr.14: 93237480-93237480 on Build GRCh38
rs943420029 Chr.14: 93237518-93237518 on Build GRCh38
rs78080111 Chr.14: 93237881-93237881 on Build GRCh38
rs561988391 Chr.14: 93237845-93237845 on Build GRCh38
rs191782726 Chr.14: 93237828-93237828 on Build GRCh38
rs1036821660 Chr.14: 93237797-93237797 on Build GRCh38
rs932011818 Chr.14: 93237755-93237756 on Build GRCh38
rs894898737 Chr.14: 93237777-93237777 on Build GRCh38
rs530806805 Chr.14: 93237851-93237851 on Build GRCh38
rs984895824 Chr.14: 93237496-93237496 on Build GRCh38
rs926179564 Chr.14: 93237750-93237750 on Build GRCh38
rs146915319 Chr.14: 93237688-93237688 on Build GRCh38
rs750656345 Chr.14: 93237873-93237873 on Build GRCh38
rs577306562 Chr.14: 93237618-93237618 on Build GRCh38
rs751959250 Chr.14: 93237609-93237609 on Build GRCh38
rs959800403 Chr.14: 93237684-93237684 on Build GRCh38
rs532932577 Chr.14: 93237929-93237929 on Build GRCh38
rs750928695 Chr.14: 93237805-93237805 on Build GRCh38
rs758793053 Chr.14: 93237491-93237491 on Build GRCh38
rs10147990 Chr.14: 93237508-93237508 on Build GRCh38

View Full Product Details