Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95709815-95710326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCL1A
Entrez Gene ID
8115
Gene Name
T-cell leukemia/lymphoma 1A
Gene Aliases
TCL1
UniGene
Hs.2484
Gene Chromosome Location
Chr.14: 95694160-95714196 on Build GRCh38
Transcripts
NM_001098725.1 NM_021966.2 NR_049726.1 XM_017021676.1 XM_017021677.1

SNP Detail

SNP ID Chromosome Location
rs148164020 Chr.14: 95709869-95709869 on Build GRCh38
rs1006033613 Chr.14: 95710153-95710153 on Build GRCh38
rs957088393 Chr.14: 95710223-95710223 on Build GRCh38
rs538360168 Chr.14: 95709939-95709939 on Build GRCh38
rs993776083 Chr.14: 95710102-95710102 on Build GRCh38
rs1017591531 Chr.14: 95710151-95710151 on Build GRCh38
rs887157411 Chr.14: 95710140-95710140 on Build GRCh38
rs45509491 Chr.14: 95710034-95710034 on Build GRCh38
rs1007628582 Chr.14: 95710279-95710279 on Build GRCh38
rs552935726 Chr.14: 95709911-95709911 on Build GRCh38
rs955265930 Chr.14: 95710213-95710213 on Build GRCh38
rs1005671823 Chr.14: 95709977-95709977 on Build GRCh38
rs530095532 Chr.14: 95710144-95710144 on Build GRCh38
rs150661254 Chr.14: 95710051-95710051 on Build GRCh38
rs899931720 Chr.14: 95710181-95710181 on Build GRCh38
rs897697652 Chr.14: 95710245-95710245 on Build GRCh38
rs958018480 Chr.14: 95710300-95710300 on Build GRCh38
rs890309740 Chr.14: 95710253-95710253 on Build GRCh38
rs986090790 Chr.14: 95709915-95709915 on Build GRCh38
rs191670127 Chr.14: 95710011-95710011 on Build GRCh38
rs777910656 Chr.14: 95710226-95710226 on Build GRCh38
rs751777129 Chr.14: 95710233-95710233 on Build GRCh38
rs565339302 Chr.14: 95710052-95710052 on Build GRCh38
rs986225118 Chr.14: 95710150-95710150 on Build GRCh38
rs977457491 Chr.14: 95710047-95710047 on Build GRCh38
rs996659886 Chr.14: 95710182-95710182 on Build GRCh38
rs559833859 Chr.14: 95709921-95709921 on Build GRCh38
rs990234261 Chr.14: 95710225-95710225 on Build GRCh38
rs993436374 Chr.14: 95710248-95710248 on Build GRCh38
rs1038482322 Chr.14: 95710166-95710166 on Build GRCh38
rs796173245 Chr.14: 95710063-95710063 on Build GRCh38
rs1045316906 Chr.14: 95709909-95709909 on Build GRCh38
rs556755593 Chr.14: 95710294-95710294 on Build GRCh38
rs567595569 Chr.14: 95709897-95709897 on Build GRCh38
rs17093288 Chr.14: 95709936-95709936 on Build GRCh38
rs182680123 Chr.14: 95710222-95710222 on Build GRCh38
rs561227835 Chr.14: 95710043-95710043 on Build GRCh38
rs948192180 Chr.14: 95710090-95710090 on Build GRCh38
rs550903655 Chr.14: 95709858-95709858 on Build GRCh38
rs958047615 Chr.14: 95710156-95710156 on Build GRCh38
rs1045226517 Chr.14: 95710096-95710096 on Build GRCh38
rs575734004 Chr.14: 95710132-95710132 on Build GRCh38
rs188793291 Chr.14: 95709956-95709956 on Build GRCh38
rs936876633 Chr.14: 95709937-95709937 on Build GRCh38
rs201931789 Chr.14: 95709897-95709898 on Build GRCh38
rs887158734 Chr.14: 95709968-95709968 on Build GRCh38
rs562874493 Chr.14: 95710130-95710130 on Build GRCh38
rs781117449 Chr.14: 95710301-95710301 on Build GRCh38
rs141964306 Chr.14: 95709879-95709879 on Build GRCh38
rs538696382 Chr.14: 95710288-95710288 on Build GRCh38
rs140113793 Chr.14: 95710070-95710070 on Build GRCh38
rs765305436 Chr.14: 95710139-95710139 on Build GRCh38
rs945563581 Chr.14: 95710185-95710185 on Build GRCh38
rs989734090 Chr.14: 95710163-95710163 on Build GRCh38
rs954743274 Chr.14: 95710142-95710142 on Build GRCh38
rs534797298 Chr.14: 95709900-95709900 on Build GRCh38
rs939598426 Chr.14: 95710291-95710291 on Build GRCh38
rs963897262 Chr.14: 95709991-95709991 on Build GRCh38
rs201350832 Chr.14: 95709896-95709897 on Build GRCh38
rs17093285 Chr.14: 95709867-95709867 on Build GRCh38
rs1017179610 Chr.14: 95709987-95709987 on Build GRCh38
rs546729242 Chr.14: 95710244-95710244 on Build GRCh38
rs1029505722 Chr.14: 95710183-95710183 on Build GRCh38
rs186505168 Chr.14: 95710249-95710249 on Build GRCh38
rs143282939 Chr.14: 95710117-95710117 on Build GRCh38
rs77989678 Chr.14: 95710220-95710220 on Build GRCh38
rs897649753 Chr.14: 95710097-95710097 on Build GRCh38
rs573241286 Chr.14: 95710048-95710048 on Build GRCh38
rs975315260 Chr.14: 95710010-95710010 on Build GRCh38
rs374503443 Chr.14: 95709985-95709985 on Build GRCh38
rs962205301 Chr.14: 95709892-95709892 on Build GRCh38
rs528444246 Chr.14: 95710243-95710243 on Build GRCh38
rs752679724 Chr.14: 95710219-95710219 on Build GRCh38

View Full Product Details