Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 100544817-100545341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEGAIN
Entrez Gene ID
57596
Gene Name
brain enriched guanylate kinase associated
Gene Aliases
-
UniGene
Hs.211751
Gene Chromosome Location
Chr.14: 100537147-100587404 on Build GRCh38
Transcripts
NM_001159531.1 NM_020836.3 XM_005267919.3 XM_005267920.4 XM_005267921.3 XM_011537031.1 XM_017021521.1 XM_017021522.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4049089 c.238C>T p.R80C Chr.14: 100545005-100545005 on Build GRCh38
949315 c.196G>A p.A66T Chr.14: 100545047-100545047 on Build GRCh38
147841 c.243+8C>T p.? Chr.14: 100544992-100544992 on Build GRCh38
5684531 c.216G>C p.Q72H Chr.14: 100545027-100545027 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768599874 Chr.14: 100545099-100545099 on Build GRCh38
rs745534996 Chr.14: 100545050-100545050 on Build GRCh38
rs1031970065 Chr.14: 100545148-100545148 on Build GRCh38
rs4074037 Chr.14: 100544872-100544872 on Build GRCh38
rs369042883 Chr.14: 100544991-100544991 on Build GRCh38
rs746464797 Chr.14: 100545047-100545047 on Build GRCh38
rs769354786 Chr.14: 100545294-100545294 on Build GRCh38
rs201229779 Chr.14: 100544987-100544987 on Build GRCh38
rs992595921 Chr.14: 100544930-100544930 on Build GRCh38
rs959742709 Chr.14: 100545200-100545200 on Build GRCh38
rs747505403 Chr.14: 100545182-100545182 on Build GRCh38
rs977106699 Chr.14: 100544894-100544894 on Build GRCh38
rs969763046 Chr.14: 100545270-100545270 on Build GRCh38
rs766441419 Chr.14: 100545115-100545115 on Build GRCh38
rs377175303 Chr.14: 100545021-100545021 on Build GRCh38
rs571419037 Chr.14: 100545048-100545048 on Build GRCh38
rs574924214 Chr.14: 100545312-100545312 on Build GRCh38
rs376786194 Chr.14: 100545160-100545160 on Build GRCh38
rs996648296 Chr.14: 100544867-100544867 on Build GRCh38
rs949180619 Chr.14: 100544842-100544842 on Build GRCh38
rs968618443 Chr.14: 100544892-100544892 on Build GRCh38
rs567389117 Chr.14: 100544969-100544969 on Build GRCh38
rs893466112 Chr.14: 100544947-100544947 on Build GRCh38
rs769393283 Chr.14: 100545054-100545054 on Build GRCh38
rs1046162160 Chr.14: 100544871-100544871 on Build GRCh38
rs375029308 Chr.14: 100544936-100544936 on Build GRCh38
rs776237670 Chr.14: 100545058-100545058 on Build GRCh38
rs746519830 Chr.14: 100544968-100544968 on Build GRCh38
rs998792125 Chr.14: 100545136-100545136 on Build GRCh38
rs745562081 Chr.14: 100544971-100544971 on Build GRCh38
rs980903953 Chr.14: 100544946-100544946 on Build GRCh38
rs761788351 Chr.14: 100545103-100545103 on Build GRCh38
rs368404489 Chr.14: 100544981-100544981 on Build GRCh38
rs778451982 Chr.14: 100545039-100545039 on Build GRCh38
rs181022257 Chr.14: 100545117-100545117 on Build GRCh38
rs368675167 Chr.14: 100544952-100544952 on Build GRCh38
rs373225306 Chr.14: 100545131-100545131 on Build GRCh38
rs375154513 Chr.14: 100544970-100544970 on Build GRCh38
rs959953524 Chr.14: 100544908-100544908 on Build GRCh38
rs370438499 Chr.14: 100545130-100545130 on Build GRCh38
rs754038666 Chr.14: 100544992-100544992 on Build GRCh38
rs1053539684 Chr.14: 100545140-100545140 on Build GRCh38
rs774294422 Chr.14: 100545100-100545100 on Build GRCh38
rs367672777 Chr.14: 100544939-100544939 on Build GRCh38
rs763954358 Chr.14: 100544986-100544986 on Build GRCh38
rs147714240 Chr.14: 100544951-100544951 on Build GRCh38
rs765621552 Chr.14: 100545005-100545005 on Build GRCh38
rs748684822 Chr.14: 100544953-100544953 on Build GRCh38
rs948282522 Chr.14: 100544945-100544946 on Build GRCh38
rs560917461 Chr.14: 100545216-100545216 on Build GRCh38
rs1017707223 Chr.14: 100544974-100544974 on Build GRCh38
rs145605251 Chr.14: 100544940-100544940 on Build GRCh38
rs769603490 Chr.14: 100544978-100544978 on Build GRCh38
rs1031319884 Chr.14: 100544900-100544900 on Build GRCh38
rs79064001 Chr.14: 100544982-100544982 on Build GRCh38
rs777278140 Chr.14: 100544967-100544967 on Build GRCh38
rs1042675646 Chr.14: 100545298-100545298 on Build GRCh38
rs569488036 Chr.14: 100544880-100544880 on Build GRCh38
rs114368829 Chr.14: 100545004-100545004 on Build GRCh38
rs1022753305 Chr.14: 100545262-100545262 on Build GRCh38
rs753014841 Chr.14: 100545017-100545017 on Build GRCh38
rs372358446 Chr.14: 100544958-100544958 on Build GRCh38
rs369884208 Chr.14: 100545097-100545097 on Build GRCh38
rs775300513 Chr.14: 100545076-100545076 on Build GRCh38

View Full Product Details