Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 27519840-27520345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRG3
Entrez Gene ID
2567
Gene Name
gamma-aminobutyric acid type A receptor gamma3 subunit
Gene Aliases
-
UniGene
Hs.569475
Gene Chromosome Location
Chr.15: 26971282-27537112 on Build GRCh38
Transcripts
NM_033223.4 XM_011521430.2 XM_011521431.2 XM_017022058.1 XM_017022059.1 XM_017022060.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2083089 c.721G>A p.V241I Chr.15: 27519980-27519980 on Build GRCh38
2083088 c.720T>A p.Y240* Chr.15: 27519979-27519979 on Build GRCh38
2083086 c.713-2A>T p.? Chr.15: 27519970-27519970 on Build GRCh38
1253008 c.806T>G p.V269G Chr.15: 27520065-27520065 on Build GRCh38
5733620 c.724G>A p.V242I Chr.15: 27519983-27519983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777606546 Chr.15: 27520064-27520064 on Build GRCh38
rs774709562 Chr.15: 27520146-27520146 on Build GRCh38
rs1041251727 Chr.15: 27520310-27520310 on Build GRCh38
rs1015241882 Chr.15: 27520055-27520055 on Build GRCh38
rs756430332 Chr.15: 27520283-27520283 on Build GRCh38
rs977957925 Chr.15: 27519990-27519990 on Build GRCh38
rs932450125 Chr.15: 27519866-27519866 on Build GRCh38
rs960250614 Chr.15: 27520138-27520138 on Build GRCh38
rs753820210 Chr.15: 27520031-27520031 on Build GRCh38
rs1048461661 Chr.15: 27520240-27520240 on Build GRCh38
rs966520469 Chr.15: 27519979-27519979 on Build GRCh38
rs769220987 Chr.15: 27520135-27520135 on Build GRCh38
rs34130136 Chr.15: 27520009-27520009 on Build GRCh38
rs766467935 Chr.15: 27520027-27520027 on Build GRCh38
rs765021902 Chr.15: 27520052-27520052 on Build GRCh38
rs370040504 Chr.15: 27519921-27519921 on Build GRCh38
rs754947412 Chr.15: 27520043-27520043 on Build GRCh38
rs1021119878 Chr.15: 27519903-27519903 on Build GRCh38
rs754228838 Chr.15: 27520017-27520017 on Build GRCh38
rs773420079 Chr.15: 27519966-27519966 on Build GRCh38
rs768684484 Chr.15: 27519870-27519871 on Build GRCh38
rs759961930 Chr.15: 27520091-27520091 on Build GRCh38
rs369801805 Chr.15: 27519901-27519901 on Build GRCh38
rs531803801 Chr.15: 27519875-27519875 on Build GRCh38
rs940124515 Chr.15: 27520284-27520284 on Build GRCh38
rs567628395 Chr.15: 27520180-27520180 on Build GRCh38
rs767425276 Chr.15: 27520173-27520173 on Build GRCh38
rs536777524 Chr.15: 27520229-27520229 on Build GRCh38
rs756924937 Chr.15: 27520105-27520105 on Build GRCh38
rs371238776 Chr.15: 27520158-27520158 on Build GRCh38
rs1003302264 Chr.15: 27520190-27520190 on Build GRCh38
rs752592069 Chr.15: 27520054-27520054 on Build GRCh38
rs12905441 Chr.15: 27520267-27520267 on Build GRCh38
rs547519737 Chr.15: 27520174-27520174 on Build GRCh38
rs780884337 Chr.15: 27520108-27520108 on Build GRCh38
rs752863668 Chr.15: 27520137-27520143 on Build GRCh38
rs746640486 Chr.15: 27520096-27520096 on Build GRCh38
rs757068573 Chr.15: 27519883-27519883 on Build GRCh38
rs758260967 Chr.15: 27520057-27520057 on Build GRCh38
rs376673418 Chr.15: 27520157-27520157 on Build GRCh38
rs76364643 Chr.15: 27520020-27520020 on Build GRCh38
rs888358146 Chr.15: 27519975-27519975 on Build GRCh38
rs767786656 Chr.15: 27519940-27519940 on Build GRCh38
rs745309801 Chr.15: 27520131-27520131 on Build GRCh38
rs192833610 Chr.15: 27520290-27520290 on Build GRCh38
rs187966860 Chr.15: 27520114-27520114 on Build GRCh38
rs767932957 Chr.15: 27520134-27520135 on Build GRCh38
rs748615417 Chr.15: 27520151-27520151 on Build GRCh38

View Full Product Details