Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 27527326-27527826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRG3
Entrez Gene ID
2567
Gene Name
gamma-aminobutyric acid type A receptor gamma3 subunit
Gene Aliases
-
UniGene
Hs.569475
Gene Chromosome Location
Chr.15: 26971282-27537112 on Build GRCh38
Transcripts
NM_033223.4 XM_011521430.2 XM_011521431.2 XM_017022058.1 XM_017022059.1 XM_017022060.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2083098 c.989C>A p.A330E Chr.15: 27527556-27527556 on Build GRCh38
361789 c.1022C>A p.S341Y Chr.15: 27527589-27527589 on Build GRCh38
7296395 c.875C>T p.T292M Chr.15: 27527442-27527442 on Build GRCh38
6668688 c.951C>A p.D317E Chr.15: 27527518-27527518 on Build GRCh38
1301006 c.1028G>A p.C343Y Chr.15: 27527595-27527595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373976015 Chr.15: 27527637-27527637 on Build GRCh38
rs75302071 Chr.15: 27527418-27527418 on Build GRCh38
rs372451105 Chr.15: 27527628-27527628 on Build GRCh38
rs748622911 Chr.15: 27527473-27527473 on Build GRCh38
rs764060171 Chr.15: 27527509-27527509 on Build GRCh38
rs377256790 Chr.15: 27527401-27527401 on Build GRCh38
rs556260076 Chr.15: 27527689-27527689 on Build GRCh38
rs767268527 Chr.15: 27527517-27527517 on Build GRCh38
rs372739989 Chr.15: 27527385-27527385 on Build GRCh38
rs774426380 Chr.15: 27527607-27527607 on Build GRCh38
rs755576470 Chr.15: 27527444-27527444 on Build GRCh38
rs939058248 Chr.15: 27527614-27527616 on Build GRCh38
rs781522661 Chr.15: 27527566-27527566 on Build GRCh38
rs202145457 Chr.15: 27527390-27527390 on Build GRCh38
rs775440135 Chr.15: 27527596-27527596 on Build GRCh38
rs373831797 Chr.15: 27527420-27527420 on Build GRCh38
rs370118937 Chr.15: 27527419-27527419 on Build GRCh38
rs768854851 Chr.15: 27527352-27527352 on Build GRCh38
rs543791419 Chr.15: 27527657-27527657 on Build GRCh38
rs984023036 Chr.15: 27527484-27527484 on Build GRCh38
rs2066712 Chr.15: 27527621-27527621 on Build GRCh38
rs779459678 Chr.15: 27527551-27527551 on Build GRCh38
rs922558081 Chr.15: 27527368-27527368 on Build GRCh38
rs576009445 Chr.15: 27527379-27527379 on Build GRCh38
rs932582088 Chr.15: 27527768-27527768 on Build GRCh38
rs202042920 Chr.15: 27527648-27527648 on Build GRCh38
rs1031707892 Chr.15: 27527366-27527366 on Build GRCh38
rs772138833 Chr.15: 27527554-27527554 on Build GRCh38
rs879645590 Chr.15: 27527527-27527527 on Build GRCh38
rs771612089 Chr.15: 27527656-27527656 on Build GRCh38
rs932303966 Chr.15: 27527373-27527373 on Build GRCh38
rs759698238 Chr.15: 27527489-27527489 on Build GRCh38
rs750009355 Chr.15: 27527442-27527442 on Build GRCh38
rs747315792 Chr.15: 27527479-27527479 on Build GRCh38
rs778114963 Chr.15: 27527476-27527476 on Build GRCh38
rs77630664 Chr.15: 27527439-27527439 on Build GRCh38
rs1021875140 Chr.15: 27527693-27527693 on Build GRCh38
rs781443644 Chr.15: 27527672-27527672 on Build GRCh38
rs775592973 Chr.15: 27527494-27527494 on Build GRCh38
rs755740949 Chr.15: 27527540-27527540 on Build GRCh38
rs768772490 Chr.15: 27527606-27527606 on Build GRCh38
rs1011087107 Chr.15: 27527395-27527395 on Build GRCh38
rs768139197 Chr.15: 27527683-27527683 on Build GRCh38
rs769737722 Chr.15: 27527491-27527491 on Build GRCh38
rs767195771 Chr.15: 27527611-27527611 on Build GRCh38
rs746102581 Chr.15: 27527578-27527578 on Build GRCh38
rs757731018 Chr.15: 27527667-27527667 on Build GRCh38
rs760910630 Chr.15: 27527621-27527623 on Build GRCh38
rs2303879 Chr.15: 27527746-27527746 on Build GRCh38
rs764006643 Chr.15: 27527410-27527410 on Build GRCh38
rs752038522 Chr.15: 27527660-27527660 on Build GRCh38
rs111556911 Chr.15: 27527736-27527736 on Build GRCh38
rs758920161 Chr.15: 27527557-27527557 on Build GRCh38
rs749327046 Chr.15: 27527597-27527597 on Build GRCh38
rs771165582 Chr.15: 27527561-27527561 on Build GRCh38
rs78244225 Chr.15: 27527474-27527474 on Build GRCh38
rs747436394 Chr.15: 27527558-27527558 on Build GRCh38
rs763019442 Chr.15: 27527503-27527503 on Build GRCh38
rs373413606 Chr.15: 27527446-27527446 on Build GRCh38
rs961189302 Chr.15: 27527720-27527720 on Build GRCh38
rs185908450 Chr.15: 27527351-27527351 on Build GRCh38
rs866790239 Chr.15: 27527391-27527391 on Build GRCh38
rs773191845 Chr.15: 27527613-27527613 on Build GRCh38
rs761742427 Chr.15: 27527609-27527609 on Build GRCh38
rs561530336 Chr.15: 27527584-27527584 on Build GRCh38
rs140678 Chr.15: 27527359-27527359 on Build GRCh38
rs776710232 Chr.15: 27527483-27527483 on Build GRCh38
rs751413776 Chr.15: 27527510-27527510 on Build GRCh38
rs370950953 Chr.15: 27527512-27527512 on Build GRCh38
rs771195863 Chr.15: 27527481-27527481 on Build GRCh38
rs140679 Chr.15: 27527530-27527530 on Build GRCh38
rs80141809 Chr.15: 27527472-27527472 on Build GRCh38
rs780651677 Chr.15: 27527427-27527427 on Build GRCh38
rs759516910 Chr.15: 27527387-27527387 on Build GRCh38

View Full Product Details