Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34874550-34875067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQR
Entrez Gene ID
9716
Gene Name
aquarius intron-binding spliceosomal factor
Gene Aliases
IBP160, fSAP164
UniGene
Hs.510958
Gene Chromosome Location
Chr.15: 34856351-34969794 on Build GRCh38
Transcripts
NM_014691.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
700032 c.3365G>A p.R1122H Chr.15: 34874737-34874737 on Build GRCh38
50619 c.3376G>A p.V1126I Chr.15: 34874726-34874726 on Build GRCh38
1245274 c.3386C>T p.P1129L Chr.15: 34874716-34874716 on Build GRCh38
960902 c.3285C>T p.G1095G Chr.15: 34874817-34874817 on Build GRCh38
5789718 c.3238-1G>C p.? Chr.15: 34874865-34874865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200095121 Chr.15: 34874728-34874728 on Build GRCh38
rs752121140 Chr.15: 34874844-34874844 on Build GRCh38
rs776270242 Chr.15: 34874913-34874913 on Build GRCh38
rs1056403932 Chr.15: 34875003-34875003 on Build GRCh38
rs1038372647 Chr.15: 34874934-34874934 on Build GRCh38
rs954405926 Chr.15: 34875044-34875044 on Build GRCh38
rs763802425 Chr.15: 34874762-34874762 on Build GRCh38
rs75405831 Chr.15: 34874979-34874979 on Build GRCh38
rs773831067 Chr.15: 34874726-34874726 on Build GRCh38
rs975668238 Chr.15: 34874747-34874747 on Build GRCh38
rs775479403 Chr.15: 34874574-34874574 on Build GRCh38
rs773874652 Chr.15: 34874881-34874882 on Build GRCh38
rs748894591 Chr.15: 34874709-34874709 on Build GRCh38
rs187500930 Chr.15: 34874988-34874988 on Build GRCh38
rs562834652 Chr.15: 34874889-34874889 on Build GRCh38
rs747337340 Chr.15: 34874703-34874703 on Build GRCh38
rs565314234 Chr.15: 34874590-34874590 on Build GRCh38
rs1036764108 Chr.15: 34874920-34874920 on Build GRCh38
rs567085491 Chr.15: 34874995-34874995 on Build GRCh38
rs750587400 Chr.15: 34874671-34874671 on Build GRCh38
rs778857349 Chr.15: 34874665-34874665 on Build GRCh38
rs767853291 Chr.15: 34874782-34874782 on Build GRCh38
rs937974781 Chr.15: 34875001-34875001 on Build GRCh38
rs1021190405 Chr.15: 34875043-34875043 on Build GRCh38
rs1044590882 Chr.15: 34874869-34874869 on Build GRCh38
rs756167860 Chr.15: 34874900-34874900 on Build GRCh38
rs370712067 Chr.15: 34874653-34874653 on Build GRCh38
rs771588027 Chr.15: 34874898-34874898 on Build GRCh38
rs1007071422 Chr.15: 34875045-34875045 on Build GRCh38
rs780745443 Chr.15: 34874683-34874683 on Build GRCh38
rs767028312 Chr.15: 34874884-34874885 on Build GRCh38
rs373849838 Chr.15: 34874663-34874663 on Build GRCh38
rs201062230 Chr.15: 34874814-34874814 on Build GRCh38
rs140189633 Chr.15: 34874727-34874727 on Build GRCh38
rs767152106 Chr.15: 34874910-34874911 on Build GRCh38
rs375574768 Chr.15: 34874636-34874636 on Build GRCh38
rs978081785 Chr.15: 34874652-34874652 on Build GRCh38
rs776557531 Chr.15: 34874772-34874772 on Build GRCh38
rs758581492 Chr.15: 34874817-34874817 on Build GRCh38
rs965403789 Chr.15: 34874619-34874619 on Build GRCh38
rs941430582 Chr.15: 34874929-34874929 on Build GRCh38
rs905142337 Chr.15: 34874598-34874598 on Build GRCh38
rs896365947 Chr.15: 34875019-34875019 on Build GRCh38
rs531053905 Chr.15: 34874930-34874930 on Build GRCh38
rs781402890 Chr.15: 34874874-34874874 on Build GRCh38
rs755299180 Chr.15: 34874864-34874864 on Build GRCh38
rs775432905 Chr.15: 34874737-34874737 on Build GRCh38
rs745336539 Chr.15: 34874890-34874890 on Build GRCh38
rs768697955 Chr.15: 34874908-34874908 on Build GRCh38
rs751301330 Chr.15: 34874586-34874586 on Build GRCh38
rs1009792150 Chr.15: 34874655-34874655 on Build GRCh38
rs376843690 Chr.15: 34874648-34874648 on Build GRCh38
rs754268525 Chr.15: 34875038-34875038 on Build GRCh38
rs368010115 Chr.15: 34874708-34874708 on Build GRCh38
rs995573268 Chr.15: 34874994-34874994 on Build GRCh38
rs768921138 Chr.15: 34874706-34874706 on Build GRCh38
rs759126659 Chr.15: 34874882-34874883 on Build GRCh38
rs934192507 Chr.15: 34874875-34874875 on Build GRCh38
rs760461836 Chr.15: 34874738-34874738 on Build GRCh38
rs1002073125 Chr.15: 34874618-34874618 on Build GRCh38
rs748601329 Chr.15: 34874881-34874881 on Build GRCh38
rs184526206 Chr.15: 34874662-34874662 on Build GRCh38
rs765603694 Chr.15: 34874800-34874800 on Build GRCh38
rs775339056 Chr.15: 34874901-34874901 on Build GRCh38
rs756831327 Chr.15: 34874885-34874885 on Build GRCh38
rs758948708 Chr.15: 34874679-34874679 on Build GRCh38
rs368592487 Chr.15: 34874796-34874796 on Build GRCh38
rs371226184 Chr.15: 34874715-34874715 on Build GRCh38
rs781455615 Chr.15: 34874767-34874767 on Build GRCh38
rs756949703 Chr.15: 34874654-34874655 on Build GRCh38

View Full Product Details