Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49485532-49486070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM227B
Entrez Gene ID
196951
Gene Name
family with sequence similarity 227 member B
Gene Aliases
C15orf33
UniGene
Hs.712959
Gene Chromosome Location
Chr.15: 49326997-49620931 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521321.2 XM_011521322.1 XM_011521324.1 XM_011521325.2 XM_011521326.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1 NM_002009.3

Gene Symbol
FGF7
Entrez Gene ID
2252
Gene Name
fibroblast growth factor 7
Gene Aliases
HBGF-7, KGF
UniGene
Hs.567268
Gene Chromosome Location
Chr.15: 49423178-49487326 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521321.2 XM_011521322.1 XM_011521324.1 XM_011521325.2 XM_011521326.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1 NM_002009.3

SNP Detail

SNP ID Chromosome Location
rs1057669 Chr.15: 49485947-49485947 on Build GRCh38
rs980499556 Chr.15: 49485558-49485558 on Build GRCh38
rs560983567 Chr.15: 49486032-49486032 on Build GRCh38
rs530204047 Chr.15: 49485556-49485556 on Build GRCh38
rs531547965 Chr.15: 49485689-49485689 on Build GRCh38
rs931685557 Chr.15: 49486001-49486001 on Build GRCh38
rs1057670 Chr.15: 49485954-49485954 on Build GRCh38
rs913505675 Chr.15: 49485915-49485915 on Build GRCh38
rs936255662 Chr.15: 49485583-49485583 on Build GRCh38
rs548633926 Chr.15: 49485589-49485589 on Build GRCh38
rs373173503 Chr.15: 49485696-49485696 on Build GRCh38
rs908847850 Chr.15: 49486022-49486022 on Build GRCh38
rs913369424 Chr.15: 49485617-49485617 on Build GRCh38
rs535938973 Chr.15: 49485875-49485875 on Build GRCh38
rs976648498 Chr.15: 49485962-49485962 on Build GRCh38
rs3985843 Chr.15: 49485860-49485860 on Build GRCh38
rs188586834 Chr.15: 49485669-49485669 on Build GRCh38
rs1064338 Chr.15: 49485568-49485568 on Build GRCh38
rs3985845 Chr.15: 49485937-49485937 on Build GRCh38
rs1001668552 Chr.15: 49485798-49485798 on Build GRCh38
rs944889885 Chr.15: 49485628-49485628 on Build GRCh38
rs926011788 Chr.15: 49485574-49485574 on Build GRCh38
rs1033387883 Chr.15: 49485816-49485816 on Build GRCh38
rs183756896 Chr.15: 49485936-49485936 on Build GRCh38
rs773596769 Chr.15: 49485797-49485797 on Build GRCh38
rs183166589 Chr.15: 49485552-49485552 on Build GRCh38
rs1004767045 Chr.15: 49485683-49485683 on Build GRCh38
rs879226752 Chr.15: 49485586-49485586 on Build GRCh38
rs547173407 Chr.15: 49485795-49485795 on Build GRCh38
rs3985844 Chr.15: 49485890-49485890 on Build GRCh38
rs3959665 Chr.15: 49486019-49486019 on Build GRCh38
rs988994989 Chr.15: 49485910-49485910 on Build GRCh38
rs1048809618 Chr.15: 49486021-49486021 on Build GRCh38
rs931784888 Chr.15: 49485673-49485673 on Build GRCh38
rs900235410 Chr.15: 49485660-49485660 on Build GRCh38
rs1040570030 Chr.15: 49485647-49485647 on Build GRCh38
rs989024807 Chr.15: 49485611-49485611 on Build GRCh38
rs887442124 Chr.15: 49485682-49485682 on Build GRCh38
rs1013820560 Chr.15: 49485761-49485761 on Build GRCh38
rs12440028 Chr.15: 49485703-49485703 on Build GRCh38
rs192954168 Chr.15: 49485771-49485771 on Build GRCh38
rs922162789 Chr.15: 49485988-49485988 on Build GRCh38

View Full Product Details