Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59107198-59107738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB2
Entrez Gene ID
9133
Gene Name
cyclin B2
Gene Aliases
HsT17299
UniGene
Hs.194698
Gene Chromosome Location
Chr.15: 59105085-59125045 on Build GRCh38
Transcripts
NM_004701.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5798152 c.94C>T p.R32* Chr.15: 59107391-59107391 on Build GRCh38
7415718 c.33T>G p.S11R Chr.15: 59107330-59107330 on Build GRCh38
6622926 c.228T>C p.A76A Chr.15: 59107631-59107631 on Build GRCh38
1247604 c.86C>T p.T29I Chr.15: 59107383-59107383 on Build GRCh38
963415 c.95G>A p.R32Q Chr.15: 59107392-59107392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773159638 Chr.15: 59107569-59107569 on Build GRCh38
rs189971570 Chr.15: 59107554-59107554 on Build GRCh38
rs35033821 Chr.15: 59107670-59107670 on Build GRCh38
rs866249126 Chr.15: 59107588-59107588 on Build GRCh38
rs201973541 Chr.15: 59107711-59107711 on Build GRCh38
rs150117081 Chr.15: 59107433-59107433 on Build GRCh38
rs771676922 Chr.15: 59107323-59107323 on Build GRCh38
rs749059876 Chr.15: 59107571-59107571 on Build GRCh38
rs755571015 Chr.15: 59107679-59107679 on Build GRCh38
rs770697460 Chr.15: 59107695-59107695 on Build GRCh38
rs199782001 Chr.15: 59107631-59107631 on Build GRCh38
rs745745222 Chr.15: 59107488-59107488 on Build GRCh38
rs752722691 Chr.15: 59107391-59107391 on Build GRCh38
rs776514242 Chr.15: 59107353-59107353 on Build GRCh38
rs769001548 Chr.15: 59107707-59107707 on Build GRCh38
rs34747071 Chr.15: 59107643-59107643 on Build GRCh38
rs757230414 Chr.15: 59107455-59107455 on Build GRCh38
rs764998483 Chr.15: 59107363-59107363 on Build GRCh38
rs144091405 Chr.15: 59107633-59107633 on Build GRCh38
rs761620661 Chr.15: 59107355-59107355 on Build GRCh38
rs532667545 Chr.15: 59107305-59107306 on Build GRCh38
rs767576210 Chr.15: 59107588-59107590 on Build GRCh38
rs146926787 Chr.15: 59107597-59107597 on Build GRCh38
rs369986119 Chr.15: 59107678-59107678 on Build GRCh38
rs138232014 Chr.15: 59107603-59107603 on Build GRCh38
rs778357585 Chr.15: 59107301-59107301 on Build GRCh38
rs772878851 Chr.15: 59107638-59107638 on Build GRCh38
rs777620010 Chr.15: 59107690-59107690 on Build GRCh38
rs757823735 Chr.15: 59107534-59107534 on Build GRCh38
rs779769445 Chr.15: 59107302-59107303 on Build GRCh38
rs1051832905 Chr.15: 59107543-59107543 on Build GRCh38
rs371888839 Chr.15: 59107439-59107439 on Build GRCh38
rs769644954 Chr.15: 59107494-59107494 on Build GRCh38
rs11293095 Chr.15: 59107303-59107303 on Build GRCh38
rs765713696 Chr.15: 59107536-59107536 on Build GRCh38
rs151299700 Chr.15: 59107378-59107378 on Build GRCh38
rs746535929 Chr.15: 59107485-59107485 on Build GRCh38
rs150253971 Chr.15: 59107604-59107604 on Build GRCh38
rs746653521 Chr.15: 59107548-59107548 on Build GRCh38
rs11548100 Chr.15: 59107586-59107586 on Build GRCh38
rs552798983 Chr.15: 59107303-59107305 on Build GRCh38
rs767695563 Chr.15: 59107380-59107380 on Build GRCh38
rs374923143 Chr.15: 59107510-59107510 on Build GRCh38
rs773075874 Chr.15: 59107531-59107531 on Build GRCh38
rs775731943 Chr.15: 59107517-59107517 on Build GRCh38
rs559386008 Chr.15: 59107546-59107546 on Build GRCh38
rs563802128 Chr.15: 59107285-59107285 on Build GRCh38
rs750754556 Chr.15: 59107537-59107537 on Build GRCh38
rs539408437 Chr.15: 59107552-59107552 on Build GRCh38
rs549586410 Chr.15: 59107322-59107322 on Build GRCh38
rs373482453 Chr.15: 59107697-59107697 on Build GRCh38
rs530327716 Chr.15: 59107305-59107305 on Build GRCh38
rs571854426 Chr.15: 59107492-59107492 on Build GRCh38
rs745596937 Chr.15: 59107703-59107703 on Build GRCh38
rs983351370 Chr.15: 59107361-59107361 on Build GRCh38
rs200529135 Chr.15: 59107611-59107611 on Build GRCh38
rs1021675271 Chr.15: 59107596-59107596 on Build GRCh38
rs921961093 Chr.15: 59107509-59107509 on Build GRCh38
rs760169940 Chr.15: 59107340-59107340 on Build GRCh38
rs398027505 Chr.15: 59107318-59107318 on Build GRCh38
rs547223062 Chr.15: 59107435-59107435 on Build GRCh38
rs769581400 Chr.15: 59107418-59107419 on Build GRCh38
rs751848817 Chr.15: 59107367-59107367 on Build GRCh38
rs768589001 Chr.15: 59107350-59107350 on Build GRCh38
rs765697847 Chr.15: 59107610-59107610 on Build GRCh38
rs749218830 Chr.15: 59107694-59107694 on Build GRCh38
rs779600368 Chr.15: 59107544-59107544 on Build GRCh38
rs531056842 Chr.15: 59107318-59107318 on Build GRCh38
rs775613648 Chr.15: 59107589-59107589 on Build GRCh38
rs140527154 Chr.15: 59107431-59107431 on Build GRCh38
rs367803778 Chr.15: 59107413-59107413 on Build GRCh38
rs761914242 Chr.15: 59107605-59107605 on Build GRCh38
rs201051714 Chr.15: 59107642-59107642 on Build GRCh38
rs760885492 Chr.15: 59107521-59107521 on Build GRCh38
rs138427426 Chr.15: 59107560-59107560 on Build GRCh38
rs757996150 Chr.15: 59107457-59107457 on Build GRCh38
rs781139213 Chr.15: 59107487-59107487 on Build GRCh38
rs369361404 Chr.15: 59107529-59107529 on Build GRCh38
rs771370966 Chr.15: 59107319-59107319 on Build GRCh38
rs928445072 Chr.15: 59107224-59107224 on Build GRCh38
rs762752818 Chr.15: 59107515-59107515 on Build GRCh38
rs869228566 Chr.15: 59107398-59107398 on Build GRCh38
rs767663609 Chr.15: 59107516-59107516 on Build GRCh38
rs779694436 Chr.15: 59107469-59107469 on Build GRCh38
rs575295531 Chr.15: 59107304-59107305 on Build GRCh38
rs754613732 Chr.15: 59107655-59107655 on Build GRCh38
rs11548098 Chr.15: 59107626-59107626 on Build GRCh38
rs778445498 Chr.15: 59107425-59107425 on Build GRCh38
rs747932938 Chr.15: 59107318-59107319 on Build GRCh38
rs762932691 Chr.15: 59107536-59107536 on Build GRCh38
rs757924075 Chr.15: 59107303-59107304 on Build GRCh38
rs754459555 Chr.15: 59107531-59107531 on Build GRCh38

View Full Product Details