Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59652124-59652638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2A2
Entrez Gene ID
2958
Gene Name
general transcription factor IIA subunit 2
Gene Aliases
HsT18745, T18745, TF2A2, TFIIA, TFIIA-12, TFIIA-gamma, TFIIAS
UniGene
Hs.512934
Gene Chromosome Location
Chr.15: 59638062-59657541 on Build GRCh38
Transcripts
NM_001320929.1 NM_001320930.1 NM_004492.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6077601 c.46C>T p.Q16* Chr.15: 59652232-59652232 on Build GRCh38
470901 c.56T>C p.L19P Chr.15: 59652222-59652222 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs894571418 Chr.15: 59652467-59652467 on Build GRCh38
rs377633920 Chr.15: 59652180-59652180 on Build GRCh38
rs563643148 Chr.15: 59652396-59652396 on Build GRCh38
rs964135643 Chr.15: 59652536-59652536 on Build GRCh38
rs117916609 Chr.15: 59652551-59652551 on Build GRCh38
rs551380443 Chr.15: 59652319-59652319 on Build GRCh38
rs776585142 Chr.15: 59652286-59652289 on Build GRCh38
rs774641060 Chr.15: 59652214-59652214 on Build GRCh38
rs535139879 Chr.15: 59652431-59652431 on Build GRCh38
rs746973189 Chr.15: 59652182-59652182 on Build GRCh38
rs977632342 Chr.15: 59652559-59652559 on Build GRCh38
rs549205369 Chr.15: 59652415-59652415 on Build GRCh38
rs746931307 Chr.15: 59652320-59652320 on Build GRCh38
rs1012045020 Chr.15: 59652471-59652471 on Build GRCh38
rs915965176 Chr.15: 59652584-59652584 on Build GRCh38
rs1041983288 Chr.15: 59652428-59652428 on Build GRCh38
rs759897543 Chr.15: 59652224-59652224 on Build GRCh38
rs952344247 Chr.15: 59652521-59652521 on Build GRCh38
rs147604787 Chr.15: 59652545-59652545 on Build GRCh38
rs776041166 Chr.15: 59652185-59652185 on Build GRCh38
rs183933857 Chr.15: 59652418-59652418 on Build GRCh38
rs750744900 Chr.15: 59652307-59652307 on Build GRCh38
rs1017999747 Chr.15: 59652526-59652526 on Build GRCh38
rs574535054 Chr.15: 59652488-59652488 on Build GRCh38
rs745676646 Chr.15: 59652164-59652164 on Build GRCh38
rs187761824 Chr.15: 59652525-59652525 on Build GRCh38
rs983868677 Chr.15: 59652324-59652324 on Build GRCh38
rs191658198 Chr.15: 59652306-59652306 on Build GRCh38
rs970636378 Chr.15: 59652155-59652155 on Build GRCh38
rs776042641 Chr.15: 59652255-59652255 on Build GRCh38
rs764195352 Chr.15: 59652279-59652279 on Build GRCh38
rs1055858005 Chr.15: 59652435-59652435 on Build GRCh38
rs767830055 Chr.15: 59652230-59652230 on Build GRCh38
rs752456542 Chr.15: 59652282-59652282 on Build GRCh38
rs375520518 Chr.15: 59652401-59652401 on Build GRCh38
rs762957242 Chr.15: 59652287-59652287 on Build GRCh38
rs757512686 Chr.15: 59652362-59652362 on Build GRCh38
rs781544325 Chr.15: 59652384-59652384 on Build GRCh38
rs749748672 Chr.15: 59652506-59652506 on Build GRCh38
rs547152449 Chr.15: 59652497-59652497 on Build GRCh38
rs922816248 Chr.15: 59652414-59652414 on Build GRCh38
rs191111333 Chr.15: 59652561-59652561 on Build GRCh38
rs571530006 Chr.15: 59652524-59652524 on Build GRCh38
rs530685593 Chr.15: 59652409-59652409 on Build GRCh38
rs757498406 Chr.15: 59652291-59652291 on Build GRCh38
rs540587331 Chr.15: 59652150-59652150 on Build GRCh38
rs559066872 Chr.15: 59652184-59652184 on Build GRCh38
rs1029775261 Chr.15: 59652548-59652548 on Build GRCh38
rs371091075 Chr.15: 59652221-59652221 on Build GRCh38
rs747757229 Chr.15: 59652192-59652192 on Build GRCh38
rs910990888 Chr.15: 59652351-59652351 on Build GRCh38
rs565792683 Chr.15: 59652449-59652451 on Build GRCh38
rs994083148 Chr.15: 59652505-59652505 on Build GRCh38
rs936030917 Chr.15: 59652432-59652432 on Build GRCh38
rs758348866 Chr.15: 59652167-59652167 on Build GRCh38
rs755755195 Chr.15: 59652563-59652563 on Build GRCh38
rs761128776 Chr.15: 59652266-59652266 on Build GRCh38
rs766463653 Chr.15: 59652294-59652297 on Build GRCh38
rs769457557 Chr.15: 59652206-59652206 on Build GRCh38
rs758194892 Chr.15: 59652316-59652316 on Build GRCh38
rs935994203 Chr.15: 59652576-59652576 on Build GRCh38
rs754900302 Chr.15: 59652327-59652327 on Build GRCh38
rs748477274 Chr.15: 59652580-59652580 on Build GRCh38

View Full Product Details