Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59657315-59657821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2A2
Entrez Gene ID
2958
Gene Name
general transcription factor IIA subunit 2
Gene Aliases
HsT18745, T18745, TF2A2, TFIIA, TFIIA-12, TFIIA-gamma, TFIIAS
UniGene
Hs.512934
Gene Chromosome Location
Chr.15: 59638062-59657541 on Build GRCh38
Transcripts
NM_001320930.1 NM_004492.2

SNP Detail

SNP ID Chromosome Location
rs561484260 Chr.15: 59657748-59657748 on Build GRCh38
rs970876409 Chr.15: 59657560-59657560 on Build GRCh38
rs535058445 Chr.15: 59657676-59657676 on Build GRCh38
rs936501939 Chr.15: 59657673-59657673 on Build GRCh38
rs1024748847 Chr.15: 59657445-59657445 on Build GRCh38
rs573513752 Chr.15: 59657796-59657796 on Build GRCh38
rs369082085 Chr.15: 59657423-59657423 on Build GRCh38
rs1026217088 Chr.15: 59657572-59657572 on Build GRCh38
rs543107858 Chr.15: 59657738-59657738 on Build GRCh38
rs12595090 Chr.15: 59657344-59657344 on Build GRCh38
rs7164756 Chr.15: 59657426-59657426 on Build GRCh38
rs529547893 Chr.15: 59657348-59657348 on Build GRCh38
rs763168222 Chr.15: 59657362-59657362 on Build GRCh38
rs1041083650 Chr.15: 59657480-59657480 on Build GRCh38
rs551622645 Chr.15: 59657520-59657520 on Build GRCh38
rs922660131 Chr.15: 59657627-59657627 on Build GRCh38
rs570128428 Chr.15: 59657540-59657540 on Build GRCh38
rs942662900 Chr.15: 59657598-59657598 on Build GRCh38
rs892950128 Chr.15: 59657484-59657484 on Build GRCh38
rs755094646 Chr.15: 59657457-59657457 on Build GRCh38
rs568648660 Chr.15: 59657345-59657345 on Build GRCh38
rs894783992 Chr.15: 59657532-59657532 on Build GRCh38
rs538832348 Chr.15: 59657712-59657712 on Build GRCh38
rs990554397 Chr.15: 59657427-59657427 on Build GRCh38
rs189390820 Chr.15: 59657491-59657491 on Build GRCh38
rs952941038 Chr.15: 59657574-59657574 on Build GRCh38
rs184987041 Chr.15: 59657724-59657724 on Build GRCh38
rs7163436 Chr.15: 59657571-59657571 on Build GRCh38
rs902383942 Chr.15: 59657502-59657502 on Build GRCh38
rs977008801 Chr.15: 59657792-59657792 on Build GRCh38
rs547363603 Chr.15: 59657475-59657475 on Build GRCh38
rs577842195 Chr.15: 59657773-59657773 on Build GRCh38
rs1032288924 Chr.15: 59657521-59657521 on Build GRCh38
rs1046094092 Chr.15: 59657692-59657692 on Build GRCh38
rs192633005 Chr.15: 59657552-59657552 on Build GRCh38
rs578214813 Chr.15: 59657706-59657706 on Build GRCh38
rs984363638 Chr.15: 59657466-59657466 on Build GRCh38
rs1000821168 Chr.15: 59657413-59657413 on Build GRCh38
rs997774928 Chr.15: 59657361-59657361 on Build GRCh38
rs1871500 Chr.15: 59657691-59657691 on Build GRCh38
rs1006913853 Chr.15: 59657352-59657352 on Build GRCh38
rs1001172983 Chr.15: 59657508-59657508 on Build GRCh38
rs938100191 Chr.15: 59657422-59657422 on Build GRCh38
rs1041828328 Chr.15: 59657650-59657650 on Build GRCh38
rs970882611 Chr.15: 59657448-59657448 on Build GRCh38
rs895073485 Chr.15: 59657684-59657684 on Build GRCh38
rs931556568 Chr.15: 59657460-59657460 on Build GRCh38
rs973319502 Chr.15: 59657452-59657452 on Build GRCh38
rs761112832 Chr.15: 59657695-59657695 on Build GRCh38
rs976727615 Chr.15: 59657620-59657620 on Build GRCh38
rs1041141753 Chr.15: 59657359-59657359 on Build GRCh38
rs911210972 Chr.15: 59657469-59657469 on Build GRCh38
rs564709720 Chr.15: 59657705-59657705 on Build GRCh38
rs984309809 Chr.15: 59657577-59657577 on Build GRCh38
rs1018446065 Chr.15: 59657723-59657723 on Build GRCh38
rs901657003 Chr.15: 59657360-59657360 on Build GRCh38
rs138274345 Chr.15: 59657700-59657700 on Build GRCh38
rs902433744 Chr.15: 59657384-59657384 on Build GRCh38
rs532936697 Chr.15: 59657493-59657493 on Build GRCh38
rs768039896 Chr.15: 59657652-59657652 on Build GRCh38
rs567662197 Chr.15: 59657588-59657588 on Build GRCh38
rs557150410 Chr.15: 59657721-59657721 on Build GRCh38
rs1042604082 Chr.15: 59657496-59657496 on Build GRCh38
rs946118822 Chr.15: 59657642-59657642 on Build GRCh38

View Full Product Details