Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63611868-63612400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HERC1
Entrez Gene ID
8925
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase family member 1
Gene Aliases
p532, p619
UniGene
Hs.210385
Gene Chromosome Location
Chr.15: 63608618-63833964 on Build GRCh38
Transcripts
NM_003922.3 XM_017022699.1 XM_017022700.1 XM_017022701.1 XM_017022702.1 XM_017022703.1 XM_017022704.1 XM_017022705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
963697 c.14339C>T p.S4780F Chr.15: 63612312-63612312 on Build GRCh38
6522164 c.14392G>T p.V4798F Chr.15: 63612259-63612259 on Build GRCh38
7281664 c.14400+4A>G p.? Chr.15: 63612247-63612247 on Build GRCh38
3502969 c.14341C>T p.R4781* Chr.15: 63612310-63612310 on Build GRCh38
275453 c.14397T>C p.D4799D Chr.15: 63612254-63612254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992794855 Chr.15: 63612169-63612169 on Build GRCh38
rs898767416 Chr.15: 63612213-63612213 on Build GRCh38
rs995560310 Chr.15: 63612002-63612002 on Build GRCh38
rs1003507637 Chr.15: 63611909-63611909 on Build GRCh38
rs772222697 Chr.15: 63612214-63612214 on Build GRCh38
rs116238605 Chr.15: 63611953-63611953 on Build GRCh38
rs780185997 Chr.15: 63612373-63612373 on Build GRCh38
rs1042247806 Chr.15: 63612096-63612096 on Build GRCh38
rs1049948132 Chr.15: 63612013-63612013 on Build GRCh38
rs779946629 Chr.15: 63612215-63612215 on Build GRCh38
rs972885927 Chr.15: 63611943-63611943 on Build GRCh38
rs940010045 Chr.15: 63611958-63611958 on Build GRCh38
rs1020277403 Chr.15: 63612049-63612049 on Build GRCh38
rs751489074 Chr.15: 63612242-63612243 on Build GRCh38
rs955570921 Chr.15: 63612229-63612229 on Build GRCh38
rs762753239 Chr.15: 63612265-63612265 on Build GRCh38
rs974669400 Chr.15: 63612032-63612032 on Build GRCh38
rs938642957 Chr.15: 63611938-63611938 on Build GRCh38
rs756250611 Chr.15: 63612303-63612303 on Build GRCh38
rs914332466 Chr.15: 63611904-63611904 on Build GRCh38
rs765928968 Chr.15: 63612317-63612317 on Build GRCh38
rs940593686 Chr.15: 63612228-63612228 on Build GRCh38
rs199937922 Chr.15: 63612220-63612220 on Build GRCh38
rs529636769 Chr.15: 63612171-63612171 on Build GRCh38
rs146027764 Chr.15: 63612070-63612070 on Build GRCh38
rs62012818 Chr.15: 63612081-63612081 on Build GRCh38
rs369207929 Chr.15: 63612221-63612221 on Build GRCh38
rs536292522 Chr.15: 63612010-63612010 on Build GRCh38
rs968453463 Chr.15: 63612108-63612108 on Build GRCh38
rs889932632 Chr.15: 63612014-63612014 on Build GRCh38
rs73441919 Chr.15: 63611911-63611911 on Build GRCh38
rs950471229 Chr.15: 63611963-63611963 on Build GRCh38
rs987061533 Chr.15: 63612001-63612001 on Build GRCh38
rs571820514 Chr.15: 63612365-63612365 on Build GRCh38
rs538193188 Chr.15: 63612059-63612059 on Build GRCh38
rs902619537 Chr.15: 63611901-63611901 on Build GRCh38
rs12102137 Chr.15: 63611936-63611936 on Build GRCh38
rs752320738 Chr.15: 63612354-63612354 on Build GRCh38
rs188246355 Chr.15: 63612259-63612259 on Build GRCh38
rs1017631249 Chr.15: 63611933-63611933 on Build GRCh38
rs747124430 Chr.15: 63612374-63612374 on Build GRCh38
rs562482745 Chr.15: 63612135-63612135 on Build GRCh38
rs183466963 Chr.15: 63612120-63612120 on Build GRCh38
rs138360051 Chr.15: 63612344-63612344 on Build GRCh38
rs1057414207 Chr.15: 63611944-63611944 on Build GRCh38
rs769682318 Chr.15: 63612245-63612245 on Build GRCh38
rs767218768 Chr.15: 63612346-63612346 on Build GRCh38
rs373488853 Chr.15: 63612307-63612307 on Build GRCh38
rs988610855 Chr.15: 63611897-63611897 on Build GRCh38
rs757655603 Chr.15: 63612363-63612363 on Build GRCh38
rs1056991414 Chr.15: 63612125-63612125 on Build GRCh38
rs560176021 Chr.15: 63612239-63612239 on Build GRCh38
rs142779987 Chr.15: 63612083-63612083 on Build GRCh38
rs773314378 Chr.15: 63612251-63612251 on Build GRCh38
rs575815748 Chr.15: 63612028-63612028 on Build GRCh38
rs1008518506 Chr.15: 63612047-63612047 on Build GRCh38
rs1036993323 Chr.15: 63611977-63611977 on Build GRCh38
rs898598191 Chr.15: 63611993-63611993 on Build GRCh38
rs895173739 Chr.15: 63611918-63611918 on Build GRCh38
rs374885302 Chr.15: 63612048-63612048 on Build GRCh38
rs200151191 Chr.15: 63612371-63612371 on Build GRCh38
rs535690614 Chr.15: 63611922-63611922 on Build GRCh38
rs759086324 Chr.15: 63612345-63612345 on Build GRCh38
rs748335433 Chr.15: 63612180-63612180 on Build GRCh38
rs902567582 Chr.15: 63612110-63612110 on Build GRCh38
rs761748895 Chr.15: 63612242-63612242 on Build GRCh38
rs571887592 Chr.15: 63611896-63611896 on Build GRCh38
rs576650438 Chr.15: 63612119-63612119 on Build GRCh38

View Full Product Details