Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65642816-65643316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC24A1
Entrez Gene ID
9187
Gene Name
solute carrier family 24 member 1
Gene Aliases
CSNB1D, HsT17412, NCKX, NCKX1, RODX
UniGene
Hs.173092
Gene Chromosome Location
Chr.15: 65611405-65660995 on Build GRCh38
Transcripts
NM_001254740.1 NM_001301031.1 NM_001301032.1 NM_001301033.1 NM_004727.2 XM_005254778.3 XM_005254781.3 XM_006720768.3 XM_011522219.1 XM_011522220.2 XM_011522221.2 XM_011522222.2 XM_011522226.2 XM_017022724.1 XM_017022725.1

SNP Detail

SNP ID Chromosome Location
rs140623274 Chr.15: 65642939-65642939 on Build GRCh38
rs1052872783 Chr.15: 65643277-65643277 on Build GRCh38
rs903273470 Chr.15: 65642916-65642916 on Build GRCh38
rs922181911 Chr.15: 65642892-65642892 on Build GRCh38
rs903221757 Chr.15: 65643220-65643220 on Build GRCh38
rs984482850 Chr.15: 65643026-65643026 on Build GRCh38
rs1030830950 Chr.15: 65642931-65642931 on Build GRCh38
rs951056113 Chr.15: 65643079-65643079 on Build GRCh38
rs758093292 Chr.15: 65643052-65643054 on Build GRCh38
rs544859896 Chr.15: 65642977-65642977 on Build GRCh38
rs976231807 Chr.15: 65642868-65642868 on Build GRCh38
rs1041552323 Chr.15: 65643233-65643233 on Build GRCh38
rs80200212 Chr.15: 65642961-65642961 on Build GRCh38
rs542846443 Chr.15: 65642917-65642917 on Build GRCh38
rs935304629 Chr.15: 65643232-65643232 on Build GRCh38
rs773143272 Chr.15: 65643013-65643013 on Build GRCh38
rs755690614 Chr.15: 65642946-65642946 on Build GRCh38
rs1043505439 Chr.15: 65642901-65642901 on Build GRCh38
rs772149325 Chr.15: 65643051-65643052 on Build GRCh38
rs1027331140 Chr.15: 65643054-65643054 on Build GRCh38
rs947389091 Chr.15: 65642881-65642881 on Build GRCh38
rs12101597 Chr.15: 65642857-65642857 on Build GRCh38
rs1043065183 Chr.15: 65643182-65643182 on Build GRCh38
rs991717880 Chr.15: 65643095-65643095 on Build GRCh38
rs566738277 Chr.15: 65642945-65642945 on Build GRCh38
rs550538508 Chr.15: 65643227-65643227 on Build GRCh38
rs527636293 Chr.15: 65642844-65642844 on Build GRCh38
rs1016389650 Chr.15: 65643053-65643053 on Build GRCh38
rs745349959 Chr.15: 65643083-65643083 on Build GRCh38
rs1013390493 Chr.15: 65643051-65643053 on Build GRCh38
rs779637051 Chr.15: 65643248-65643248 on Build GRCh38
rs916178686 Chr.15: 65643148-65643148 on Build GRCh38
rs1019328375 Chr.15: 65643096-65643096 on Build GRCh38
rs972259898 Chr.15: 65643067-65643067 on Build GRCh38
rs901664991 Chr.15: 65643276-65643276 on Build GRCh38
rs530784949 Chr.15: 65643160-65643160 on Build GRCh38
rs953615411 Chr.15: 65643184-65643185 on Build GRCh38
rs928775633 Chr.15: 65643069-65643069 on Build GRCh38
rs953023755 Chr.15: 65642996-65642996 on Build GRCh38
rs979678474 Chr.15: 65643202-65643202 on Build GRCh38
rs947335559 Chr.15: 65643179-65643179 on Build GRCh38
rs757893843 Chr.15: 65643110-65643110 on Build GRCh38
rs753075064 Chr.15: 65642919-65642919 on Build GRCh38
rs961820901 Chr.15: 65643064-65643064 on Build GRCh38
rs912318841 Chr.15: 65642849-65642849 on Build GRCh38
rs1052504031 Chr.15: 65642960-65642960 on Build GRCh38
rs564952731 Chr.15: 65643125-65643125 on Build GRCh38
rs774562377 Chr.15: 65643054-65643055 on Build GRCh38
rs557122521 Chr.15: 65642850-65642850 on Build GRCh38

View Full Product Details