Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 66702238-66702774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAD6
Entrez Gene ID
4091
Gene Name
SMAD family member 6
Gene Aliases
AOVD2, HsT17432, MADH6, MADH7
UniGene
Hs.153863
Gene Chromosome Location
Chr.15: 66702110-66782000 on Build GRCh38
Transcripts
NM_005585.4 NR_027654.1

SNP Detail

SNP ID Chromosome Location
rs965774411 Chr.15: 66702534-66702534 on Build GRCh38
rs753438218 Chr.15: 66702516-66702516 on Build GRCh38
rs777632523 Chr.15: 66702588-66702588 on Build GRCh38
rs1046103423 Chr.15: 66702638-66702638 on Build GRCh38
rs989742026 Chr.15: 66702586-66702586 on Build GRCh38
rs556096586 Chr.15: 66702709-66702710 on Build GRCh38
rs1034268201 Chr.15: 66702571-66702571 on Build GRCh38
rs34643453 Chr.15: 66702737-66702737 on Build GRCh38
rs1002260847 Chr.15: 66702549-66702549 on Build GRCh38
rs994715544 Chr.15: 66702471-66702471 on Build GRCh38
rs989694477 Chr.15: 66702347-66702347 on Build GRCh38
rs1024784982 Chr.15: 66702307-66702307 on Build GRCh38
rs956642131 Chr.15: 66702275-66702275 on Build GRCh38
rs914183425 Chr.15: 66702606-66702606 on Build GRCh38
rs913586851 Chr.15: 66702363-66702363 on Build GRCh38
rs994704153 Chr.15: 66702286-66702286 on Build GRCh38
rs903609665 Chr.15: 66702400-66702400 on Build GRCh38
rs879650048 Chr.15: 66702310-66702311 on Build GRCh38
rs543137828 Chr.15: 66702518-66702518 on Build GRCh38
rs558891070 Chr.15: 66702484-66702484 on Build GRCh38
rs545634412 Chr.15: 66702448-66702448 on Build GRCh38
rs921520374 Chr.15: 66702552-66702552 on Build GRCh38
rs987580858 Chr.15: 66702637-66702637 on Build GRCh38
rs148563981 Chr.15: 66702311-66702312 on Build GRCh38
rs908514986 Chr.15: 66702502-66702502 on Build GRCh38
rs970820372 Chr.15: 66702321-66702321 on Build GRCh38
rs879836195 Chr.15: 66702447-66702448 on Build GRCh38
rs943119720 Chr.15: 66702653-66702653 on Build GRCh38
rs926806528 Chr.15: 66702329-66702329 on Build GRCh38
rs751515365 Chr.15: 66702554-66702554 on Build GRCh38
rs1024973414 Chr.15: 66702499-66702499 on Build GRCh38
rs567690010 Chr.15: 66702317-66702317 on Build GRCh38
rs1050098913 Chr.15: 66702397-66702397 on Build GRCh38
rs1025318462 Chr.15: 66702438-66702438 on Build GRCh38
rs1013474086 Chr.15: 66702436-66702436 on Build GRCh38
rs920010859 Chr.15: 66702314-66702314 on Build GRCh38
rs936407849 Chr.15: 66702584-66702584 on Build GRCh38
rs754325733 Chr.15: 66702306-66702306 on Build GRCh38
rs1002034622 Chr.15: 66702477-66702477 on Build GRCh38
rs981387424 Chr.15: 66702324-66702324 on Build GRCh38
rs930507320 Chr.15: 66702705-66702705 on Build GRCh38
rs573453313 Chr.15: 66702434-66702434 on Build GRCh38
rs866061987 Chr.15: 66702442-66702442 on Build GRCh38
rs1002148833 Chr.15: 66702686-66702686 on Build GRCh38
rs958542086 Chr.15: 66702333-66702333 on Build GRCh38
rs1043198843 Chr.15: 66702574-66702574 on Build GRCh38
rs1047608691 Chr.15: 66702476-66702476 on Build GRCh38
rs542208222 Chr.15: 66702719-66702719 on Build GRCh38
rs141197789 Chr.15: 66702652-66702652 on Build GRCh38
rs966931624 Chr.15: 66702626-66702626 on Build GRCh38
rs1038741815 Chr.15: 66702684-66702684 on Build GRCh38
rs944992593 Chr.15: 66702368-66702368 on Build GRCh38
rs905100270 Chr.15: 66702660-66702660 on Build GRCh38
rs952541258 Chr.15: 66702710-66702710 on Build GRCh38
rs553459285 Chr.15: 66702319-66702319 on Build GRCh38
rs911652692 Chr.15: 66702401-66702401 on Build GRCh38
rs74327941 Chr.15: 66702709-66702709 on Build GRCh38
rs62005619 Chr.15: 66702492-66702492 on Build GRCh38
rs921415389 Chr.15: 66702345-66702345 on Build GRCh38
rs958171994 Chr.15: 66702580-66702580 on Build GRCh38
rs553667868 Chr.15: 66702344-66702344 on Build GRCh38
rs898963434 Chr.15: 66702464-66702464 on Build GRCh38
rs1052095199 Chr.15: 66702431-66702431 on Build GRCh38
rs1051413954 Chr.15: 66702378-66702378 on Build GRCh38
rs757307293 Chr.15: 66702316-66702316 on Build GRCh38
rs920442133 Chr.15: 66702690-66702690 on Build GRCh38
rs184224213 Chr.15: 66702718-66702718 on Build GRCh38
rs1052625362 Chr.15: 66702598-66702600 on Build GRCh38
rs892150221 Chr.15: 66702432-66702432 on Build GRCh38

View Full Product Details