Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 66708450-66708970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAD6
Entrez Gene ID
4091
Gene Name
SMAD family member 6
Gene Aliases
AOVD2, HsT17432, MADH6, MADH7
UniGene
Hs.153863
Gene Chromosome Location
Chr.15: 66702110-66782000 on Build GRCh38
Transcripts
NM_005585.4 NR_027654.1 XM_011521561.2

SNP Detail

SNP ID Chromosome Location
rs747049504 Chr.15: 66708798-66708798 on Build GRCh38
rs902808266 Chr.15: 66708773-66708773 on Build GRCh38
rs763177723 Chr.15: 66708657-66708657 on Build GRCh38
rs142756715 Chr.15: 66708816-66708816 on Build GRCh38
rs756262890 Chr.15: 66708711-66708711 on Build GRCh38
rs1015293089 Chr.15: 66708666-66708666 on Build GRCh38
rs947004527 Chr.15: 66708504-66708504 on Build GRCh38
rs963507517 Chr.15: 66708746-66708746 on Build GRCh38
rs981334919 Chr.15: 66708872-66708872 on Build GRCh38
rs891024941 Chr.15: 66708590-66708590 on Build GRCh38
rs148738763 Chr.15: 66708894-66708894 on Build GRCh38
rs770220183 Chr.15: 66708785-66708785 on Build GRCh38
rs768882941 Chr.15: 66708876-66708876 on Build GRCh38
rs752887529 Chr.15: 66708702-66708702 on Build GRCh38
rs768079244 Chr.15: 66708520-66708520 on Build GRCh38
rs755274303 Chr.15: 66708683-66708683 on Build GRCh38
rs902746530 Chr.15: 66708555-66708555 on Build GRCh38
rs1040633057 Chr.15: 66708942-66708942 on Build GRCh38
rs1039631562 Chr.15: 66708819-66708819 on Build GRCh38
rs1011411932 Chr.15: 66708855-66708855 on Build GRCh38
rs751699745 Chr.15: 66708678-66708678 on Build GRCh38
rs924087571 Chr.15: 66708926-66708926 on Build GRCh38
rs1042559335 Chr.15: 66708728-66708728 on Build GRCh38
rs779333249 Chr.15: 66708564-66708564 on Build GRCh38
rs1027501663 Chr.15: 66708629-66708629 on Build GRCh38
rs576500148 Chr.15: 66708938-66708938 on Build GRCh38
rs1005155354 Chr.15: 66708830-66708830 on Build GRCh38
rs1035033749 Chr.15: 66708874-66708874 on Build GRCh38
rs971307914 Chr.15: 66708857-66708857 on Build GRCh38
rs971294762 Chr.15: 66708667-66708667 on Build GRCh38
rs991761074 Chr.15: 66708719-66708719 on Build GRCh38
rs1038095553 Chr.15: 66708519-66708519 on Build GRCh38
rs918317723 Chr.15: 66708793-66708793 on Build GRCh38
rs995858122 Chr.15: 66708618-66708618 on Build GRCh38
rs952000350 Chr.15: 66708640-66708640 on Build GRCh38
rs542599133 Chr.15: 66708599-66708599 on Build GRCh38
rs1014723871 Chr.15: 66708833-66708833 on Build GRCh38
rs892976347 Chr.15: 66708641-66708641 on Build GRCh38
rs539852832 Chr.15: 66708560-66708560 on Build GRCh38
rs554896732 Chr.15: 66708815-66708815 on Build GRCh38
rs944971743 Chr.15: 66708927-66708927 on Build GRCh38
rs995784667 Chr.15: 66708814-66708814 on Build GRCh38
rs938097860 Chr.15: 66708695-66708695 on Build GRCh38
rs544751160 Chr.15: 66708904-66708904 on Build GRCh38
rs893091703 Chr.15: 66708850-66708850 on Build GRCh38
rs968152651 Chr.15: 66708921-66708921 on Build GRCh38
rs184581334 Chr.15: 66708561-66708561 on Build GRCh38
rs946898512 Chr.15: 66708725-66708725 on Build GRCh38
rs776932207 Chr.15: 66708923-66708923 on Build GRCh38
rs767689205 Chr.15: 66708686-66708686 on Build GRCh38
rs929643485 Chr.15: 66708568-66708568 on Build GRCh38
rs756974446 Chr.15: 66708784-66708784 on Build GRCh38
rs1040033960 Chr.15: 66708790-66708790 on Build GRCh38
rs942573511 Chr.15: 66708819-66708819 on Build GRCh38
rs990817368 Chr.15: 66708891-66708891 on Build GRCh38
rs138179716 Chr.15: 66708777-66708777 on Build GRCh38
rs927911947 Chr.15: 66708692-66708692 on Build GRCh38
rs777808877 Chr.15: 66708727-66708727 on Build GRCh38
rs773784098 Chr.15: 66708892-66708892 on Build GRCh38
rs536774009 Chr.15: 66708485-66708485 on Build GRCh38
rs765907179 Chr.15: 66708871-66708872 on Build GRCh38
rs189474858 Chr.15: 66708571-66708571 on Build GRCh38
rs772612672 Chr.15: 66708691-66708691 on Build GRCh38
rs981420668 Chr.15: 66708688-66708688 on Build GRCh38
rs757551291 Chr.15: 66708529-66708529 on Build GRCh38
rs1020372116 Chr.15: 66708673-66708673 on Build GRCh38
rs146106987 Chr.15: 66708827-66708827 on Build GRCh38
rs766796320 Chr.15: 66708669-66708669 on Build GRCh38
rs1041594693 Chr.15: 66708886-66708886 on Build GRCh38
rs1135009 Chr.15: 66708717-66708717 on Build GRCh38
rs915319066 Chr.15: 66708715-66708715 on Build GRCh38

View Full Product Details