Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90084921-90085418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDH2
Entrez Gene ID
3418
Gene Name
isocitrate dehydrogenase (NADP(+)) 2, mitochondrial
Gene Aliases
D2HGA2, ICD-M, IDH, IDHM, IDP, IDPM, mNADP-IDH
UniGene
Hs.596461
Gene Chromosome Location
Chr.15: 90083978-90102554 on Build GRCh38
Transcripts
NM_001289910.1 NM_001290114.1 NM_002168.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6282012 c.1101C>G p.N367K Chr.15: 90085078-90085078 on Build GRCh38
6683617 c.1107C>T p.I369I Chr.15: 90085072-90085072 on Build GRCh38
6318877 c.1050C>A p.T350T Chr.15: 90085305-90085305 on Build GRCh38
4652657 c.1086G>A p.R362R Chr.15: 90085093-90085093 on Build GRCh38
2139728 c.1129C>T p.R377C Chr.15: 90085050-90085050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61737002 Chr.15: 90085362-90085362 on Build GRCh38
rs758449799 Chr.15: 90085395-90085395 on Build GRCh38
rs1045351891 Chr.15: 90085105-90085105 on Build GRCh38
rs748038847 Chr.15: 90085283-90085283 on Build GRCh38
rs147578332 Chr.15: 90085094-90085094 on Build GRCh38
rs770164108 Chr.15: 90084979-90084979 on Build GRCh38
rs140596855 Chr.15: 90085352-90085352 on Build GRCh38
rs770548230 Chr.15: 90085301-90085301 on Build GRCh38
rs751842511 Chr.15: 90085146-90085146 on Build GRCh38
rs536380173 Chr.15: 90085158-90085158 on Build GRCh38
rs774623780 Chr.15: 90085343-90085343 on Build GRCh38
rs770463292 Chr.15: 90085047-90085048 on Build GRCh38
rs112703257 Chr.15: 90085111-90085111 on Build GRCh38
rs915076191 Chr.15: 90085068-90085068 on Build GRCh38
rs145046086 Chr.15: 90085041-90085041 on Build GRCh38
rs927842567 Chr.15: 90085330-90085330 on Build GRCh38
rs372543878 Chr.15: 90084981-90084981 on Build GRCh38
rs758669079 Chr.15: 90084988-90084988 on Build GRCh38
rs753119952 Chr.15: 90085069-90085069 on Build GRCh38
rs201125516 Chr.15: 90085107-90085107 on Build GRCh38
rs377420001 Chr.15: 90085049-90085049 on Build GRCh38
rs748020391 Chr.15: 90085108-90085108 on Build GRCh38
rs28397672 Chr.15: 90085153-90085153 on Build GRCh38
rs747181428 Chr.15: 90085006-90085006 on Build GRCh38
rs965672106 Chr.15: 90085170-90085170 on Build GRCh38
rs770852509 Chr.15: 90085010-90085010 on Build GRCh38
rs1036260002 Chr.15: 90085333-90085333 on Build GRCh38
rs149060243 Chr.15: 90085289-90085289 on Build GRCh38
rs61737003 Chr.15: 90085359-90085359 on Build GRCh38
rs897159348 Chr.15: 90085336-90085336 on Build GRCh38
rs965380139 Chr.15: 90084982-90084982 on Build GRCh38
rs11540478 Chr.15: 90085305-90085305 on Build GRCh38
rs751677526 Chr.15: 90085078-90085078 on Build GRCh38
rs768606943 Chr.15: 90085016-90085016 on Build GRCh38
rs1024150410 Chr.15: 90085230-90085230 on Build GRCh38
rs34807593 Chr.15: 90085172-90085172 on Build GRCh38
rs375124611 Chr.15: 90085127-90085127 on Build GRCh38
rs765909746 Chr.15: 90085303-90085304 on Build GRCh38
rs777959383 Chr.15: 90084993-90084993 on Build GRCh38
rs760087240 Chr.15: 90084948-90084948 on Build GRCh38
rs775911070 Chr.15: 90085134-90085134 on Build GRCh38
rs761675086 Chr.15: 90085028-90085028 on Build GRCh38
rs952872041 Chr.15: 90085147-90085147 on Build GRCh38
rs749134763 Chr.15: 90085012-90085012 on Build GRCh38
rs550377664 Chr.15: 90085370-90085370 on Build GRCh38
rs974075713 Chr.15: 90085067-90085067 on Build GRCh38
rs61733005 Chr.15: 90085358-90085358 on Build GRCh38
rs745349081 Chr.15: 90085184-90085184 on Build GRCh38
rs375346245 Chr.15: 90085287-90085287 on Build GRCh38
rs764320528 Chr.15: 90085144-90085144 on Build GRCh38
rs763324120 Chr.15: 90085141-90085141 on Build GRCh38
rs769180013 Chr.15: 90085335-90085335 on Build GRCh38
rs776053421 Chr.15: 90085128-90085128 on Build GRCh38
rs190078206 Chr.15: 90085317-90085317 on Build GRCh38
rs1033656067 Chr.15: 90085276-90085276 on Build GRCh38
rs755012033 Chr.15: 90085106-90085106 on Build GRCh38
rs778995704 Chr.15: 90085250-90085250 on Build GRCh38
rs113616815 Chr.15: 90085191-90085191 on Build GRCh38
rs751178080 Chr.15: 90085395-90085396 on Build GRCh38
rs780056514 Chr.15: 90085011-90085011 on Build GRCh38
rs867770113 Chr.15: 90085003-90085003 on Build GRCh38
rs765748772 Chr.15: 90085045-90085045 on Build GRCh38
rs750522764 Chr.15: 90085237-90085237 on Build GRCh38
rs143339918 Chr.15: 90085296-90085296 on Build GRCh38
rs970708194 Chr.15: 90084949-90084949 on Build GRCh38
rs577018029 Chr.15: 90084956-90084956 on Build GRCh38
rs564700723 Chr.15: 90085345-90085345 on Build GRCh38
rs767061831 Chr.15: 90085316-90085316 on Build GRCh38
rs989223286 Chr.15: 90085286-90085286 on Build GRCh38
rs906576308 Chr.15: 90085231-90085231 on Build GRCh38
rs756256300 Chr.15: 90085245-90085245 on Build GRCh38
rs752711856 Chr.15: 90085368-90085368 on Build GRCh38
rs935922182 Chr.15: 90085171-90085171 on Build GRCh38
rs771906210 Chr.15: 90085030-90085030 on Build GRCh38
rs1027096352 Chr.15: 90084959-90084959 on Build GRCh38
rs773746533 Chr.15: 90085087-90085087 on Build GRCh38
rs765712414 Chr.15: 90085050-90085050 on Build GRCh38
rs766516876 Chr.15: 90085365-90085365 on Build GRCh38
rs201734043 Chr.15: 90084970-90084970 on Build GRCh38
rs202014285 Chr.15: 90085081-90085081 on Build GRCh38
rs549189801 Chr.15: 90085048-90085048 on Build GRCh38
rs1002717781 Chr.15: 90085235-90085235 on Build GRCh38
rs776344212 Chr.15: 90085303-90085303 on Build GRCh38
rs368451876 Chr.15: 90085095-90085095 on Build GRCh38
rs548976610 Chr.15: 90085236-90085236 on Build GRCh38
rs977439502 Chr.15: 90085186-90085186 on Build GRCh38
rs199985988 Chr.15: 90085072-90085072 on Build GRCh38
rs757465848 Chr.15: 90085149-90085149 on Build GRCh38
rs370350675 Chr.15: 90085332-90085332 on Build GRCh38
rs1014128390 Chr.15: 90085196-90085196 on Build GRCh38
rs772930285 Chr.15: 90085121-90085121 on Build GRCh38
rs935057402 Chr.15: 90085027-90085027 on Build GRCh38
rs199778006 Chr.15: 90084964-90084964 on Build GRCh38
rs199967272 Chr.15: 90085260-90085260 on Build GRCh38
rs200676110 Chr.15: 90084980-90084980 on Build GRCh38
rs774323865 Chr.15: 90085023-90085023 on Build GRCh38
rs745389409 Chr.15: 90085304-90085304 on Build GRCh38
rs368018432 Chr.15: 90085132-90085132 on Build GRCh38
rs976444997 Chr.15: 90084983-90084983 on Build GRCh38
rs762428788 Chr.15: 90085130-90085130 on Build GRCh38

View Full Product Details