Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 98485788-98486241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM169B
Entrez Gene ID
283777
Gene Name
family with sequence similarity 169 member B
Gene Aliases
-
UniGene
Hs.668070
Gene Chromosome Location
Chr.15: 98437162-98514382 on Build GRCh38
Transcripts
NM_182562.2

SNP Detail

SNP ID Chromosome Location
rs768315262 Chr.15: 98486023-98486023 on Build GRCh38
rs760758413 Chr.15: 98485893-98485893 on Build GRCh38
rs1013774439 Chr.15: 98486211-98486211 on Build GRCh38
rs749770271 Chr.15: 98486140-98486140 on Build GRCh38
rs549200772 Chr.15: 98485959-98485959 on Build GRCh38
rs374371128 Chr.15: 98485848-98485848 on Build GRCh38
rs775880572 Chr.15: 98486178-98486178 on Build GRCh38
rs371583913 Chr.15: 98486128-98486128 on Build GRCh38
rs1037017154 Chr.15: 98486000-98486000 on Build GRCh38
rs754121406 Chr.15: 98485915-98485915 on Build GRCh38
rs1049026157 Chr.15: 98486093-98486093 on Build GRCh38
rs758084936 Chr.15: 98485985-98485985 on Build GRCh38
rs776252054 Chr.15: 98485860-98485860 on Build GRCh38
rs944152553 Chr.15: 98485970-98485970 on Build GRCh38
rs1021408740 Chr.15: 98486011-98486011 on Build GRCh38
rs547115017 Chr.15: 98485968-98485968 on Build GRCh38
rs755419371 Chr.15: 98485928-98485928 on Build GRCh38
rs778577753 Chr.15: 98485850-98485850 on Build GRCh38
rs773804609 Chr.15: 98486035-98486035 on Build GRCh38
rs746558221 Chr.15: 98486074-98486074 on Build GRCh38
rs374721430 Chr.15: 98485941-98485941 on Build GRCh38
rs771257755 Chr.15: 98486155-98486155 on Build GRCh38
rs746491123 Chr.15: 98485961-98485961 on Build GRCh38
rs917949256 Chr.15: 98486097-98486097 on Build GRCh38
rs367933501 Chr.15: 98485892-98485892 on Build GRCh38
rs929534934 Chr.15: 98486125-98486125 on Build GRCh38
rs571255584 Chr.15: 98485897-98485897 on Build GRCh38
rs992587409 Chr.15: 98486095-98486095 on Build GRCh38
rs565959474 Chr.15: 98486141-98486141 on Build GRCh38
rs942183350 Chr.15: 98486149-98486149 on Build GRCh38
rs937998721 Chr.15: 98486078-98486078 on Build GRCh38
rs779735220 Chr.15: 98486070-98486070 on Build GRCh38
rs988771157 Chr.15: 98485992-98485992 on Build GRCh38
rs900266327 Chr.15: 98486164-98486164 on Build GRCh38
rs768209268 Chr.15: 98486077-98486077 on Build GRCh38
rs759420034 Chr.15: 98485864-98485864 on Build GRCh38
rs572175005 Chr.15: 98485922-98485922 on Build GRCh38
rs779906444 Chr.15: 98485953-98485953 on Build GRCh38
rs745743680 Chr.15: 98485880-98485880 on Build GRCh38
rs887573012 Chr.15: 98486129-98486129 on Build GRCh38
rs549875956 Chr.15: 98485865-98485865 on Build GRCh38
rs371837340 Chr.15: 98485940-98485940 on Build GRCh38
rs757973678 Chr.15: 98485881-98485881 on Build GRCh38
rs993386404 Chr.15: 98486066-98486066 on Build GRCh38

View Full Product Details