Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 684839-685359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR24
Entrez Gene ID
84219
Gene Name
WD repeat domain 24
Gene Aliases
C16orf21, JFP7
UniGene
Hs.459632
Gene Chromosome Location
Chr.16: 684622-690661 on Build GRCh38
Transcripts
NM_032259.3 XM_011522699.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1678742 c.2383C>T p.R795C Chr.16: 685283-685283 on Build GRCh38
4638964 c.2121C>T p.C707C Chr.16: 685075-685075 on Build GRCh38
1479083 c.2087_2088insT p.V697fs*>95 Chr.16: 685108-685109 on Build GRCh38
6821629 c.2323G>A p.A775T Chr.16: 685343-685343 on Build GRCh38
353929 c.2364C>T p.I788I Chr.16: 685302-685302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971649562 Chr.16: 685176-685176 on Build GRCh38
rs764671494 Chr.16: 685217-685217 on Build GRCh38
rs375979254 Chr.16: 684955-684955 on Build GRCh38
rs760598918 Chr.16: 685221-685221 on Build GRCh38
rs770970421 Chr.16: 685269-685269 on Build GRCh38
rs1012977534 Chr.16: 685277-685277 on Build GRCh38
rs773302472 Chr.16: 685230-685231 on Build GRCh38
rs569009168 Chr.16: 684920-684920 on Build GRCh38
rs966980616 Chr.16: 684909-684909 on Build GRCh38
rs775021676 Chr.16: 684995-684995 on Build GRCh38
rs566276565 Chr.16: 684942-684942 on Build GRCh38
rs778441917 Chr.16: 685206-685206 on Build GRCh38
rs374129029 Chr.16: 685332-685332 on Build GRCh38
rs760514508 Chr.16: 684968-684968 on Build GRCh38
rs769058662 Chr.16: 685281-685281 on Build GRCh38
rs779667150 Chr.16: 685140-685140 on Build GRCh38
rs570332169 Chr.16: 685244-685244 on Build GRCh38
rs146884857 Chr.16: 685289-685289 on Build GRCh38
rs757853004 Chr.16: 685264-685264 on Build GRCh38
rs773293239 Chr.16: 685301-685301 on Build GRCh38
rs770181400 Chr.16: 685216-685216 on Build GRCh38
rs754092421 Chr.16: 684888-684888 on Build GRCh38
rs747464095 Chr.16: 684947-684947 on Build GRCh38
rs752975290 Chr.16: 685178-685178 on Build GRCh38
rs369416601 Chr.16: 684891-684891 on Build GRCh38
rs371024925 Chr.16: 685242-685242 on Build GRCh38
rs1047776515 Chr.16: 684958-684958 on Build GRCh38
rs762444832 Chr.16: 684996-684996 on Build GRCh38
rs750134597 Chr.16: 685047-685047 on Build GRCh38
rs776822735 Chr.16: 684959-684959 on Build GRCh38
rs751422521 Chr.16: 684866-684866 on Build GRCh38
rs768245315 Chr.16: 685205-685205 on Build GRCh38
rs763315710 Chr.16: 685230-685230 on Build GRCh38
rs377244545 Chr.16: 685224-685224 on Build GRCh38
rs773040198 Chr.16: 685220-685220 on Build GRCh38
rs964490472 Chr.16: 684940-684940 on Build GRCh38
rs769556203 Chr.16: 685138-685138 on Build GRCh38
rs748879826 Chr.16: 685144-685144 on Build GRCh38
rs758568260 Chr.16: 685088-685088 on Build GRCh38
rs377552218 Chr.16: 685233-685233 on Build GRCh38
rs554901459 Chr.16: 684964-684964 on Build GRCh38
rs763231021 Chr.16: 685179-685179 on Build GRCh38
rs373185778 Chr.16: 685049-685049 on Build GRCh38
rs749723922 Chr.16: 685231-685231 on Build GRCh38
rs142550128 Chr.16: 685278-685278 on Build GRCh38
rs766223526 Chr.16: 685174-685174 on Build GRCh38
rs1039814748 Chr.16: 684939-684939 on Build GRCh38
rs747781138 Chr.16: 685262-685262 on Build GRCh38
rs751571231 Chr.16: 685183-685183 on Build GRCh38
rs755274152 Chr.16: 685048-685048 on Build GRCh38
rs752317430 Chr.16: 685238-685238 on Build GRCh38
rs747578055 Chr.16: 685213-685213 on Build GRCh38
rs989806705 Chr.16: 685190-685190 on Build GRCh38
rs767372966 Chr.16: 685045-685045 on Build GRCh38
rs910328343 Chr.16: 684928-684928 on Build GRCh38
rs200744818 Chr.16: 685166-685166 on Build GRCh38
rs181750639 Chr.16: 685246-685246 on Build GRCh38
rs879778382 Chr.16: 685244-685244 on Build GRCh38
rs370832813 Chr.16: 685199-685199 on Build GRCh38
rs762928513 Chr.16: 684859-684859 on Build GRCh38
rs9930020 Chr.16: 685195-685195 on Build GRCh38
rs201721281 Chr.16: 685153-685153 on Build GRCh38
rs536126296 Chr.16: 685132-685132 on Build GRCh38
rs750856494 Chr.16: 685194-685194 on Build GRCh38
rs1054112455 Chr.16: 684875-684875 on Build GRCh38
rs757059860 Chr.16: 684876-684876 on Build GRCh38
rs371070359 Chr.16: 685228-685228 on Build GRCh38
rs371647091 Chr.16: 684925-684925 on Build GRCh38
rs777300758 Chr.16: 685268-685268 on Build GRCh38
rs766836292 Chr.16: 685318-685318 on Build GRCh38
rs750899485 Chr.16: 685236-685236 on Build GRCh38
rs757822854 Chr.16: 685187-685187 on Build GRCh38
rs573578176 Chr.16: 685056-685056 on Build GRCh38
rs774713417 Chr.16: 685249-685250 on Build GRCh38
rs763987449 Chr.16: 684863-684863 on Build GRCh38
rs376665117 Chr.16: 685271-685271 on Build GRCh38
rs767524698 Chr.16: 685292-685292 on Build GRCh38
rs752327977 Chr.16: 685338-685338 on Build GRCh38
rs922768944 Chr.16: 685222-685222 on Build GRCh38
rs772108403 Chr.16: 685218-685218 on Build GRCh38
rs761587001 Chr.16: 685035-685035 on Build GRCh38
rs766311823 Chr.16: 685030-685030 on Build GRCh38
rs769309492 Chr.16: 684973-684973 on Build GRCh38
rs778422411 Chr.16: 685252-685252 on Build GRCh38
rs752887367 Chr.16: 685050-685050 on Build GRCh38
rs374342069 Chr.16: 685164-685164 on Build GRCh38
rs562033335 Chr.16: 685329-685329 on Build GRCh38
rs924271671 Chr.16: 684860-684860 on Build GRCh38
rs774283981 Chr.16: 685021-685021 on Build GRCh38
rs774448989 Chr.16: 685283-685283 on Build GRCh38
rs764077208 Chr.16: 685014-685014 on Build GRCh38
rs957056065 Chr.16: 685189-685189 on Build GRCh38
rs550620862 Chr.16: 684911-684911 on Build GRCh38
rs199891509 Chr.16: 685177-685177 on Build GRCh38
rs781657443 Chr.16: 685114-685114 on Build GRCh38
rs138956993 Chr.16: 685319-685319 on Build GRCh38
rs866195139 Chr.16: 685053-685053 on Build GRCh38
rs764382850 Chr.16: 685231-685231 on Build GRCh38
rs374885525 Chr.16: 685115-685115 on Build GRCh38
rs1029854104 Chr.16: 684924-684924 on Build GRCh38
rs764293109 Chr.16: 685181-685181 on Build GRCh38
rs781547839 Chr.16: 685191-685191 on Build GRCh38
rs867367783 Chr.16: 685288-685288 on Build GRCh38
rs751730087 Chr.16: 685232-685232 on Build GRCh38
rs539287295 Chr.16: 684921-684921 on Build GRCh38
rs1017028504 Chr.16: 685080-685080 on Build GRCh38
rs199825206 Chr.16: 684952-684952 on Build GRCh38
rs747552035 Chr.16: 685102-685102 on Build GRCh38
rs369281013 Chr.16: 685012-685012 on Build GRCh38
rs147086833 Chr.16: 685171-685171 on Build GRCh38
rs370025736 Chr.16: 685150-685150 on Build GRCh38
rs757658621 Chr.16: 685110-685110 on Build GRCh38
rs767158640 Chr.16: 684877-684877 on Build GRCh38
rs777935852 Chr.16: 685095-685095 on Build GRCh38
rs761215863 Chr.16: 685308-685308 on Build GRCh38

View Full Product Details