Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2979526-2980017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKMYT1
Entrez Gene ID
9088
Gene Name
protein kinase, membrane associated tyrosine/threonine 1
Gene Aliases
MYT1, PPP1R126
UniGene
Hs.732385
Gene Chromosome Location
Chr.16: 2972791-2980539 on Build GRCh38
Transcripts
NM_001258450.1 NM_001258451.1 NM_004203.4 NM_182687.2 XM_011522734.2 XM_011522735.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1262307 c.10+1G>T p.? Chr.16: 2979647-2979647 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs951007536 Chr.16: 2979591-2979591 on Build GRCh38
rs748966961 Chr.16: 2979599-2979599 on Build GRCh38
rs959874182 Chr.16: 2979822-2979822 on Build GRCh38
rs756893831 Chr.16: 2979682-2979682 on Build GRCh38
rs4149770 Chr.16: 2979761-2979761 on Build GRCh38
rs1038907718 Chr.16: 2979947-2979947 on Build GRCh38
rs775542785 Chr.16: 2979640-2979640 on Build GRCh38
rs748354751 Chr.16: 2979698-2979698 on Build GRCh38
rs564833107 Chr.16: 2979955-2979955 on Build GRCh38
rs754991377 Chr.16: 2979627-2979627 on Build GRCh38
rs997974744 Chr.16: 2979612-2979612 on Build GRCh38
rs772084344 Chr.16: 2979631-2979631 on Build GRCh38
rs763582603 Chr.16: 2979680-2979680 on Build GRCh38
rs550001452 Chr.16: 2979598-2979598 on Build GRCh38
rs773589053 Chr.16: 2979670-2979670 on Build GRCh38
rs888713313 Chr.16: 2979589-2979589 on Build GRCh38
rs200568818 Chr.16: 2979678-2979678 on Build GRCh38
rs943267098 Chr.16: 2979924-2979924 on Build GRCh38
rs1012908261 Chr.16: 2979993-2979993 on Build GRCh38
rs768848141 Chr.16: 2979641-2979641 on Build GRCh38
rs572533715 Chr.16: 2979983-2979999 on Build GRCh38
rs373940742 Chr.16: 2979954-2979954 on Build GRCh38
rs371743482 Chr.16: 2979752-2979752 on Build GRCh38
rs757015342 Chr.16: 2979602-2979602 on Build GRCh38
rs878882953 Chr.16: 2979753-2979753 on Build GRCh38
rs528147853 Chr.16: 2979757-2979757 on Build GRCh38
rs1016184912 Chr.16: 2979597-2979597 on Build GRCh38
rs934284606 Chr.16: 2979871-2979871 on Build GRCh38
rs746926825 Chr.16: 2979692-2979692 on Build GRCh38
rs752032639 Chr.16: 2979675-2979675 on Build GRCh38
rs765529436 Chr.16: 2979668-2979668 on Build GRCh38
rs571458666 Chr.16: 2979988-2979988 on Build GRCh38
rs551649159 Chr.16: 2979879-2979879 on Build GRCh38
rs536853506 Chr.16: 2979556-2979556 on Build GRCh38
rs573457928 Chr.16: 2979813-2979813 on Build GRCh38
rs1016667424 Chr.16: 2979984-2979984 on Build GRCh38
rs929494398 Chr.16: 2979758-2979758 on Build GRCh38
rs111899226 Chr.16: 2979872-2979872 on Build GRCh38
rs745822713 Chr.16: 2979630-2979630 on Build GRCh38
rs1005773079 Chr.16: 2979595-2979595 on Build GRCh38
rs181949038 Chr.16: 2979960-2979960 on Build GRCh38
rs776773461 Chr.16: 2979651-2979651 on Build GRCh38
rs1029442137 Chr.16: 2979921-2979921 on Build GRCh38
rs557221650 Chr.16: 2979856-2979857 on Build GRCh38
rs370007178 Chr.16: 2979820-2979820 on Build GRCh38
rs762041837 Chr.16: 2979655-2979655 on Build GRCh38
rs770013129 Chr.16: 2979700-2979700 on Build GRCh38
rs1007369013 Chr.16: 2979718-2979718 on Build GRCh38
rs570170668 Chr.16: 2979672-2979672 on Build GRCh38
rs777746072 Chr.16: 2979709-2979709 on Build GRCh38
rs756980373 Chr.16: 2979557-2979558 on Build GRCh38
rs748395615 Chr.16: 2979622-2979634 on Build GRCh38
rs576123313 Chr.16: 2979913-2979913 on Build GRCh38
rs751498050 Chr.16: 2979765-2979765 on Build GRCh38
rs892972269 Chr.16: 2979714-2979714 on Build GRCh38
rs897362074 Chr.16: 2979891-2979891 on Build GRCh38
rs778702698 Chr.16: 2979609-2979609 on Build GRCh38
rs558558630 Chr.16: 2979693-2979693 on Build GRCh38
rs886461832 Chr.16: 2979964-2979966 on Build GRCh38
rs1016494749 Chr.16: 2979816-2979816 on Build GRCh38
rs1026265708 Chr.16: 2979588-2979588 on Build GRCh38
rs562436597 Chr.16: 2979848-2979848 on Build GRCh38
rs141021541 Chr.16: 2979648-2979648 on Build GRCh38
rs113871907 Chr.16: 2979920-2979920 on Build GRCh38
rs542856724 Chr.16: 2979823-2979823 on Build GRCh38
rs572130432 Chr.16: 2979703-2979703 on Build GRCh38
rs971173320 Chr.16: 2979850-2979851 on Build GRCh38
rs750279591 Chr.16: 2979689-2979689 on Build GRCh38
rs901645467 Chr.16: 2979952-2979952 on Build GRCh38
rs866783117 Chr.16: 2979855-2979855 on Build GRCh38
rs753437637 Chr.16: 2979681-2979681 on Build GRCh38
rs957532340 Chr.16: 2979745-2979745 on Build GRCh38
rs769523724 Chr.16: 2979944-2979944 on Build GRCh38
rs970939325 Chr.16: 2979993-2979994 on Build GRCh38
rs1046492346 Chr.16: 2979797-2979797 on Build GRCh38
rs928032226 Chr.16: 2979819-2979819 on Build GRCh38
rs1039261582 Chr.16: 2979740-2979740 on Build GRCh38
rs780202950 Chr.16: 2979691-2979691 on Build GRCh38
rs781269797 Chr.16: 2979694-2979694 on Build GRCh38
rs770283312 Chr.16: 2979692-2979692 on Build GRCh38
rs757053582 Chr.16: 2979868-2979868 on Build GRCh38
rs1006202821 Chr.16: 2979971-2979971 on Build GRCh38
rs553661069 Chr.16: 2979803-2979803 on Build GRCh38

View Full Product Details