Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3068825-3069371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL32
Entrez Gene ID
9235
Gene Name
interleukin 32
Gene Aliases
IL-32alpha, IL-32beta, IL-32delta, IL-32gamma, NK4, TAIF, TAIFa, TAIFb, TAIFc, TAIFd
UniGene
Hs.943
Gene Chromosome Location
Chr.16: 3065312-3069819 on Build GRCh38
Transcripts
NM_001012631.1 NM_001012632.1 NM_001012633.1 NM_001012634.1 NM_001012635.1 NM_001012636.1 NM_001012718.1 NM_001308078.1 NM_004221.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6144339 c.521A>T p.E174V Chr.16: 3069309-3069309 on Build GRCh38
2157043 c.444C>T p.F148F Chr.16: 3069232-3069232 on Build GRCh38
281907 c.251G>A p.G84D Chr.16: 3069039-3069039 on Build GRCh38
6144340 c.474C>G p.L158L Chr.16: 3069262-3069262 on Build GRCh38
6079037 c.523G>T p.E175* Chr.16: 3069311-3069311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771750080 Chr.16: 3069186-3069186 on Build GRCh38
rs920380792 Chr.16: 3069111-3069111 on Build GRCh38
rs143941291 Chr.16: 3069338-3069338 on Build GRCh38
rs571369564 Chr.16: 3068966-3068966 on Build GRCh38
rs758251866 Chr.16: 3069235-3069235 on Build GRCh38
rs767320072 Chr.16: 3069079-3069079 on Build GRCh38
rs750400589 Chr.16: 3069014-3069015 on Build GRCh38
rs71818662 Chr.16: 3069296-3069297 on Build GRCh38
rs200868372 Chr.16: 3069058-3069058 on Build GRCh38
rs772061055 Chr.16: 3069147-3069147 on Build GRCh38
rs766011549 Chr.16: 3069320-3069320 on Build GRCh38
rs34184287 Chr.16: 3069166-3069166 on Build GRCh38
rs148189652 Chr.16: 3069286-3069286 on Build GRCh38
rs759204196 Chr.16: 3069143-3069143 on Build GRCh38
rs755581741 Chr.16: 3069173-3069174 on Build GRCh38
rs748768387 Chr.16: 3069178-3069178 on Build GRCh38
rs143049351 Chr.16: 3069246-3069246 on Build GRCh38
rs752085136 Chr.16: 3069174-3069174 on Build GRCh38
rs746754097 Chr.16: 3069240-3069240 on Build GRCh38
rs201698562 Chr.16: 3068951-3068951 on Build GRCh38
rs745680533 Chr.16: 3069184-3069184 on Build GRCh38
rs780363363 Chr.16: 3069116-3069116 on Build GRCh38
rs766634535 Chr.16: 3069198-3069198 on Build GRCh38
rs958331534 Chr.16: 3069121-3069121 on Build GRCh38
rs371449782 Chr.16: 3069039-3069039 on Build GRCh38
rs150738098 Chr.16: 3069312-3069312 on Build GRCh38
rs574601205 Chr.16: 3069210-3069210 on Build GRCh38
rs774320014 Chr.16: 3069141-3069141 on Build GRCh38
rs757665795 Chr.16: 3069092-3069092 on Build GRCh38
rs780936859 Chr.16: 3069330-3069330 on Build GRCh38
rs751706652 Chr.16: 3068943-3068943 on Build GRCh38
rs887401579 Chr.16: 3068970-3068970 on Build GRCh38
rs377536439 Chr.16: 3069308-3069308 on Build GRCh38
rs752904266 Chr.16: 3069018-3069018 on Build GRCh38
rs559090050 Chr.16: 3069163-3069163 on Build GRCh38
rs372215212 Chr.16: 3069169-3069169 on Build GRCh38
rs762774579 Chr.16: 3068992-3068992 on Build GRCh38
rs779743927 Chr.16: 3069238-3069238 on Build GRCh38
rs775205321 Chr.16: 3069300-3069301 on Build GRCh38
rs758963925 Chr.16: 3069113-3069113 on Build GRCh38
rs570334137 Chr.16: 3069125-3069125 on Build GRCh38
rs12922880 Chr.16: 3068848-3068848 on Build GRCh38
rs779696582 Chr.16: 3069287-3069287 on Build GRCh38
rs778169419 Chr.16: 3068958-3068958 on Build GRCh38
rs775284592 Chr.16: 3069156-3069156 on Build GRCh38
rs761457048 Chr.16: 3069277-3069277 on Build GRCh38
rs992493049 Chr.16: 3069069-3069069 on Build GRCh38
rs749912248 Chr.16: 3069314-3069314 on Build GRCh38
rs770464943 Chr.16: 3069298-3069299 on Build GRCh38
rs368991361 Chr.16: 3069261-3069261 on Build GRCh38
rs779453646 Chr.16: 3069038-3069038 on Build GRCh38
rs140102091 Chr.16: 3069325-3069325 on Build GRCh38
rs746055082 Chr.16: 3069345-3069345 on Build GRCh38
rs531505867 Chr.16: 3068932-3068932 on Build GRCh38
rs768197322 Chr.16: 3068972-3068972 on Build GRCh38
rs147297738 Chr.16: 3069343-3069343 on Build GRCh38
rs12934561 Chr.16: 3068864-3068864 on Build GRCh38
rs940293113 Chr.16: 3069174-3069174 on Build GRCh38
rs955913137 Chr.16: 3069255-3069255 on Build GRCh38
rs752150665 Chr.16: 3069199-3069199 on Build GRCh38
rs771147572 Chr.16: 3069254-3069254 on Build GRCh38
rs761820961 Chr.16: 3069192-3069192 on Build GRCh38
rs113259139 Chr.16: 3068900-3068900 on Build GRCh38
rs547462668 Chr.16: 3069346-3069346 on Build GRCh38
rs138083601 Chr.16: 3069190-3069190 on Build GRCh38
rs748058182 Chr.16: 3069296-3069296 on Build GRCh38
rs750709861 Chr.16: 3069110-3069110 on Build GRCh38
rs772017218 Chr.16: 3069299-3069300 on Build GRCh38
rs984076512 Chr.16: 3068878-3068878 on Build GRCh38
rs1019695099 Chr.16: 3068853-3068853 on Build GRCh38
rs752845473 Chr.16: 3069082-3069082 on Build GRCh38
rs1034287357 Chr.16: 3069114-3069114 on Build GRCh38
rs768591207 Chr.16: 3069243-3069243 on Build GRCh38
rs139497186 Chr.16: 3069200-3069200 on Build GRCh38
rs768143359 Chr.16: 3069276-3069276 on Build GRCh38
rs772552901 Chr.16: 3069307-3069307 on Build GRCh38
rs774483172 Chr.16: 3069067-3069067 on Build GRCh38
rs978675059 Chr.16: 3068887-3068887 on Build GRCh38
rs754206288 Chr.16: 3069030-3069030 on Build GRCh38
rs748929635 Chr.16: 3069131-3069131 on Build GRCh38
rs113528935 Chr.16: 3068917-3068917 on Build GRCh38
rs770892725 Chr.16: 3069137-3069137 on Build GRCh38
rs968597217 Chr.16: 3068885-3068885 on Build GRCh38
rs781339380 Chr.16: 3068950-3068950 on Build GRCh38
rs751542510 Chr.16: 3068999-3068999 on Build GRCh38
rs181077045 Chr.16: 3069319-3069319 on Build GRCh38
rs776194529 Chr.16: 3068979-3068979 on Build GRCh38
rs780054734 Chr.16: 3069161-3069161 on Build GRCh38
rs1025895864 Chr.16: 3069083-3069083 on Build GRCh38
rs760865971 Chr.16: 3069158-3069158 on Build GRCh38
rs541511099 Chr.16: 3069188-3069188 on Build GRCh38
rs901230804 Chr.16: 3069034-3069034 on Build GRCh38
rs2981599 Chr.16: 3069303-3069303 on Build GRCh38
rs755073142 Chr.16: 3069005-3069005 on Build GRCh38
rs749342733 Chr.16: 3069299-3069299 on Build GRCh38
rs746588354 Chr.16: 3068971-3068971 on Build GRCh38
rs749333838 Chr.16: 3069340-3069340 on Build GRCh38
rs551525801 Chr.16: 3068941-3068941 on Build GRCh38
rs757614578 Chr.16: 3069035-3069035 on Build GRCh38
rs767615947 Chr.16: 3069006-3069006 on Build GRCh38
rs398100042 Chr.16: 3069303-3069304 on Build GRCh38
rs951828852 Chr.16: 3069301-3069301 on Build GRCh38
rs778372895 Chr.16: 3069182-3069182 on Build GRCh38
rs201024346 Chr.16: 3069052-3069052 on Build GRCh38
rs759487686 Chr.16: 3069073-3069073 on Build GRCh38
rs781656938 Chr.16: 3069295-3069296 on Build GRCh38
rs111283867 Chr.16: 3068852-3068852 on Build GRCh38
rs934305559 Chr.16: 3068899-3068899 on Build GRCh38
rs879059449 Chr.16: 3068889-3068889 on Build GRCh38
rs771451025 Chr.16: 3068965-3068965 on Build GRCh38
rs751355275 Chr.16: 3069292-3069292 on Build GRCh38
rs762454460 Chr.16: 3069316-3069316 on Build GRCh38
rs773160368 Chr.16: 3068988-3068988 on Build GRCh38
rs751197524 Chr.16: 3069323-3069323 on Build GRCh38
rs773298486 Chr.16: 3069195-3069195 on Build GRCh38
rs768090259 Chr.16: 3069202-3069202 on Build GRCh38
rs904645833 Chr.16: 3069234-3069234 on Build GRCh38
rs756994398 Chr.16: 3069181-3069181 on Build GRCh38
rs1031589372 Chr.16: 3069062-3069062 on Build GRCh38
rs745486455 Chr.16: 3069216-3069216 on Build GRCh38
rs538977484 Chr.16: 3069161-3069161 on Build GRCh38
rs763437038 Chr.16: 3069197-3069197 on Build GRCh38
rs374583841 Chr.16: 3069167-3069167 on Build GRCh38
rs374680477 Chr.16: 3069130-3069130 on Build GRCh38
rs924238516 Chr.16: 3068890-3068890 on Build GRCh38
rs763056959 Chr.16: 3069251-3069251 on Build GRCh38
rs764612413 Chr.16: 3069313-3069313 on Build GRCh38
rs141220342 Chr.16: 3069302-3069302 on Build GRCh38
rs776711769 Chr.16: 3069191-3069191 on Build GRCh38
rs113049976 Chr.16: 3068922-3068922 on Build GRCh38
rs752453644 Chr.16: 3069331-3069331 on Build GRCh38
rs371244098 Chr.16: 3069019-3069019 on Build GRCh38
rs916999945 Chr.16: 3068921-3068921 on Build GRCh38
rs148655642 Chr.16: 3068866-3068866 on Build GRCh38
rs779217049 Chr.16: 3069094-3069094 on Build GRCh38
rs931479975 Chr.16: 3069122-3069122 on Build GRCh38
rs766427838 Chr.16: 3068997-3068997 on Build GRCh38
rs748964193 Chr.16: 3069297-3069298 on Build GRCh38
rs760045825 Chr.16: 3069304-3069304 on Build GRCh38
rs756761335 Chr.16: 3069206-3069206 on Build GRCh38
rs776480395 Chr.16: 3069245-3069245 on Build GRCh38
rs556206733 Chr.16: 3069088-3069088 on Build GRCh38
rs764366058 Chr.16: 3069090-3069090 on Build GRCh38
rs567269635 Chr.16: 3069057-3069057 on Build GRCh38
rs765446264 Chr.16: 3069194-3069194 on Build GRCh38
rs749614384 Chr.16: 3068960-3068960 on Build GRCh38
rs760465682 Chr.16: 3069301-3069302 on Build GRCh38
rs781160848 Chr.16: 3069293-3069293 on Build GRCh38
rs753414314 Chr.16: 3069205-3069205 on Build GRCh38
rs773536097 Chr.16: 3069249-3069249 on Build GRCh38
rs755737640 Chr.16: 3069171-3069171 on Build GRCh38
rs773468767 Chr.16: 3069320-3069320 on Build GRCh38
rs777924344 Chr.16: 3069029-3069029 on Build GRCh38
rs371460714 Chr.16: 3069140-3069140 on Build GRCh38
rs756486490 Chr.16: 3068955-3068955 on Build GRCh38
rs759924800 Chr.16: 3069268-3069268 on Build GRCh38
rs967532044 Chr.16: 3069151-3069151 on Build GRCh38
rs764135077 Chr.16: 3069160-3069160 on Build GRCh38
rs530081335 Chr.16: 3068877-3068877 on Build GRCh38
rs527364961 Chr.16: 3068981-3068981 on Build GRCh38
rs897865773 Chr.16: 3069264-3069264 on Build GRCh38
rs752000903 Chr.16: 3069170-3069170 on Build GRCh38
rs761494040 Chr.16: 3068980-3068980 on Build GRCh38
rs768089141 Chr.16: 3069048-3069048 on Build GRCh38
rs758499890 Chr.16: 3069130-3069142 on Build GRCh38
rs531600758 Chr.16: 3069297-3069297 on Build GRCh38
rs779733588 Chr.16: 3069214-3069214 on Build GRCh38
rs373154056 Chr.16: 3069279-3069279 on Build GRCh38

View Full Product Details