Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3440728-3441239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF597
Entrez Gene ID
146434
Gene Name
zinc finger protein 597
Gene Aliases
HIT-4
UniGene
Hs.133338
Gene Chromosome Location
Chr.16: 3432422-3443537 on Build GRCh38
Transcripts
NM_152457.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
970451 c.133G>T p.E45* Chr.16: 3440834-3440834 on Build GRCh38
7177632 c.114C>T p.L38L Chr.16: 3440853-3440853 on Build GRCh38
7184101 c.73C>G p.Q25E Chr.16: 3440894-3440894 on Build GRCh38
4837908 c.133G>C p.E45Q Chr.16: 3440834-3440834 on Build GRCh38
1725161 c.37C>G p.P13A Chr.16: 3440930-3440930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2270496 Chr.16: 3441067-3441067 on Build GRCh38
rs78463667 Chr.16: 3441155-3441155 on Build GRCh38
rs756185895 Chr.16: 3440924-3440924 on Build GRCh38
rs377060983 Chr.16: 3440798-3440798 on Build GRCh38
rs115389621 Chr.16: 3441085-3441085 on Build GRCh38
rs1036170609 Chr.16: 3440965-3440965 on Build GRCh38
rs933189921 Chr.16: 3441134-3441134 on Build GRCh38
rs769741254 Chr.16: 3440883-3440883 on Build GRCh38
rs939585654 Chr.16: 3441144-3441144 on Build GRCh38
rs183222316 Chr.16: 3440944-3440944 on Build GRCh38
rs1004127254 Chr.16: 3440882-3440882 on Build GRCh38
rs753108515 Chr.16: 3441044-3441044 on Build GRCh38
rs749631398 Chr.16: 3440853-3440853 on Build GRCh38
rs369748871 Chr.16: 3440818-3440818 on Build GRCh38
rs932744957 Chr.16: 3441181-3441181 on Build GRCh38
rs376011364 Chr.16: 3440973-3440973 on Build GRCh38
rs1035966257 Chr.16: 3440848-3440848 on Build GRCh38
rs769508666 Chr.16: 3440835-3440835 on Build GRCh38
rs981520329 Chr.16: 3441056-3441056 on Build GRCh38
rs992458412 Chr.16: 3441159-3441159 on Build GRCh38
rs1045856573 Chr.16: 3440925-3440925 on Build GRCh38
rs193294026 Chr.16: 3441196-3441196 on Build GRCh38
rs754321944 Chr.16: 3440930-3440930 on Build GRCh38
rs766516287 Chr.16: 3440993-3440993 on Build GRCh38
rs1020416930 Chr.16: 3441031-3441031 on Build GRCh38
rs766550281 Chr.16: 3440811-3440811 on Build GRCh38
rs747370056 Chr.16: 3441204-3441204 on Build GRCh38
rs769211624 Chr.16: 3440962-3440962 on Build GRCh38
rs372252543 Chr.16: 3440757-3440757 on Build GRCh38
rs776268734 Chr.16: 3440874-3440874 on Build GRCh38
rs2270495 Chr.16: 3440933-3440933 on Build GRCh38
rs1009677956 Chr.16: 3440916-3440916 on Build GRCh38
rs926260253 Chr.16: 3440827-3440827 on Build GRCh38
rs980423534 Chr.16: 3441109-3441111 on Build GRCh38
rs1000516334 Chr.16: 3441015-3441015 on Build GRCh38
rs762806919 Chr.16: 3440972-3440972 on Build GRCh38
rs781258220 Chr.16: 3440833-3440833 on Build GRCh38
rs373859952 Chr.16: 3440955-3440955 on Build GRCh38
rs751717460 Chr.16: 3440912-3440912 on Build GRCh38
rs868311395 Chr.16: 3441102-3441102 on Build GRCh38
rs576545882 Chr.16: 3441012-3441012 on Build GRCh38
rs147007149 Chr.16: 3440952-3440952 on Build GRCh38
rs940587106 Chr.16: 3441096-3441096 on Build GRCh38
rs531863578 Chr.16: 3441033-3441033 on Build GRCh38
rs762762995 Chr.16: 3440904-3440904 on Build GRCh38
rs1020993991 Chr.16: 3441210-3441210 on Build GRCh38
rs1045328136 Chr.16: 3440870-3440870 on Build GRCh38
rs771730194 Chr.16: 3440767-3440767 on Build GRCh38
rs1000588466 Chr.16: 3441171-3441171 on Build GRCh38
rs575391198 Chr.16: 3440787-3440787 on Build GRCh38
rs144056794 Chr.16: 3441177-3441177 on Build GRCh38
rs888889102 Chr.16: 3441167-3441167 on Build GRCh38
rs2270494 Chr.16: 3440922-3440922 on Build GRCh38
rs751567575 Chr.16: 3440975-3440975 on Build GRCh38
rs763718130 Chr.16: 3440784-3440784 on Build GRCh38
rs759754321 Chr.16: 3440919-3440919 on Build GRCh38
rs868314146 Chr.16: 3440906-3440906 on Build GRCh38
rs78646901 Chr.16: 3441029-3441029 on Build GRCh38
rs369017760 Chr.16: 3440948-3440948 on Build GRCh38
rs139189056 Chr.16: 3440932-3440932 on Build GRCh38
rs762698312 Chr.16: 3440809-3440809 on Build GRCh38
rs953053062 Chr.16: 3440773-3440773 on Build GRCh38
rs747656867 Chr.16: 3440961-3440961 on Build GRCh38
rs756627180 Chr.16: 3440842-3440842 on Build GRCh38
rs889011207 Chr.16: 3441030-3441030 on Build GRCh38
rs779802564 Chr.16: 3440862-3440862 on Build GRCh38
rs373855901 Chr.16: 3440943-3440943 on Build GRCh38
rs547464572 Chr.16: 3441117-3441117 on Build GRCh38
rs926334001 Chr.16: 3441136-3441136 on Build GRCh38
rs778119523 Chr.16: 3441100-3441100 on Build GRCh38
rs961943400 Chr.16: 3441064-3441064 on Build GRCh38
rs903817853 Chr.16: 3441023-3441023 on Build GRCh38
rs778189322 Chr.16: 3440845-3440845 on Build GRCh38
rs551959393 Chr.16: 3441063-3441063 on Build GRCh38
rs751805268 Chr.16: 3440813-3440813 on Build GRCh38
rs149503721 Chr.16: 3440988-3440988 on Build GRCh38
rs575500769 Chr.16: 3441202-3441202 on Build GRCh38
rs957621109 Chr.16: 3441020-3441020 on Build GRCh38
rs1005228798 Chr.16: 3440964-3440964 on Build GRCh38
rs778433317 Chr.16: 3441107-3441107 on Build GRCh38
rs766177607 Chr.16: 3440910-3440910 on Build GRCh38
rs37831 Chr.16: 3441209-3441209 on Build GRCh38
rs750211917 Chr.16: 3440804-3440804 on Build GRCh38
rs759214484 Chr.16: 3440820-3440820 on Build GRCh38
rs746673058 Chr.16: 3440869-3440869 on Build GRCh38
rs774114637 Chr.16: 3440976-3440976 on Build GRCh38
rs201912721 Chr.16: 3440873-3440873 on Build GRCh38
rs754046674 Chr.16: 3441014-3441014 on Build GRCh38
rs760221241 Chr.16: 3440774-3440774 on Build GRCh38
rs1009225810 Chr.16: 3441199-3441201 on Build GRCh38
rs138236442 Chr.16: 3440956-3440956 on Build GRCh38
rs2270493 Chr.16: 3440879-3440879 on Build GRCh38
rs775655410 Chr.16: 3440779-3440779 on Build GRCh38
rs879851565 Chr.16: 3441170-3441170 on Build GRCh38
rs747573099 Chr.16: 3441192-3441192 on Build GRCh38
rs984830951 Chr.16: 3441082-3441082 on Build GRCh38
rs971820319 Chr.16: 3440960-3440960 on Build GRCh38
rs911642918 Chr.16: 3440779-3440779 on Build GRCh38
rs887726122 Chr.16: 3440957-3440957 on Build GRCh38
rs773147069 Chr.16: 3440963-3440963 on Build GRCh38
rs879318630 Chr.16: 3440788-3440788 on Build GRCh38
rs752796522 Chr.16: 3440923-3440923 on Build GRCh38
rs771229009 Chr.16: 3440856-3440856 on Build GRCh38
rs773056591 Chr.16: 3440894-3440894 on Build GRCh38
rs1033661281 Chr.16: 3441018-3441018 on Build GRCh38
rs957758718 Chr.16: 3440855-3440855 on Build GRCh38
rs765146442 Chr.16: 3440802-3440802 on Build GRCh38

View Full Product Details