Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 21968495-21968946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDZD9
Entrez Gene ID
255762
Gene Name
PDZ domain containing 9
Gene Aliases
C16orf65
UniGene
Hs.98244
Gene Chromosome Location
Chr.16: 21957540-22001137 on Build GRCh38
Transcripts
XM_011545785.2 XM_017023109.1 XM_017023110.1 NM_003366.3

Gene Symbol
UQCRC2
Entrez Gene ID
7385
Gene Name
ubiquinol-cytochrome c reductase core protein II
Gene Aliases
MC3DN5, QCR2, UQCR2
UniGene
Hs.528803
Gene Chromosome Location
Chr.16: 21953064-21983660 on Build GRCh38
Transcripts
XM_011545785.2 XM_017023109.1 XM_017023110.1 NM_003366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3507587 c.634C>A p.H212N Chr.16: 21968649-21968649 on Build GRCh38
3817506 c.630G>A p.Q210Q Chr.16: 21968645-21968645 on Build GRCh38
5414141 c.634C>T p.H212Y Chr.16: 21968649-21968649 on Build GRCh38
6001979 c.670+5G>A p.? Chr.16: 21968690-21968690 on Build GRCh38
32283 c.623T>A p.F208Y Chr.16: 21968638-21968638 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767810414 Chr.16: 21968674-21968674 on Build GRCh38
rs919059873 Chr.16: 21968883-21968883 on Build GRCh38
rs749807095 Chr.16: 21968583-21968583 on Build GRCh38
rs903120059 Chr.16: 21968847-21968847 on Build GRCh38
rs113206588 Chr.16: 21968813-21968813 on Build GRCh38
rs778037846 Chr.16: 21968684-21968684 on Build GRCh38
rs753648710 Chr.16: 21968625-21968625 on Build GRCh38
rs774068164 Chr.16: 21968598-21968598 on Build GRCh38
rs186427842 Chr.16: 21968561-21968561 on Build GRCh38
rs528631587 Chr.16: 21968904-21968904 on Build GRCh38
rs368615913 Chr.16: 21968644-21968644 on Build GRCh38
rs1012550374 Chr.16: 21968744-21968744 on Build GRCh38
rs997738115 Chr.16: 21968806-21968806 on Build GRCh38
rs938523554 Chr.16: 21968891-21968891 on Build GRCh38
rs957131468 Chr.16: 21968787-21968787 on Build GRCh38
rs949919126 Chr.16: 21968758-21968758 on Build GRCh38
rs543451561 Chr.16: 21968697-21968697 on Build GRCh38
rs775234681 Chr.16: 21968729-21968729 on Build GRCh38
rs996223114 Chr.16: 21968912-21968912 on Build GRCh38
rs150477770 Chr.16: 21968565-21968565 on Build GRCh38
rs763749530 Chr.16: 21968616-21968616 on Build GRCh38
rs375775122 Chr.16: 21968753-21968753 on Build GRCh38
rs771859641 Chr.16: 21968602-21968602 on Build GRCh38
rs908154840 Chr.16: 21968574-21968574 on Build GRCh38
rs765156352 Chr.16: 21968640-21968640 on Build GRCh38
rs200763856 Chr.16: 21968666-21968666 on Build GRCh38
rs1032728197 Chr.16: 21968873-21968873 on Build GRCh38
rs752812494 Chr.16: 21968677-21968677 on Build GRCh38
rs771671613 Chr.16: 21968718-21968718 on Build GRCh38
rs138354399 Chr.16: 21968702-21968702 on Build GRCh38
rs756334914 Chr.16: 21968678-21968678 on Build GRCh38
rs948663133 Chr.16: 21968922-21968922 on Build GRCh38
rs746691349 Chr.16: 21968736-21968736 on Build GRCh38
rs572638074 Chr.16: 21968545-21968545 on Build GRCh38
rs915833223 Chr.16: 21968918-21968918 on Build GRCh38
rs539271487 Chr.16: 21968523-21968523 on Build GRCh38
rs759117109 Chr.16: 21968817-21968817 on Build GRCh38
rs575935732 Chr.16: 21968645-21968645 on Build GRCh38
rs893819973 Chr.16: 21968910-21968910 on Build GRCh38
rs953328772 Chr.16: 21968834-21968834 on Build GRCh38
rs1055480443 Chr.16: 21968749-21968749 on Build GRCh38
rs753495328 Chr.16: 21968547-21968547 on Build GRCh38
rs976774484 Chr.16: 21968527-21968527 on Build GRCh38
rs9924083 Chr.16: 21968532-21968532 on Build GRCh38
rs544156901 Chr.16: 21968655-21968655 on Build GRCh38
rs761780092 Chr.16: 21968639-21968639 on Build GRCh38
rs779076372 Chr.16: 21968801-21968801 on Build GRCh38
rs1029131234 Chr.16: 21968541-21968541 on Build GRCh38
rs1000135137 Chr.16: 21968865-21968865 on Build GRCh38
rs909233986 Chr.16: 21968860-21968860 on Build GRCh38
rs374488411 Chr.16: 21968577-21968577 on Build GRCh38
rs941817346 Chr.16: 21968611-21968611 on Build GRCh38
rs760553949 Chr.16: 21968615-21968615 on Build GRCh38
rs771532333 Chr.16: 21968595-21968595 on Build GRCh38
rs753087368 Chr.16: 21968878-21968878 on Build GRCh38
rs371218885 Chr.16: 21968709-21968709 on Build GRCh38
rs573549977 Chr.16: 21968614-21968614 on Build GRCh38
rs563990646 Chr.16: 21968662-21968662 on Build GRCh38

View Full Product Details