Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28606623-28607156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SULT1A1
Entrez Gene ID
6817
Gene Name
sulfotransferase family 1A member 1
Gene Aliases
HAST1/HAST2, P-PST, PST, ST1A1, ST1A3, STP, STP1, TSPST1
UniGene
Hs.567342
Gene Chromosome Location
Chr.16: 28605587-28623586 on Build GRCh38
Transcripts
NM_001055.3 NM_177529.2 NM_177530.2 NM_177534.2 NM_177536.3 XM_017023604.1 XM_017023605.1 XM_017023606.1 XM_017023607.1 XM_017023608.1 XM_017023609.1 XM_017023610.1 XM_017023611.1 XM_017023612.1 XM_017023613.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5772300 c.175G>T p.V59F Chr.16: 28607041-28607041 on Build GRCh38
3508395 c.358G>A p.E120K Chr.16: 28606763-28606763 on Build GRCh38
5850615 c.317G>A p.R106H Chr.16: 28606804-28606804 on Build GRCh38
6792974 c.445C>T p.H149Y Chr.16: 28607005-28607005 on Build GRCh38
3508399 c.279C>T p.S93S Chr.16: 28606842-28606842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763511802 Chr.16: 28606866-28606866 on Build GRCh38
rs763385134 Chr.16: 28606932-28606932 on Build GRCh38
rs755497597 Chr.16: 28606752-28606752 on Build GRCh38
rs780554364 Chr.16: 28606718-28606718 on Build GRCh38
rs767411375 Chr.16: 28606712-28606712 on Build GRCh38
rs766014920 Chr.16: 28606952-28606952 on Build GRCh38
rs4149391 Chr.16: 28606650-28606650 on Build GRCh38
rs771144668 Chr.16: 28606847-28606847 on Build GRCh38
rs748940168 Chr.16: 28606763-28606763 on Build GRCh38
rs764706137 Chr.16: 28606927-28606927 on Build GRCh38
rs74752114 Chr.16: 28606747-28606747 on Build GRCh38
rs764616712 Chr.16: 28606866-28606866 on Build GRCh38
rs781598005 Chr.16: 28606722-28606722 on Build GRCh38
rs750088694 Chr.16: 28606880-28606880 on Build GRCh38
rs74016817 Chr.16: 28606751-28606751 on Build GRCh38
rs776934662 Chr.16: 28606746-28606747 on Build GRCh38
rs189186124 Chr.16: 28607037-28607037 on Build GRCh38
rs138013613 Chr.16: 28607056-28607056 on Build GRCh38
rs757965560 Chr.16: 28606942-28606942 on Build GRCh38
rs770428518 Chr.16: 28607074-28607074 on Build GRCh38
rs770600494 Chr.16: 28606764-28606764 on Build GRCh38
rs776432354 Chr.16: 28606838-28606838 on Build GRCh38
rs376199654 Chr.16: 28606815-28606815 on Build GRCh38
rs777536647 Chr.16: 28606760-28606760 on Build GRCh38
rs777965756 Chr.16: 28606814-28606814 on Build GRCh38
rs773911794 Chr.16: 28607089-28607089 on Build GRCh38
rs1042011 Chr.16: 28606999-28606999 on Build GRCh38
rs759643466 Chr.16: 28606914-28606914 on Build GRCh38
rs759716692 Chr.16: 28606849-28606849 on Build GRCh38
rs879714457 Chr.16: 28606916-28606917 on Build GRCh38
rs141878102 Chr.16: 28606957-28606957 on Build GRCh38
rs3020805 Chr.16: 28606750-28606750 on Build GRCh38
rs778957485 Chr.16: 28606714-28606714 on Build GRCh38
rs761059678 Chr.16: 28606862-28606862 on Build GRCh38
rs368643324 Chr.16: 28606743-28606743 on Build GRCh38
rs766979489 Chr.16: 28607039-28607039 on Build GRCh38
rs774493600 Chr.16: 28606736-28606736 on Build GRCh38
rs756996605 Chr.16: 28606818-28606818 on Build GRCh38
rs1043824267 Chr.16: 28606669-28606669 on Build GRCh38
rs200643489 Chr.16: 28606898-28606898 on Build GRCh38
rs760535405 Chr.16: 28606782-28606782 on Build GRCh38
rs368819430 Chr.16: 28606805-28606805 on Build GRCh38
rs761555076 Chr.16: 28606845-28606845 on Build GRCh38
rs751208498 Chr.16: 28607034-28607034 on Build GRCh38
rs767472727 Chr.16: 28606738-28606738 on Build GRCh38
rs768960535 Chr.16: 28606718-28606718 on Build GRCh38
rs140504750 Chr.16: 28606934-28606934 on Build GRCh38
rs776831209 Chr.16: 28607065-28607065 on Build GRCh38
rs11546425 Chr.16: 28607068-28607068 on Build GRCh38
rs757482973 Chr.16: 28606713-28606713 on Build GRCh38
rs780098334 Chr.16: 28606830-28606830 on Build GRCh38
rs748070353 Chr.16: 28606963-28606963 on Build GRCh38
rs747947906 Chr.16: 28606905-28606905 on Build GRCh38
rs779399958 Chr.16: 28606894-28606894 on Build GRCh38
rs201262577 Chr.16: 28606834-28606834 on Build GRCh38
rs377234606 Chr.16: 28606732-28606732 on Build GRCh38
rs754340946 Chr.16: 28606928-28606928 on Build GRCh38
rs763784335 Chr.16: 28606785-28606785 on Build GRCh38
rs747252974 Chr.16: 28607061-28607061 on Build GRCh38
rs757455705 Chr.16: 28606678-28606678 on Build GRCh38
rs763310205 Chr.16: 28607024-28607024 on Build GRCh38
rs747017564 Chr.16: 28606895-28606895 on Build GRCh38
rs767487725 Chr.16: 28606850-28606850 on Build GRCh38
rs34954534 Chr.16: 28606697-28606697 on Build GRCh38
rs771427204 Chr.16: 28606829-28606829 on Build GRCh38
rs763283299 Chr.16: 28607080-28607080 on Build GRCh38
rs781162410 Chr.16: 28606959-28606959 on Build GRCh38
rs41278156 Chr.16: 28606716-28606716 on Build GRCh38
rs768899038 Chr.16: 28607062-28607062 on Build GRCh38
rs750555116 Chr.16: 28606746-28606746 on Build GRCh38
rs773550526 Chr.16: 28606790-28606790 on Build GRCh38
rs760771748 Chr.16: 28606921-28606921 on Build GRCh38
rs759163694 Chr.16: 28607092-28607092 on Build GRCh38
rs748548173 Chr.16: 28606757-28606757 on Build GRCh38
rs2925623 Chr.16: 28607125-28607125 on Build GRCh38
rs757994677 Chr.16: 28606889-28606889 on Build GRCh38
rs777517408 Chr.16: 28606975-28606975 on Build GRCh38
rs9282862 Chr.16: 28606916-28606916 on Build GRCh38
rs116120224 Chr.16: 28606770-28606770 on Build GRCh38
rs748412001 Chr.16: 28606724-28606724 on Build GRCh38
rs777643918 Chr.16: 28607055-28607055 on Build GRCh38
rs4149389 Chr.16: 28606690-28606690 on Build GRCh38
rs200705344 Chr.16: 28606976-28606976 on Build GRCh38
rs7499134 Chr.16: 28606917-28606917 on Build GRCh38
rs148109436 Chr.16: 28606825-28606825 on Build GRCh38
rs55664460 Chr.16: 28606698-28606698 on Build GRCh38
rs745664429 Chr.16: 28606716-28606716 on Build GRCh38
rs200013410 Chr.16: 28607011-28607011 on Build GRCh38
rs763956125 Chr.16: 28607114-28607114 on Build GRCh38
rs370484353 Chr.16: 28606982-28606982 on Build GRCh38
rs376722115 Chr.16: 28606897-28606897 on Build GRCh38
rs765694696 Chr.16: 28606937-28606937 on Build GRCh38
rs751668521 Chr.16: 28606799-28606799 on Build GRCh38
rs772081539 Chr.16: 28606776-28606776 on Build GRCh38
rs754982744 Chr.16: 28606800-28606800 on Build GRCh38
rs41278158 Chr.16: 28606672-28606672 on Build GRCh38
rs1042014 Chr.16: 28606997-28606997 on Build GRCh38
rs778723776 Chr.16: 28606871-28606871 on Build GRCh38
rs766883133 Chr.16: 28606937-28606945 on Build GRCh38
rs755941191 Chr.16: 28606974-28606974 on Build GRCh38
rs756565728 Chr.16: 28606807-28606807 on Build GRCh38
rs369221760 Chr.16: 28606884-28606884 on Build GRCh38
rs201305654 Chr.16: 28607098-28607098 on Build GRCh38
rs756063880 Chr.16: 28607047-28607047 on Build GRCh38
rs376366647 Chr.16: 28606912-28606912 on Build GRCh38
rs368295553 Chr.16: 28607100-28607100 on Build GRCh38
rs55879441 Chr.16: 28606687-28606687 on Build GRCh38
rs763131488 Chr.16: 28606791-28606791 on Build GRCh38
rs766539781 Chr.16: 28606795-28606795 on Build GRCh38
rs752231054 Chr.16: 28607094-28607094 on Build GRCh38
rs765402332 Chr.16: 28606757-28606757 on Build GRCh38
rs145721404 Chr.16: 28606804-28606804 on Build GRCh38
rs1042008 Chr.16: 28607005-28607005 on Build GRCh38
rs781509368 Chr.16: 28606754-28606754 on Build GRCh38
rs755401865 Chr.16: 28606720-28606720 on Build GRCh38
rs138833811 Chr.16: 28606767-28606767 on Build GRCh38
rs76357215 Chr.16: 28606737-28606737 on Build GRCh38
rs4149388 Chr.16: 28606726-28606726 on Build GRCh38
rs754451964 Chr.16: 28606958-28606958 on Build GRCh38
rs780201783 Chr.16: 28607058-28607058 on Build GRCh38
rs373126091 Chr.16: 28606801-28606801 on Build GRCh38
rs1042000 Chr.16: 28607042-28607042 on Build GRCh38
rs746917796 Chr.16: 28606831-28606831 on Build GRCh38
rs765502732 Chr.16: 28606745-28606745 on Build GRCh38
rs757631776 Chr.16: 28606870-28606870 on Build GRCh38
rs143774918 Chr.16: 28607057-28607057 on Build GRCh38
rs754289699 Chr.16: 28606867-28606867 on Build GRCh38
rs3020806 Chr.16: 28606744-28606744 on Build GRCh38
rs756373864 Chr.16: 28606759-28606759 on Build GRCh38
rs539253610 Chr.16: 28606993-28606993 on Build GRCh38
rs753803229 Chr.16: 28607124-28607124 on Build GRCh38
rs773590770 Chr.16: 28606860-28606860 on Build GRCh38
rs386790190 Chr.16: 28606916-28606917 on Build GRCh38
rs1042019 Chr.16: 28606824-28606824 on Build GRCh38
rs887461171 Chr.16: 28606854-28606854 on Build GRCh38
rs1042005 Chr.16: 28607009-28607009 on Build GRCh38
rs867573386 Chr.16: 28607109-28607109 on Build GRCh38
rs4149390 Chr.16: 28606653-28606653 on Build GRCh38
rs775226927 Chr.16: 28606781-28606781 on Build GRCh38
rs775719447 Chr.16: 28606984-28606984 on Build GRCh38
rs147218626 Chr.16: 28607078-28607078 on Build GRCh38
rs753283272 Chr.16: 28606853-28606853 on Build GRCh38
rs750141699 Chr.16: 28606938-28606938 on Build GRCh38
rs761780927 Chr.16: 28606749-28606752 on Build GRCh38
rs1126462 Chr.16: 28606848-28606848 on Build GRCh38
rs373444797 Chr.16: 28606911-28606911 on Build GRCh38
rs766759792 Chr.16: 28607083-28607083 on Build GRCh38
rs754548195 Chr.16: 28607038-28607038 on Build GRCh38
rs769515649 Chr.16: 28606725-28606725 on Build GRCh38

View Full Product Details