Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30525063-30525566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF768
Entrez Gene ID
79724
Gene Name
zinc finger protein 768
Gene Aliases
-
UniGene
Hs.85658
Gene Chromosome Location
Chr.16: 30524001-30532367 on Build GRCh38
Transcripts
NM_024671.3 XM_017023665.1 XM_017023666.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7215780 c.645T>C p.P215P Chr.16: 30525495-30525495 on Build GRCh38
3508838 c.1074C>T p.S358S Chr.16: 30525066-30525066 on Build GRCh38
6219289 c.810C>T p.F270F Chr.16: 30525330-30525330 on Build GRCh38
7229254 c.789T>C p.C263C Chr.16: 30525351-30525351 on Build GRCh38
4744921 c.844A>G p.I282V Chr.16: 30525296-30525296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763672570 Chr.16: 30525217-30525217 on Build GRCh38
rs757510388 Chr.16: 30525386-30525386 on Build GRCh38
rs771991672 Chr.16: 30525291-30525291 on Build GRCh38
rs774869388 Chr.16: 30525296-30525296 on Build GRCh38
rs762935221 Chr.16: 30525242-30525243 on Build GRCh38
rs759860061 Chr.16: 30525515-30525515 on Build GRCh38
rs745612939 Chr.16: 30525397-30525397 on Build GRCh38
rs753224952 Chr.16: 30525439-30525439 on Build GRCh38
rs766752472 Chr.16: 30525315-30525315 on Build GRCh38
rs756242157 Chr.16: 30525362-30525362 on Build GRCh38
rs776231152 Chr.16: 30525311-30525311 on Build GRCh38
rs745818910 Chr.16: 30525289-30525289 on Build GRCh38
rs185407031 Chr.16: 30525416-30525416 on Build GRCh38
rs750841686 Chr.16: 30525243-30525243 on Build GRCh38
rs761282524 Chr.16: 30525517-30525517 on Build GRCh38
rs376531475 Chr.16: 30525497-30525497 on Build GRCh38
rs138236969 Chr.16: 30525477-30525477 on Build GRCh38
rs774644153 Chr.16: 30525427-30525427 on Build GRCh38
rs761942042 Chr.16: 30525538-30525538 on Build GRCh38
rs774409725 Chr.16: 30525146-30525146 on Build GRCh38
rs551039193 Chr.16: 30525520-30525520 on Build GRCh38
rs200448261 Chr.16: 30525543-30525543 on Build GRCh38
rs779726263 Chr.16: 30525496-30525496 on Build GRCh38
rs971102346 Chr.16: 30525085-30525085 on Build GRCh38
rs747651334 Chr.16: 30525509-30525509 on Build GRCh38
rs748190420 Chr.16: 30525272-30525272 on Build GRCh38
rs141607219 Chr.16: 30525087-30525087 on Build GRCh38
rs139435151 Chr.16: 30525516-30525516 on Build GRCh38
rs754042148 Chr.16: 30525537-30525537 on Build GRCh38
rs758178426 Chr.16: 30525470-30525470 on Build GRCh38
rs755462642 Chr.16: 30525255-30525255 on Build GRCh38
rs774160369 Chr.16: 30525288-30525288 on Build GRCh38
rs561390852 Chr.16: 30525417-30525417 on Build GRCh38
rs768181947 Chr.16: 30525412-30525412 on Build GRCh38
rs545507206 Chr.16: 30525273-30525273 on Build GRCh38
rs752033647 Chr.16: 30525249-30525249 on Build GRCh38
rs750044408 Chr.16: 30525453-30525453 on Build GRCh38
rs749554021 Chr.16: 30525379-30525379 on Build GRCh38
rs746559698 Chr.16: 30525409-30525409 on Build GRCh38
rs759127147 Chr.16: 30525165-30525165 on Build GRCh38
rs1056488333 Chr.16: 30525501-30525501 on Build GRCh38
rs770081378 Chr.16: 30525084-30525084 on Build GRCh38
rs35230431 Chr.16: 30525279-30525280 on Build GRCh38
rs771051847 Chr.16: 30525104-30525104 on Build GRCh38
rs377230108 Chr.16: 30525261-30525261 on Build GRCh38
rs370166983 Chr.16: 30525282-30525282 on Build GRCh38
rs769389051 Chr.16: 30525510-30525510 on Build GRCh38
rs778589873 Chr.16: 30525388-30525388 on Build GRCh38
rs750231065 Chr.16: 30525468-30525468 on Build GRCh38
rs866153106 Chr.16: 30525436-30525436 on Build GRCh38
rs780017304 Chr.16: 30525401-30525401 on Build GRCh38
rs778129017 Chr.16: 30525276-30525276 on Build GRCh38
rs944747783 Chr.16: 30525378-30525378 on Build GRCh38
rs769419547 Chr.16: 30525418-30525418 on Build GRCh38
rs779194247 Chr.16: 30525459-30525459 on Build GRCh38
rs145451015 Chr.16: 30525192-30525192 on Build GRCh38
rs202117110 Chr.16: 30525096-30525096 on Build GRCh38
rs749145456 Chr.16: 30525280-30525280 on Build GRCh38
rs758736587 Chr.16: 30525244-30525244 on Build GRCh38
rs773143945 Chr.16: 30525095-30525095 on Build GRCh38
rs766970842 Chr.16: 30525180-30525180 on Build GRCh38
rs3751847 Chr.16: 30525351-30525351 on Build GRCh38
rs189981520 Chr.16: 30525530-30525530 on Build GRCh38
rs755443193 Chr.16: 30525342-30525342 on Build GRCh38
rs750686634 Chr.16: 30525545-30525545 on Build GRCh38
rs761411417 Chr.16: 30525314-30525314 on Build GRCh38
rs867070538 Chr.16: 30525158-30525158 on Build GRCh38
rs768062322 Chr.16: 30525350-30525350 on Build GRCh38
rs528697308 Chr.16: 30525434-30525434 on Build GRCh38
rs778181999 Chr.16: 30525371-30525371 on Build GRCh38
rs766907897 Chr.16: 30525246-30525246 on Build GRCh38
rs546832648 Chr.16: 30525437-30525437 on Build GRCh38
rs763782885 Chr.16: 30525309-30525309 on Build GRCh38
rs922240785 Chr.16: 30525368-30525368 on Build GRCh38
rs761223651 Chr.16: 30525451-30525451 on Build GRCh38
rs926933624 Chr.16: 30525228-30525228 on Build GRCh38
rs765415245 Chr.16: 30525540-30525540 on Build GRCh38
rs1043056333 Chr.16: 30525438-30525438 on Build GRCh38
rs1000200368 Chr.16: 30525419-30525419 on Build GRCh38
rs1027732100 Chr.16: 30525528-30525528 on Build GRCh38
rs530906636 Chr.16: 30525400-30525400 on Build GRCh38
rs369207286 Chr.16: 30525514-30525514 on Build GRCh38
rs775019894 Chr.16: 30525182-30525182 on Build GRCh38
rs751893545 Chr.16: 30525317-30525317 on Build GRCh38
rs1024069610 Chr.16: 30525445-30525445 on Build GRCh38
rs371944438 Chr.16: 30525486-30525486 on Build GRCh38
rs753914674 Chr.16: 30525457-30525457 on Build GRCh38
rs746966256 Chr.16: 30525482-30525482 on Build GRCh38
rs780683574 Chr.16: 30525502-30525502 on Build GRCh38
rs760162989 Chr.16: 30525299-30525299 on Build GRCh38

View Full Product Details