Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31216643-31217139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PYDC1
Entrez Gene ID
260434
Gene Name
pyrin domain containing 1
Gene Aliases
ASC2, POP1, PYC1, cPOP1
UniGene
Hs.58314
Gene Chromosome Location
Chr.16: 31215962-31217093 on Build GRCh38
Transcripts
NM_152901.3 NM_001008274.3 XM_017023254.1

Gene Symbol
TRIM72
Entrez Gene ID
493829
Gene Name
tripartite motif containing 72
Gene Aliases
MG53
UniGene
Hs.661859
Gene Chromosome Location
Chr.16: 31214021-31225656 on Build GRCh38
Transcripts
NM_152901.3 NM_001008274.3 XM_017023254.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6555644 c.240G>A p.E80E Chr.16: 31216789-31216789 on Build GRCh38
6555645 c.122G>A p.R41Q Chr.16: 31216907-31216907 on Build GRCh38
5897122 c.176C>T p.S59F Chr.16: 31216853-31216853 on Build GRCh38
6277342 c.91C>T p.P31S Chr.16: 31216938-31216938 on Build GRCh38
6760736 c.131T>C p.L44P Chr.16: 31216898-31216898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375157236 Chr.16: 31216746-31216746 on Build GRCh38
rs143371283 Chr.16: 31216796-31216796 on Build GRCh38
rs544084252 Chr.16: 31217106-31217106 on Build GRCh38
rs577520252 Chr.16: 31216742-31216742 on Build GRCh38
rs747858063 Chr.16: 31216994-31216994 on Build GRCh38
rs919866264 Chr.16: 31216964-31216964 on Build GRCh38
rs74015098 Chr.16: 31216897-31216897 on Build GRCh38
rs1012320148 Chr.16: 31216799-31216799 on Build GRCh38
rs865966029 Chr.16: 31216919-31216919 on Build GRCh38
rs773582347 Chr.16: 31216709-31216709 on Build GRCh38
rs752162442 Chr.16: 31216824-31216824 on Build GRCh38
rs778842963 Chr.16: 31216752-31216752 on Build GRCh38
rs768050384 Chr.16: 31216777-31216777 on Build GRCh38
rs556939015 Chr.16: 31216801-31216801 on Build GRCh38
rs776142717 Chr.16: 31216783-31216783 on Build GRCh38
rs756131042 Chr.16: 31216733-31216733 on Build GRCh38
rs377601641 Chr.16: 31216809-31216809 on Build GRCh38
rs372347969 Chr.16: 31216937-31216937 on Build GRCh38
rs779403119 Chr.16: 31217027-31217027 on Build GRCh38
rs748819441 Chr.16: 31217008-31217008 on Build GRCh38
rs780733594 Chr.16: 31216911-31216911 on Build GRCh38
rs758177015 Chr.16: 31216881-31216881 on Build GRCh38
rs753905315 Chr.16: 31216833-31216833 on Build GRCh38
rs1053894172 Chr.16: 31216778-31216778 on Build GRCh38
rs755421162 Chr.16: 31216724-31216724 on Build GRCh38
rs902521216 Chr.16: 31216748-31216748 on Build GRCh38
rs973998160 Chr.16: 31216962-31216962 on Build GRCh38
rs757358945 Chr.16: 31216848-31216848 on Build GRCh38
rs140681493 Chr.16: 31216684-31216684 on Build GRCh38
rs936641351 Chr.16: 31216763-31216763 on Build GRCh38
rs775035208 Chr.16: 31216802-31216802 on Build GRCh38
rs952070930 Chr.16: 31216967-31216967 on Build GRCh38
rs200629994 Chr.16: 31216731-31216731 on Build GRCh38
rs771894790 Chr.16: 31216764-31216764 on Build GRCh38
rs756337396 Chr.16: 31216806-31216806 on Build GRCh38
rs372785932 Chr.16: 31216672-31216672 on Build GRCh38
rs375073317 Chr.16: 31216941-31216941 on Build GRCh38
rs560556446 Chr.16: 31216927-31216927 on Build GRCh38
rs775893534 Chr.16: 31217016-31217016 on Build GRCh38
rs1041369316 Chr.16: 31217110-31217110 on Build GRCh38
rs780870987 Chr.16: 31216992-31216992 on Build GRCh38
rs757128378 Chr.16: 31216745-31216745 on Build GRCh38
rs1015149556 Chr.16: 31216858-31216858 on Build GRCh38
rs961082709 Chr.16: 31216870-31216870 on Build GRCh38
rs778791543 Chr.16: 31216850-31216850 on Build GRCh38
rs370295635 Chr.16: 31216826-31216826 on Build GRCh38
rs773221445 Chr.16: 31217039-31217039 on Build GRCh38
rs754765294 Chr.16: 31216987-31216987 on Build GRCh38
rs747659143 Chr.16: 31216918-31216918 on Build GRCh38
rs761070959 Chr.16: 31217018-31217018 on Build GRCh38
rs369666205 Chr.16: 31217067-31217067 on Build GRCh38
rs769473593 Chr.16: 31216920-31216920 on Build GRCh38
rs762091328 Chr.16: 31217031-31217031 on Build GRCh38
rs766304046 Chr.16: 31217054-31217054 on Build GRCh38
rs767279385 Chr.16: 31217065-31217065 on Build GRCh38
rs932517832 Chr.16: 31216688-31216688 on Build GRCh38
rs373491537 Chr.16: 31217070-31217070 on Build GRCh38
rs746735109 Chr.16: 31216772-31216772 on Build GRCh38
rs150364107 Chr.16: 31217026-31217026 on Build GRCh38
rs879408321 Chr.16: 31217092-31217092 on Build GRCh38
rs760184701 Chr.16: 31216929-31216929 on Build GRCh38
rs765452497 Chr.16: 31217037-31217037 on Build GRCh38
rs749331770 Chr.16: 31216744-31216744 on Build GRCh38
rs546281460 Chr.16: 31217021-31217021 on Build GRCh38
rs749355448 Chr.16: 31216903-31216904 on Build GRCh38
rs201482571 Chr.16: 31216819-31216819 on Build GRCh38
rs767002234 Chr.16: 31216713-31216713 on Build GRCh38
rs202126650 Chr.16: 31216712-31216712 on Build GRCh38
rs777350501 Chr.16: 31216996-31216996 on Build GRCh38
rs772581790 Chr.16: 31217009-31217009 on Build GRCh38
rs763047132 Chr.16: 31217046-31217046 on Build GRCh38
rs750411206 Chr.16: 31216875-31216875 on Build GRCh38
rs1054702349 Chr.16: 31217056-31217056 on Build GRCh38
rs759953624 Chr.16: 31216806-31216806 on Build GRCh38
rs368720244 Chr.16: 31216909-31216909 on Build GRCh38
rs751918609 Chr.16: 31216723-31216723 on Build GRCh38
rs769420582 Chr.16: 31216995-31216995 on Build GRCh38
rs868032860 Chr.16: 31216905-31216905 on Build GRCh38
rs900687330 Chr.16: 31216821-31216821 on Build GRCh38
rs778023939 Chr.16: 31216895-31216914 on Build GRCh38
rs775103425 Chr.16: 31216770-31216770 on Build GRCh38
rs750358061 Chr.16: 31216969-31216969 on Build GRCh38
rs758267007 Chr.16: 31216978-31216978 on Build GRCh38
rs765360475 Chr.16: 31216968-31216968 on Build GRCh38
rs146708916 Chr.16: 31217072-31217072 on Build GRCh38
rs751355576 Chr.16: 31216779-31216780 on Build GRCh38
rs746787173 Chr.16: 31216898-31216898 on Build GRCh38
rs368309851 Chr.16: 31217055-31217055 on Build GRCh38
rs73532241 Chr.16: 31216697-31216697 on Build GRCh38
rs533900923 Chr.16: 31216730-31216730 on Build GRCh38
rs770946351 Chr.16: 31216945-31216945 on Build GRCh38
rs754047946 Chr.16: 31216942-31216942 on Build GRCh38
rs754784977 Chr.16: 31217062-31217062 on Build GRCh38
rs745624026 Chr.16: 31216755-31216755 on Build GRCh38
rs751323039 Chr.16: 31216983-31216983 on Build GRCh38
rs958802795 Chr.16: 31216943-31216943 on Build GRCh38
rs140415359 Chr.16: 31216933-31216933 on Build GRCh38
rs1005139861 Chr.16: 31216840-31216840 on Build GRCh38
rs779872702 Chr.16: 31216979-31216979 on Build GRCh38

View Full Product Details