Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50673415-50673921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX20
Entrez Gene ID
124460
Gene Name
sorting nexin 20
Gene Aliases
SLIC1
UniGene
Hs.460777
Gene Chromosome Location
Chr.16: 50666300-50681353 on Build GRCh38
Transcripts
NM_001144972.1 NM_153337.2 NM_182854.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1227060 c.904C>T p.R302* Chr.16: 50673453-50673453 on Build GRCh38
4593315 c.723C>A p.D241E Chr.16: 50673634-50673634 on Build GRCh38
6786485 c.725G>A p.R242H Chr.16: 50673632-50673632 on Build GRCh38
3510073 c.571G>A p.E191K Chr.16: 50673786-50673786 on Build GRCh38
3510071 c.753G>A p.E251E Chr.16: 50673604-50673604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374773567 Chr.16: 50673838-50673838 on Build GRCh38
rs762265145 Chr.16: 50673443-50673443 on Build GRCh38
rs770818368 Chr.16: 50673879-50673879 on Build GRCh38
rs534067357 Chr.16: 50673630-50673630 on Build GRCh38
rs772786803 Chr.16: 50673873-50673873 on Build GRCh38
rs760316629 Chr.16: 50673876-50673876 on Build GRCh38
rs766233288 Chr.16: 50673815-50673815 on Build GRCh38
rs761260114 Chr.16: 50673504-50673504 on Build GRCh38
rs774896005 Chr.16: 50673628-50673628 on Build GRCh38
rs748469553 Chr.16: 50673585-50673585 on Build GRCh38
rs1007232450 Chr.16: 50673633-50673633 on Build GRCh38
rs748499124 Chr.16: 50673494-50673494 on Build GRCh38
rs773792568 Chr.16: 50673503-50673503 on Build GRCh38
rs368454557 Chr.16: 50673548-50673548 on Build GRCh38
rs746355889 Chr.16: 50673783-50673783 on Build GRCh38
rs779141373 Chr.16: 50673583-50673583 on Build GRCh38
rs777479037 Chr.16: 50673463-50673463 on Build GRCh38
rs763751801 Chr.16: 50673518-50673518 on Build GRCh38
rs781049068 Chr.16: 50673478-50673478 on Build GRCh38
rs902879100 Chr.16: 50673614-50673614 on Build GRCh38
rs761491948 Chr.16: 50673788-50673788 on Build GRCh38
rs372727469 Chr.16: 50673849-50673849 on Build GRCh38
rs969526797 Chr.16: 50673679-50673679 on Build GRCh38
rs768031335 Chr.16: 50673445-50673445 on Build GRCh38
rs755179588 Chr.16: 50673560-50673560 on Build GRCh38
rs770169868 Chr.16: 50673855-50673855 on Build GRCh38
rs762464935 Chr.16: 50673642-50673642 on Build GRCh38
rs762369910 Chr.16: 50673711-50673711 on Build GRCh38
rs545584172 Chr.16: 50673808-50673808 on Build GRCh38
rs940679203 Chr.16: 50673455-50673455 on Build GRCh38
rs768889840 Chr.16: 50673490-50673490 on Build GRCh38
rs7198294 Chr.16: 50673871-50673871 on Build GRCh38
rs758353023 Chr.16: 50673488-50673489 on Build GRCh38
rs762555491 Chr.16: 50673513-50673513 on Build GRCh38
rs375118669 Chr.16: 50673521-50673521 on Build GRCh38
rs982196607 Chr.16: 50673699-50673699 on Build GRCh38
rs576966789 Chr.16: 50673673-50673673 on Build GRCh38
rs750413433 Chr.16: 50673544-50673544 on Build GRCh38
rs772722665 Chr.16: 50673500-50673500 on Build GRCh38
rs774119531 Chr.16: 50673657-50673657 on Build GRCh38
rs140872915 Chr.16: 50673484-50673484 on Build GRCh38
rs773010904 Chr.16: 50673801-50673801 on Build GRCh38
rs554052528 Chr.16: 50673660-50673660 on Build GRCh38
rs920530340 Chr.16: 50673753-50673753 on Build GRCh38
rs940726034 Chr.16: 50673746-50673746 on Build GRCh38
rs930507012 Chr.16: 50673777-50673777 on Build GRCh38
rs747339593 Chr.16: 50673751-50673751 on Build GRCh38
rs751323411 Chr.16: 50673520-50673520 on Build GRCh38
rs867890625 Chr.16: 50673510-50673510 on Build GRCh38
rs763657317 Chr.16: 50673643-50673643 on Build GRCh38
rs889072534 Chr.16: 50673578-50673578 on Build GRCh38
rs771534628 Chr.16: 50673625-50673625 on Build GRCh38
rs915500358 Chr.16: 50673460-50673460 on Build GRCh38
rs752464534 Chr.16: 50673452-50673452 on Build GRCh38
rs751376297 Chr.16: 50673860-50673861 on Build GRCh38
rs998985483 Chr.16: 50673672-50673672 on Build GRCh38
rs776303744 Chr.16: 50673481-50673481 on Build GRCh38
rs778514926 Chr.16: 50673861-50673861 on Build GRCh38
rs759377980 Chr.16: 50673822-50673822 on Build GRCh38
rs776272427 Chr.16: 50673885-50673885 on Build GRCh38
rs868799465 Chr.16: 50673690-50673690 on Build GRCh38
rs754770486 Chr.16: 50673891-50673891 on Build GRCh38
rs200752656 Chr.16: 50673895-50673895 on Build GRCh38
rs201531002 Chr.16: 50673752-50673752 on Build GRCh38
rs751339391 Chr.16: 50673467-50673467 on Build GRCh38
rs57853215 Chr.16: 50673450-50673450 on Build GRCh38
rs745950787 Chr.16: 50673479-50673479 on Build GRCh38
rs747606360 Chr.16: 50673870-50673870 on Build GRCh38
rs761442088 Chr.16: 50673667-50673667 on Build GRCh38
rs749518961 Chr.16: 50673488-50673488 on Build GRCh38
rs752797483 Chr.16: 50673824-50673824 on Build GRCh38
rs1036310310 Chr.16: 50673471-50673471 on Build GRCh38
rs775044496 Chr.16: 50673511-50673511 on Build GRCh38
rs755022847 Chr.16: 50673691-50673691 on Build GRCh38
rs749346547 Chr.16: 50673557-50673557 on Build GRCh38
rs758057584 Chr.16: 50673457-50673457 on Build GRCh38
rs772636555 Chr.16: 50673590-50673590 on Build GRCh38
rs757162450 Chr.16: 50673468-50673468 on Build GRCh38
rs766326460 Chr.16: 50673705-50673706 on Build GRCh38
rs767345248 Chr.16: 50673671-50673671 on Build GRCh38
rs753879529 Chr.16: 50673686-50673686 on Build GRCh38
rs778323232 Chr.16: 50673750-50673750 on Build GRCh38
rs781674959 Chr.16: 50673768-50673768 on Build GRCh38
rs764990101 Chr.16: 50673451-50673451 on Build GRCh38
rs1048975860 Chr.16: 50673563-50673563 on Build GRCh38
rs758671707 Chr.16: 50673735-50673735 on Build GRCh38
rs778857780 Chr.16: 50673706-50673706 on Build GRCh38
rs766257486 Chr.16: 50673685-50673685 on Build GRCh38
rs919062067 Chr.16: 50673535-50673535 on Build GRCh38
rs780144839 Chr.16: 50673555-50673555 on Build GRCh38
rs778094287 Chr.16: 50673842-50673842 on Build GRCh38
rs372280737 Chr.16: 50673491-50673491 on Build GRCh38
rs944507574 Chr.16: 50673829-50673829 on Build GRCh38
rs771671749 Chr.16: 50673791-50673791 on Build GRCh38
rs769928339 Chr.16: 50673482-50673482 on Build GRCh38
rs746432403 Chr.16: 50673850-50673850 on Build GRCh38
rs773425123 Chr.16: 50673817-50673817 on Build GRCh38
rs368969544 Chr.16: 50673446-50673446 on Build GRCh38
rs201673911 Chr.16: 50673597-50673597 on Build GRCh38
rs764898274 Chr.16: 50673508-50673508 on Build GRCh38
rs545833578 Chr.16: 50673749-50673749 on Build GRCh38
rs773707336 Chr.16: 50673787-50673787 on Build GRCh38
rs768270570 Chr.16: 50673784-50673784 on Build GRCh38
rs562563894 Chr.16: 50673843-50673843 on Build GRCh38
rs113387142 Chr.16: 50673542-50673542 on Build GRCh38
rs990098618 Chr.16: 50673453-50673453 on Build GRCh38
rs957056780 Chr.16: 50673701-50673701 on Build GRCh38

View Full Product Details