Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57985256-57985751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEPP
Entrez Gene ID
374739
Gene Name
testis, prostate and placenta expressed
Gene Aliases
-
UniGene
Hs.132976
Gene Chromosome Location
Chr.16: 57976435-57988116 on Build GRCh38
Transcripts
NM_199046.2 NM_199456.2 XM_017023218.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6427390 c.690G>T p.Q230H Chr.16: 57985403-57985403 on Build GRCh38
2992572 c.833+3A>G p.? Chr.16: 57985549-57985549 on Build GRCh38
4129151 c.779G>A p.S260N Chr.16: 57985492-57985492 on Build GRCh38
2992571 c.743G>A p.R248H Chr.16: 57985456-57985456 on Build GRCh38
5363756 c.773G>A p.C258Y Chr.16: 57985486-57985486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575779645 Chr.16: 57985458-57985458 on Build GRCh38
rs769756941 Chr.16: 57985375-57985375 on Build GRCh38
rs768503790 Chr.16: 57985356-57985357 on Build GRCh38
rs1047826657 Chr.16: 57985419-57985419 on Build GRCh38
rs774371731 Chr.16: 57985391-57985391 on Build GRCh38
rs978525340 Chr.16: 57985345-57985345 on Build GRCh38
rs1026007100 Chr.16: 57985699-57985699 on Build GRCh38
rs770956594 Chr.16: 57985538-57985538 on Build GRCh38
rs554741652 Chr.16: 57985708-57985708 on Build GRCh38
rs566311990 Chr.16: 57985623-57985623 on Build GRCh38
rs1005882332 Chr.16: 57985612-57985612 on Build GRCh38
rs767288689 Chr.16: 57985563-57985563 on Build GRCh38
rs576150360 Chr.16: 57985342-57985342 on Build GRCh38
rs778542622 Chr.16: 57985518-57985518 on Build GRCh38
rs2304486 Chr.16: 57985349-57985349 on Build GRCh38
rs768939142 Chr.16: 57985389-57985389 on Build GRCh38
rs752388065 Chr.16: 57985355-57985355 on Build GRCh38
rs374687332 Chr.16: 57985374-57985374 on Build GRCh38
rs560594217 Chr.16: 57985433-57985433 on Build GRCh38
rs761744244 Chr.16: 57985301-57985301 on Build GRCh38
rs780174769 Chr.16: 57985514-57985514 on Build GRCh38
rs777143832 Chr.16: 57985543-57985543 on Build GRCh38
rs970746709 Chr.16: 57985344-57985344 on Build GRCh38
rs923908887 Chr.16: 57985477-57985477 on Build GRCh38
rs766887230 Chr.16: 57985484-57985484 on Build GRCh38
rs765470243 Chr.16: 57985493-57985493 on Build GRCh38
rs773276401 Chr.16: 57985464-57985464 on Build GRCh38
rs9934227 Chr.16: 57985492-57985492 on Build GRCh38
rs753858373 Chr.16: 57985574-57985574 on Build GRCh38
rs755621426 Chr.16: 57985511-57985511 on Build GRCh38
rs147821456 Chr.16: 57985459-57985459 on Build GRCh38
rs978218995 Chr.16: 57985468-57985468 on Build GRCh38
rs754854508 Chr.16: 57985517-57985517 on Build GRCh38
rs747094701 Chr.16: 57985532-57985532 on Build GRCh38
rs766797583 Chr.16: 57985408-57985408 on Build GRCh38
rs761582049 Chr.16: 57985560-57985560 on Build GRCh38
rs376372652 Chr.16: 57985441-57985441 on Build GRCh38
rs959207083 Chr.16: 57985323-57985323 on Build GRCh38
rs748470488 Chr.16: 57985519-57985519 on Build GRCh38
rs770229974 Chr.16: 57985558-57985558 on Build GRCh38
rs749585071 Chr.16: 57985382-57985382 on Build GRCh38
rs746540934 Chr.16: 57985358-57985358 on Build GRCh38
rs369142450 Chr.16: 57985348-57985348 on Build GRCh38
rs201813826 Chr.16: 57985571-57985571 on Build GRCh38
rs200222278 Chr.16: 57985368-57985368 on Build GRCh38
rs767450827 Chr.16: 57985393-57985393 on Build GRCh38
rs763241303 Chr.16: 57985290-57985290 on Build GRCh38
rs752450425 Chr.16: 57985598-57985598 on Build GRCh38
rs775873787 Chr.16: 57985281-57985281 on Build GRCh38
rs756825037 Chr.16: 57985495-57985495 on Build GRCh38
rs1041260004 Chr.16: 57985567-57985567 on Build GRCh38
rs199701351 Chr.16: 57985549-57985549 on Build GRCh38
rs761988788 Chr.16: 57985392-57985392 on Build GRCh38
rs771317662 Chr.16: 57985549-57985550 on Build GRCh38
rs754867558 Chr.16: 57985586-57985586 on Build GRCh38
rs777916286 Chr.16: 57985531-57985531 on Build GRCh38
rs756813925 Chr.16: 57985435-57985435 on Build GRCh38
rs572054542 Chr.16: 57985284-57985284 on Build GRCh38
rs886065932 Chr.16: 57985599-57985599 on Build GRCh38
rs765446238 Chr.16: 57985462-57985462 on Build GRCh38
rs750894417 Chr.16: 57985319-57985319 on Build GRCh38
rs146257859 Chr.16: 57985455-57985455 on Build GRCh38
rs942845779 Chr.16: 57985370-57985370 on Build GRCh38
rs776251963 Chr.16: 57985397-57985399 on Build GRCh38
rs374742693 Chr.16: 57985318-57985318 on Build GRCh38
rs780823079 Chr.16: 57985436-57985436 on Build GRCh38
rs774660170 Chr.16: 57985456-57985456 on Build GRCh38
rs201219668 Chr.16: 57985278-57985278 on Build GRCh38
rs376915152 Chr.16: 57985453-57985453 on Build GRCh38
rs200939193 Chr.16: 57985379-57985379 on Build GRCh38
rs780049741 Chr.16: 57985452-57985452 on Build GRCh38
rs754190012 Chr.16: 57985409-57985409 on Build GRCh38
rs775709851 Chr.16: 57985559-57985559 on Build GRCh38
rs889581405 Chr.16: 57985621-57985621 on Build GRCh38
rs542525208 Chr.16: 57985674-57985674 on Build GRCh38
rs756113605 Chr.16: 57985443-57985443 on Build GRCh38
rs777055561 Chr.16: 57985489-57985489 on Build GRCh38
rs761230306 Chr.16: 57985470-57985470 on Build GRCh38
rs547674673 Chr.16: 57985600-57985600 on Build GRCh38
rs962875643 Chr.16: 57985643-57985644 on Build GRCh38
rs764118083 Chr.16: 57985291-57985291 on Build GRCh38
rs1038798247 Chr.16: 57985377-57985377 on Build GRCh38
rs554603938 Chr.16: 57985338-57985338 on Build GRCh38
rs753674239 Chr.16: 57985516-57985516 on Build GRCh38
rs2304487 Chr.16: 57985711-57985711 on Build GRCh38
rs751650634 Chr.16: 57985300-57985300 on Build GRCh38
rs751269919 Chr.16: 57985494-57985494 on Build GRCh38
rs895097191 Chr.16: 57985634-57985634 on Build GRCh38
rs777207612 Chr.16: 57985357-57985357 on Build GRCh38
rs140883587 Chr.16: 57985434-57985434 on Build GRCh38
rs755238557 Chr.16: 57985430-57985430 on Build GRCh38
rs758043944 Chr.16: 57985356-57985356 on Build GRCh38
rs371867703 Chr.16: 57985606-57985606 on Build GRCh38
rs772172993 Chr.16: 57985526-57985526 on Build GRCh38
rs770278376 Chr.16: 57985361-57985361 on Build GRCh38
rs757845520 Chr.16: 57985486-57985486 on Build GRCh38
rs761140068 Chr.16: 57985403-57985403 on Build GRCh38
rs780374952 Chr.16: 57985353-57985353 on Build GRCh38
rs200645231 Chr.16: 57985573-57985573 on Build GRCh38
rs761645352 Chr.16: 57985492-57985492 on Build GRCh38
rs773757475 Chr.16: 57985398-57985398 on Build GRCh38
rs369893338 Chr.16: 57985496-57985496 on Build GRCh38
rs536200369 Chr.16: 57985337-57985337 on Build GRCh38
rs778827252 Chr.16: 57985589-57985589 on Build GRCh38

View Full Product Details