Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66808300-66808801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAE1
Entrez Gene ID
8883
Gene Name
NEDD8 activating enzyme E1 subunit 1
Gene Aliases
A-116A10.1, APPBP1, HPP1, ula-1
UniGene
Hs.460978
Gene Chromosome Location
Chr.16: 66802878-66830977 on Build GRCh38
Transcripts
NM_001018159.1 NM_001018160.1 NM_001286500.1 NM_003905.3 XM_005256215.1 XM_011523422.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6144996 c.1297G>C p.D433H Chr.16: 66808554-66808554 on Build GRCh38
6721688 c.1288C>T p.R430W Chr.16: 66808563-66808563 on Build GRCh38
1640518 c.1238-8_1238-7insT p.? Chr.16: 66808620-66808621 on Build GRCh38
5513205 c.1238-9delT p.? Chr.16: 66808622-66808622 on Build GRCh38
4061691 c.1306C>A p.H436N Chr.16: 66808545-66808545 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766440475 Chr.16: 66808639-66808639 on Build GRCh38
rs1039017025 Chr.16: 66808761-66808761 on Build GRCh38
rs776646555 Chr.16: 66808555-66808555 on Build GRCh38
rs916166341 Chr.16: 66808408-66808408 on Build GRCh38
rs763861962 Chr.16: 66808633-66808634 on Build GRCh38
rs568489849 Chr.16: 66808767-66808767 on Build GRCh38
rs866919019 Chr.16: 66808648-66808648 on Build GRCh38
rs761064262 Chr.16: 66808555-66808564 on Build GRCh38
rs139404942 Chr.16: 66808587-66808587 on Build GRCh38
rs772379487 Chr.16: 66808534-66808534 on Build GRCh38
rs531963847 Chr.16: 66808398-66808398 on Build GRCh38
rs753118465 Chr.16: 66808548-66808551 on Build GRCh38
rs363205 Chr.16: 66808776-66808776 on Build GRCh38
rs363168 Chr.16: 66808774-66808774 on Build GRCh38
rs186468179 Chr.16: 66808759-66808759 on Build GRCh38
rs779751094 Chr.16: 66808470-66808470 on Build GRCh38
rs754549391 Chr.16: 66808654-66808654 on Build GRCh38
rs754637368 Chr.16: 66808483-66808483 on Build GRCh38
rs753332794 Chr.16: 66808645-66808645 on Build GRCh38
rs578205054 Chr.16: 66808688-66808688 on Build GRCh38
rs565276191 Chr.16: 66808529-66808529 on Build GRCh38
rs982293880 Chr.16: 66808333-66808333 on Build GRCh38
rs767281309 Chr.16: 66808623-66808623 on Build GRCh38
rs1034764319 Chr.16: 66808384-66808384 on Build GRCh38
rs770980813 Chr.16: 66808549-66808549 on Build GRCh38
rs893895185 Chr.16: 66808746-66808746 on Build GRCh38
rs759639950 Chr.16: 66808560-66808560 on Build GRCh38
rs545304045 Chr.16: 66808472-66808472 on Build GRCh38
rs773767225 Chr.16: 66808542-66808542 on Build GRCh38
rs890542042 Chr.16: 66808722-66808722 on Build GRCh38
rs1052314428 Chr.16: 66808620-66808620 on Build GRCh38
rs973555811 Chr.16: 66808532-66808532 on Build GRCh38
rs775586914 Chr.16: 66808601-66808601 on Build GRCh38
rs527972853 Chr.16: 66808626-66808626 on Build GRCh38
rs999729085 Chr.16: 66808718-66808718 on Build GRCh38
rs376356697 Chr.16: 66808543-66808543 on Build GRCh38
rs760698544 Chr.16: 66808736-66808736 on Build GRCh38
rs112307898 Chr.16: 66808528-66808528 on Build GRCh38
rs1010803145 Chr.16: 66808771-66808771 on Build GRCh38
rs530951488 Chr.16: 66808368-66808369 on Build GRCh38
rs761651914 Chr.16: 66808606-66808606 on Build GRCh38
rs929477022 Chr.16: 66808553-66808553 on Build GRCh38
rs543116587 Chr.16: 66808328-66808328 on Build GRCh38
rs75370624 Chr.16: 66808392-66808392 on Build GRCh38
rs894106612 Chr.16: 66808545-66808545 on Build GRCh38
rs942253451 Chr.16: 66808434-66808434 on Build GRCh38
rs748488879 Chr.16: 66808533-66808533 on Build GRCh38
rs941066503 Chr.16: 66808393-66808393 on Build GRCh38
rs756168849 Chr.16: 66808630-66808630 on Build GRCh38
rs778616877 Chr.16: 66808662-66808662 on Build GRCh38

View Full Product Details