Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 74300665-74301199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMD7
Entrez Gene ID
5713
Gene Name
proteasome 26S subunit, non-ATPase 7
Gene Aliases
MOV34, P40, Rpn8, S12
UniGene
Hs.440604
Gene Chromosome Location
Chr.16: 74296775-74306288 on Build GRCh38
Transcripts
NM_002811.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
973598 c.167T>C p.V56A Chr.16: 74301052-74301052 on Build GRCh38
5773727 c.258T>A p.N86K Chr.16: 74301143-74301143 on Build GRCh38
1730821 c.236A>G p.Y79C Chr.16: 74301121-74301121 on Build GRCh38
4471288 c.170C>T p.P57L Chr.16: 74301055-74301055 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759055013 Chr.16: 74301014-74301014 on Build GRCh38
rs745915653 Chr.16: 74301027-74301030 on Build GRCh38
rs576988646 Chr.16: 74301024-74301028 on Build GRCh38
rs988476898 Chr.16: 74300911-74300911 on Build GRCh38
rs1019812771 Chr.16: 74300892-74300892 on Build GRCh38
rs772704482 Chr.16: 74301004-74301004 on Build GRCh38
rs781726127 Chr.16: 74301143-74301143 on Build GRCh38
rs1051133535 Chr.16: 74301134-74301134 on Build GRCh38
rs570674709 Chr.16: 74300924-74300924 on Build GRCh38
rs776381295 Chr.16: 74301013-74301013 on Build GRCh38
rs765123498 Chr.16: 74301077-74301077 on Build GRCh38
rs769607012 Chr.16: 74301165-74301165 on Build GRCh38
rs777971936 Chr.16: 74301007-74301007 on Build GRCh38
rs982694789 Chr.16: 74300722-74300722 on Build GRCh38
rs761784390 Chr.16: 74301046-74301046 on Build GRCh38
rs562836191 Chr.16: 74300780-74300780 on Build GRCh38
rs965269497 Chr.16: 74300733-74300733 on Build GRCh38
rs775116853 Chr.16: 74301167-74301167 on Build GRCh38
rs189790319 Chr.16: 74300972-74300972 on Build GRCh38
rs563182701 Chr.16: 74301031-74301031 on Build GRCh38
rs764140182 Chr.16: 74301054-74301054 on Build GRCh38
rs747458591 Chr.16: 74301008-74301008 on Build GRCh38
rs768491215 Chr.16: 74300968-74300968 on Build GRCh38
rs934921595 Chr.16: 74300822-74300822 on Build GRCh38
rs745508996 Chr.16: 74301164-74301164 on Build GRCh38
rs775204865 Chr.16: 74301022-74301022 on Build GRCh38
rs373632849 Chr.16: 74300997-74300997 on Build GRCh38
rs905963320 Chr.16: 74300912-74300912 on Build GRCh38
rs746836036 Chr.16: 74300869-74300869 on Build GRCh38
rs955772957 Chr.16: 74300905-74300905 on Build GRCh38
rs778369795 Chr.16: 74301125-74301125 on Build GRCh38
rs376081054 Chr.16: 74301162-74301162 on Build GRCh38
rs527730798 Chr.16: 74300883-74300883 on Build GRCh38
rs772759169 Chr.16: 74301034-74301034 on Build GRCh38
rs1044533308 Chr.16: 74300887-74300887 on Build GRCh38
rs980429071 Chr.16: 74300981-74300981 on Build GRCh38
rs776112942 Chr.16: 74301033-74301034 on Build GRCh38
rs770445982 Chr.16: 74301009-74301009 on Build GRCh38
rs531604653 Chr.16: 74300789-74300789 on Build GRCh38
rs575462071 Chr.16: 74301083-74301083 on Build GRCh38
rs932357100 Chr.16: 74300782-74300782 on Build GRCh38
rs747300059 Chr.16: 74301126-74301126 on Build GRCh38
rs377722617 Chr.16: 74300862-74300862 on Build GRCh38
rs905699214 Chr.16: 74300750-74300750 on Build GRCh38
rs748827639 Chr.16: 74301176-74301176 on Build GRCh38
rs749339366 Chr.16: 74300753-74300753 on Build GRCh38
rs908383895 Chr.16: 74300724-74300724 on Build GRCh38
rs952738984 Chr.16: 74300691-74300691 on Build GRCh38
rs765299100 Chr.16: 74301107-74301107 on Build GRCh38
rs947301908 Chr.16: 74300711-74300711 on Build GRCh38
rs74028225 Chr.16: 74300831-74300831 on Build GRCh38
rs1015533324 Chr.16: 74300978-74300978 on Build GRCh38
rs758813349 Chr.16: 74301121-74301121 on Build GRCh38
rs950259742 Chr.16: 74300841-74300841 on Build GRCh38
rs537436459 Chr.16: 74301078-74301078 on Build GRCh38
rs545745493 Chr.16: 74300773-74300773 on Build GRCh38
rs150308557 Chr.16: 74301101-74301101 on Build GRCh38
rs940748859 Chr.16: 74301036-74301036 on Build GRCh38
rs766443918 Chr.16: 74301099-74301099 on Build GRCh38
rs771281162 Chr.16: 74300757-74300757 on Build GRCh38
rs570377167 Chr.16: 74300990-74300990 on Build GRCh38
rs138324633 Chr.16: 74301140-74301140 on Build GRCh38
rs761190927 Chr.16: 74301047-74301048 on Build GRCh38
rs773966773 Chr.16: 74301056-74301056 on Build GRCh38
rs369328599 Chr.16: 74301018-74301018 on Build GRCh38
rs1056950639 Chr.16: 74300998-74300998 on Build GRCh38
rs754073708 Chr.16: 74301100-74301100 on Build GRCh38
rs774574121 Chr.16: 74300790-74300790 on Build GRCh38
rs987629021 Chr.16: 74300870-74300870 on Build GRCh38
rs968320525 Chr.16: 74300956-74300956 on Build GRCh38
rs1053404071 Chr.16: 74300830-74300830 on Build GRCh38
rs752815587 Chr.16: 74301117-74301117 on Build GRCh38
rs547505348 Chr.16: 74300923-74300923 on Build GRCh38
rs776469705 Chr.16: 74300982-74300982 on Build GRCh38
rs550182085 Chr.16: 74300989-74300989 on Build GRCh38
rs1044269803 Chr.16: 74300717-74300717 on Build GRCh38
rs1028086797 Chr.16: 74300955-74300955 on Build GRCh38
rs1029210411 Chr.16: 74300864-74300864 on Build GRCh38

View Full Product Details