Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 80540873-80541375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNLRB2
Entrez Gene ID
83657
Gene Name
dynein light chain roadblock-type 2
Gene Aliases
DNCL2B, DNLC2B, ROBLD2
UniGene
Hs.98849
Gene Chromosome Location
Chr.16: 80540259-80550811 on Build GRCh38
Transcripts
NM_001305017.1 NM_130897.2 NR_130942.1 NR_130943.1 XM_006721295.3 XM_006721296.3 XM_011523370.2 NR_120307.1

Gene Symbol
LOC102724084
Entrez Gene ID
102724084
Gene Name
uncharacterized LOC102724084
Gene Aliases
-
UniGene
Hs.147884
Gene Chromosome Location
Chr.16: 80155958-80563135 on Build GRCh38
Transcripts
NM_001305017.1 NM_130897.2 NR_130942.1 NR_130943.1 XM_006721295.3 XM_006721296.3 XM_011523370.2 NR_120307.1

SNP Detail

SNP ID Chromosome Location
rs562865294 Chr.16: 80541056-80541056 on Build GRCh38
rs761453209 Chr.16: 80541090-80541090 on Build GRCh38
rs1002483437 Chr.16: 80540946-80540946 on Build GRCh38
rs181419897 Chr.16: 80541336-80541336 on Build GRCh38
rs573206693 Chr.16: 80541022-80541022 on Build GRCh38
rs750520266 Chr.16: 80541207-80541207 on Build GRCh38
rs749728820 Chr.16: 80541129-80541129 on Build GRCh38
rs889916661 Chr.16: 80541012-80541012 on Build GRCh38
rs772315633 Chr.16: 80541058-80541058 on Build GRCh38
rs999058563 Chr.16: 80541339-80541339 on Build GRCh38
rs781247522 Chr.16: 80541043-80541043 on Build GRCh38
rs189034057 Chr.16: 80541167-80541167 on Build GRCh38
rs1048878723 Chr.16: 80540895-80540895 on Build GRCh38
rs781150308 Chr.16: 80541106-80541106 on Build GRCh38
rs575252004 Chr.16: 80541224-80541224 on Build GRCh38
rs772190891 Chr.16: 80540893-80540893 on Build GRCh38
rs965445362 Chr.16: 80541204-80541204 on Build GRCh38
rs79420254 Chr.16: 80541317-80541317 on Build GRCh38
rs564071479 Chr.16: 80540995-80540995 on Build GRCh38
rs77727997 Chr.16: 80541032-80541032 on Build GRCh38
rs765662097 Chr.16: 80541031-80541031 on Build GRCh38
rs568288323 Chr.16: 80541193-80541193 on Build GRCh38
rs965393413 Chr.16: 80540993-80540993 on Build GRCh38
rs9922406 Chr.16: 80540989-80540989 on Build GRCh38
rs921521789 Chr.16: 80541120-80541120 on Build GRCh38
rs909118959 Chr.16: 80541263-80541263 on Build GRCh38
rs551723425 Chr.16: 80540902-80540902 on Build GRCh38
rs763798060 Chr.16: 80541036-80541036 on Build GRCh38
rs533431541 Chr.16: 80541060-80541060 on Build GRCh38
rs940421234 Chr.16: 80541190-80541190 on Build GRCh38
rs561988065 Chr.16: 80541030-80541030 on Build GRCh38
rs535572003 Chr.16: 80540951-80540951 on Build GRCh38
rs373786586 Chr.16: 80541128-80541128 on Build GRCh38
rs539469878 Chr.16: 80541262-80541262 on Build GRCh38
rs921404731 Chr.16: 80540947-80540947 on Build GRCh38
rs999317498 Chr.16: 80541291-80541291 on Build GRCh38
rs534875209 Chr.16: 80540927-80540927 on Build GRCh38
rs1055683892 Chr.16: 80541208-80541208 on Build GRCh38
rs776025182 Chr.16: 80541075-80541075 on Build GRCh38
rs535309988 Chr.16: 80541200-80541200 on Build GRCh38
rs7202484 Chr.16: 80541349-80541349 on Build GRCh38
rs757403347 Chr.16: 80541105-80541105 on Build GRCh38
rs750324717 Chr.16: 80541045-80541045 on Build GRCh38
rs534956728 Chr.16: 80541158-80541158 on Build GRCh38
rs894391739 Chr.16: 80541223-80541223 on Build GRCh38
rs760634706 Chr.16: 80541028-80541028 on Build GRCh38
rs570773902 Chr.16: 80540905-80540905 on Build GRCh38
rs370337674 Chr.16: 80541093-80541093 on Build GRCh38
rs374137799 Chr.16: 80541065-80541065 on Build GRCh38
rs934128029 Chr.16: 80541145-80541145 on Build GRCh38
rs928423692 Chr.16: 80541272-80541272 on Build GRCh38
rs890193834 Chr.16: 80540980-80540980 on Build GRCh38
rs1039961996 Chr.16: 80540934-80540934 on Build GRCh38
rs763851304 Chr.16: 80541077-80541077 on Build GRCh38
rs977819435 Chr.16: 80540919-80540919 on Build GRCh38
rs186083546 Chr.16: 80540974-80540974 on Build GRCh38
rs1036101047 Chr.16: 80541049-80541049 on Build GRCh38
rs751083610 Chr.16: 80541037-80541037 on Build GRCh38
rs540561011 Chr.16: 80541310-80541310 on Build GRCh38
rs74729796 Chr.16: 80540933-80540933 on Build GRCh38
rs1019800754 Chr.16: 80541341-80541341 on Build GRCh38
rs7202313 Chr.16: 80541270-80541270 on Build GRCh38
rs762065617 Chr.16: 80541250-80541250 on Build GRCh38
rs763341973 Chr.16: 80541076-80541076 on Build GRCh38
rs1033117017 Chr.16: 80541300-80541300 on Build GRCh38
rs965495971 Chr.16: 80541347-80541347 on Build GRCh38
rs12920691 Chr.16: 80541324-80541324 on Build GRCh38
rs545499976 Chr.16: 80540990-80540990 on Build GRCh38
rs551083473 Chr.16: 80541053-80541053 on Build GRCh38
rs753638151 Chr.16: 80541096-80541096 on Build GRCh38
rs770373346 Chr.16: 80541074-80541074 on Build GRCh38
rs778119951 Chr.16: 80541099-80541099 on Build GRCh38
rs984736276 Chr.16: 80541225-80541225 on Build GRCh38
rs777231204 Chr.16: 80541067-80541067 on Build GRCh38
rs201613754 Chr.16: 80541086-80541086 on Build GRCh38
rs144837151 Chr.16: 80541110-80541110 on Build GRCh38
rs773303146 Chr.16: 80541027-80541027 on Build GRCh38
rs1036124893 Chr.16: 80541192-80541192 on Build GRCh38
rs774262378 Chr.16: 80541252-80541254 on Build GRCh38
rs1004674640 Chr.16: 80540982-80540982 on Build GRCh38
rs574675369 Chr.16: 80540943-80540943 on Build GRCh38
rs906944547 Chr.16: 80541064-80541064 on Build GRCh38
rs1011631560 Chr.16: 80541334-80541334 on Build GRCh38
rs751774105 Chr.16: 80541008-80541008 on Build GRCh38
rs766265562 Chr.16: 80541095-80541095 on Build GRCh38
rs747195901 Chr.16: 80541100-80541100 on Build GRCh38
rs756663810 Chr.16: 80541042-80541042 on Build GRCh38
rs908732940 Chr.16: 80541189-80541189 on Build GRCh38
rs368032058 Chr.16: 80541066-80541066 on Build GRCh38
rs1024178429 Chr.16: 80541047-80541047 on Build GRCh38
rs887571595 Chr.16: 80540913-80540913 on Build GRCh38
rs769604176 Chr.16: 80540901-80540901 on Build GRCh38
rs755943372 Chr.16: 80541046-80541046 on Build GRCh38
rs746350545 Chr.16: 80541070-80541070 on Build GRCh38
rs990760734 Chr.16: 80540965-80540965 on Build GRCh38
rs774560482 Chr.16: 80541027-80541028 on Build GRCh38
rs766907236 Chr.16: 80541092-80541092 on Build GRCh38
rs762714884 Chr.16: 80541035-80541035 on Build GRCh38
rs7185063 Chr.16: 80541281-80541281 on Build GRCh38
rs756070080 Chr.16: 80541094-80541094 on Build GRCh38
rs533269900 Chr.16: 80541351-80541351 on Build GRCh38
rs1043351930 Chr.16: 80541335-80541335 on Build GRCh38
rs557082458 Chr.16: 80541222-80541222 on Build GRCh38
rs975869782 Chr.16: 80541218-80541218 on Build GRCh38
rs540524209 Chr.16: 80541029-80541029 on Build GRCh38
rs1036152550 Chr.16: 80541271-80541271 on Build GRCh38
rs770428374 Chr.16: 80541122-80541122 on Build GRCh38

View Full Product Details