Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84221455-84221957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNG4
Entrez Gene ID
93107
Gene Name
potassium voltage-gated channel modifier subfamily G member 4
Gene Aliases
KV6.3, KV6.4
UniGene
Hs.335877
Gene Chromosome Location
Chr.16: 84221135-84239750 on Build GRCh38
Transcripts
NM_172347.2

SNP Detail

SNP ID Chromosome Location
rs59140300 Chr.16: 84221830-84221830 on Build GRCh38
rs1036105000 Chr.16: 84221729-84221729 on Build GRCh38
rs16963074 Chr.16: 84221846-84221846 on Build GRCh38
rs753810008 Chr.16: 84221704-84221704 on Build GRCh38
rs1009943758 Chr.16: 84221489-84221489 on Build GRCh38
rs983476881 Chr.16: 84221757-84221757 on Build GRCh38
rs1006598540 Chr.16: 84221875-84221875 on Build GRCh38
rs1033019594 Chr.16: 84221773-84221773 on Build GRCh38
rs59521089 Chr.16: 84221510-84221511 on Build GRCh38
rs977485202 Chr.16: 84221769-84221769 on Build GRCh38
rs574828889 Chr.16: 84221603-84221603 on Build GRCh38
rs532296172 Chr.16: 84221531-84221531 on Build GRCh38
rs753151985 Chr.16: 84221792-84221792 on Build GRCh38
rs1042780815 Chr.16: 84221868-84221868 on Build GRCh38
rs60559486 Chr.16: 84221509-84221510 on Build GRCh38
rs540266395 Chr.16: 84221492-84221492 on Build GRCh38
rs766459587 Chr.16: 84221667-84221667 on Build GRCh38
rs35508178 Chr.16: 84221568-84221568 on Build GRCh38
rs1056982899 Chr.16: 84221654-84221654 on Build GRCh38
rs967775952 Chr.16: 84221767-84221767 on Build GRCh38
rs963535018 Chr.16: 84221516-84221516 on Build GRCh38
rs183066639 Chr.16: 84221887-84221887 on Build GRCh38
rs927126586 Chr.16: 84221643-84221643 on Build GRCh38
rs1030445229 Chr.16: 84221736-84221736 on Build GRCh38
rs1018204766 Chr.16: 84221541-84221541 on Build GRCh38
rs921745412 Chr.16: 84221794-84221794 on Build GRCh38
rs117479346 Chr.16: 84221653-84221653 on Build GRCh38
rs767944213 Chr.16: 84221497-84221510 on Build GRCh38
rs759732709 Chr.16: 84221537-84221537 on Build GRCh38
rs111841509 Chr.16: 84221752-84221752 on Build GRCh38
rs754102587 Chr.16: 84221496-84221497 on Build GRCh38
rs767648153 Chr.16: 84221502-84221503 on Build GRCh38
rs12919548 Chr.16: 84221900-84221900 on Build GRCh38
rs570881669 Chr.16: 84221627-84221627 on Build GRCh38
rs921830587 Chr.16: 84221638-84221638 on Build GRCh38
rs866224408 Chr.16: 84221503-84221503 on Build GRCh38
rs1035473904 Chr.16: 84221478-84221478 on Build GRCh38
rs748819998 Chr.16: 84221888-84221891 on Build GRCh38
rs964875628 Chr.16: 84221547-84221547 on Build GRCh38
rs367628482 Chr.16: 84221877-84221877 on Build GRCh38
rs980428902 Chr.16: 84221813-84221813 on Build GRCh38
rs568777040 Chr.16: 84221565-84221565 on Build GRCh38
rs923812020 Chr.16: 84221587-84221587 on Build GRCh38
rs186712007 Chr.16: 84221514-84221514 on Build GRCh38
rs994966089 Chr.16: 84221748-84221748 on Build GRCh38
rs142851377 Chr.16: 84221761-84221761 on Build GRCh38
rs957886266 Chr.16: 84221778-84221778 on Build GRCh38
rs927270905 Chr.16: 84221816-84221816 on Build GRCh38
rs1043137301 Chr.16: 84221675-84221675 on Build GRCh38
rs931938991 Chr.16: 84221805-84221805 on Build GRCh38
rs39549 Chr.16: 84221510-84221510 on Build GRCh38
rs891583990 Chr.16: 84221663-84221663 on Build GRCh38
rs891469466 Chr.16: 84221488-84221488 on Build GRCh38
rs752091714 Chr.16: 84221874-84221877 on Build GRCh38
rs904192175 Chr.16: 84221873-84221873 on Build GRCh38
rs1057015493 Chr.16: 84221840-84221840 on Build GRCh38
rs1017818194 Chr.16: 84221682-84221682 on Build GRCh38
rs548051042 Chr.16: 84221728-84221728 on Build GRCh38
rs778494577 Chr.16: 84221899-84221899 on Build GRCh38
rs576463206 Chr.16: 84221914-84221914 on Build GRCh38
rs565658317 Chr.16: 84221909-84221910 on Build GRCh38
rs936476636 Chr.16: 84221594-84221594 on Build GRCh38
rs941960930 Chr.16: 84221625-84221625 on Build GRCh38
rs995819429 Chr.16: 84221480-84221480 on Build GRCh38
rs911594469 Chr.16: 84221864-84221864 on Build GRCh38
rs909197265 Chr.16: 84221617-84221617 on Build GRCh38
rs529903956 Chr.16: 84221649-84221649 on Build GRCh38
rs1039771126 Chr.16: 84221885-84221885 on Build GRCh38
rs1001535030 Chr.16: 84221681-84221681 on Build GRCh38
rs940303670 Chr.16: 84221880-84221880 on Build GRCh38
rs938570500 Chr.16: 84221820-84221820 on Build GRCh38
rs192497144 Chr.16: 84221572-84221572 on Build GRCh38
rs945794310 Chr.16: 84221862-84221862 on Build GRCh38
rs900793449 Chr.16: 84221886-84221886 on Build GRCh38
rs375775550 Chr.16: 84221500-84221513 on Build GRCh38
rs977475123 Chr.16: 84221584-84221584 on Build GRCh38
rs964930109 Chr.16: 84221726-84221726 on Build GRCh38
rs548494549 Chr.16: 84221564-84221564 on Build GRCh38
rs779618599 Chr.16: 84221508-84221509 on Build GRCh38
rs757753073 Chr.16: 84221879-84221879 on Build GRCh38
rs553540807 Chr.16: 84221679-84221679 on Build GRCh38
rs140352628 Chr.16: 84221480-84221481 on Build GRCh38
rs1005024347 Chr.16: 84221496-84221496 on Build GRCh38
rs779479861 Chr.16: 84221885-84221886 on Build GRCh38

View Full Product Details