Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 90029089-90029584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS8
Entrez Gene ID
2622
Gene Name
growth arrest specific 8
Gene Aliases
CILD33, DRC4, GAS11
UniGene
Hs.431792
Gene Chromosome Location
Chr.16: 90019629-90044971 on Build GRCh38
Transcripts
NM_001286205.1 NM_001286208.1 NM_001286209.1 NM_001481.2 XM_005256304.4 XM_005256309.4 XM_006721175.3 XM_011522990.2 XM_011522991.1 XM_011522992.2 XM_017023122.1 XM_017023123.1 XM_017023124.1 XM_017023125.1 NR_122031.1

Gene Symbol
GAS8-AS1
Entrez Gene ID
750
Gene Name
GAS8 antisense RNA 1
Gene Aliases
C16orf3
UniGene
Hs.633032
Gene Chromosome Location
Chr.16: 90028908-90029901 on Build GRCh38
Transcripts
NM_001286205.1 NM_001286208.1 NM_001286209.1 NM_001481.2 XM_005256304.4 XM_005256309.4 XM_006721175.3 XM_011522990.2 XM_011522991.1 XM_011522992.2 XM_017023122.1 XM_017023123.1 XM_017023124.1 XM_017023125.1 NR_122031.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7428916 c.133_134ins12 p.V44_A45insGCPA Chr.16: 90029209-90029210 on Build GRCh38
4129536 c.248G>T p.W83L Chr.16: 90029095-90029095 on Build GRCh38
7358100 c.143T>C p.V48A Chr.16: 90029200-90029200 on Build GRCh38
3957942 c.165C>A p.P55P Chr.16: 90029178-90029178 on Build GRCh38
4063714 c.110G>A p.C37Y Chr.16: 90029233-90029233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900252794 Chr.16: 90029527-90029527 on Build GRCh38
rs781342272 Chr.16: 90029274-90029274 on Build GRCh38
rs77382359 Chr.16: 90029165-90029165 on Build GRCh38
rs542497003 Chr.16: 90029275-90029275 on Build GRCh38
rs539417855 Chr.16: 90029176-90029176 on Build GRCh38
rs777740957 Chr.16: 90029386-90029387 on Build GRCh38
rs768481971 Chr.16: 90029162-90029163 on Build GRCh38
rs773519784 Chr.16: 90029238-90029238 on Build GRCh38
rs553077045 Chr.16: 90029235-90029235 on Build GRCh38
rs202095648 Chr.16: 90029200-90029200 on Build GRCh38
rs555737522 Chr.16: 90029499-90029499 on Build GRCh38
rs897909981 Chr.16: 90029162-90029162 on Build GRCh38
rs761727857 Chr.16: 90029537-90029537 on Build GRCh38
rs552590997 Chr.16: 90029135-90029146 on Build GRCh38
rs999839812 Chr.16: 90029532-90029532 on Build GRCh38
rs754162787 Chr.16: 90029112-90029112 on Build GRCh38
rs761914372 Chr.16: 90029178-90029178 on Build GRCh38
rs766188860 Chr.16: 90029323-90029323 on Build GRCh38
rs750801215 Chr.16: 90029138-90029138 on Build GRCh38
rs766371390 Chr.16: 90029473-90029473 on Build GRCh38
rs748113286 Chr.16: 90029165-90029173 on Build GRCh38
rs554912454 Chr.16: 90029213-90029213 on Build GRCh38
rs185580603 Chr.16: 90029164-90029164 on Build GRCh38
rs779148014 Chr.16: 90029384-90029384 on Build GRCh38
rs777634190 Chr.16: 90029196-90029197 on Build GRCh38
rs777712402 Chr.16: 90029361-90029361 on Build GRCh38
rs76322535 Chr.16: 90029201-90029201 on Build GRCh38
rs767948663 Chr.16: 90029202-90029202 on Build GRCh38
rs770393003 Chr.16: 90029197-90029198 on Build GRCh38
rs779785952 Chr.16: 90029264-90029264 on Build GRCh38
rs541125315 Chr.16: 90029230-90029230 on Build GRCh38
rs748759188 Chr.16: 90029154-90029154 on Build GRCh38
rs752484462 Chr.16: 90029222-90029223 on Build GRCh38
rs141306543 Chr.16: 90029265-90029265 on Build GRCh38
rs771158938 Chr.16: 90029288-90029288 on Build GRCh38
rs778373318 Chr.16: 90029161-90029161 on Build GRCh38
rs200068483 Chr.16: 90029141-90029141 on Build GRCh38
rs758664017 Chr.16: 90029142-90029189 on Build GRCh38
rs747428627 Chr.16: 90029268-90029268 on Build GRCh38
rs55742939 Chr.16: 90029188-90029188 on Build GRCh38
rs535195978 Chr.16: 90029552-90029552 on Build GRCh38
rs767784679 Chr.16: 90029220-90029221 on Build GRCh38
rs777152825 Chr.16: 90029211-90029212 on Build GRCh38
rs772790659 Chr.16: 90029175-90029198 on Build GRCh38
rs988614940 Chr.16: 90029294-90029294 on Build GRCh38
rs148430683 Chr.16: 90029110-90029110 on Build GRCh38
rs529378637 Chr.16: 90029140-90029140 on Build GRCh38
rs891704859 Chr.16: 90029549-90029549 on Build GRCh38
rs746759021 Chr.16: 90029223-90029223 on Build GRCh38
rs753361636 Chr.16: 90029204-90029204 on Build GRCh38
rs966273969 Chr.16: 90029180-90029180 on Build GRCh38
rs755939214 Chr.16: 90029365-90029365 on Build GRCh38
rs780199710 Chr.16: 90029150-90029151 on Build GRCh38
rs776649947 Chr.16: 90029227-90029227 on Build GRCh38
rs145735978 Chr.16: 90029197-90029197 on Build GRCh38
rs201366409 Chr.16: 90029205-90029205 on Build GRCh38
rs751248218 Chr.16: 90029334-90029334 on Build GRCh38
rs1038625783 Chr.16: 90029167-90029167 on Build GRCh38
rs755205392 Chr.16: 90029221-90029222 on Build GRCh38
rs565800945 Chr.16: 90029322-90029322 on Build GRCh38
rs780017069 Chr.16: 90029222-90029222 on Build GRCh38
rs144239317 Chr.16: 90029556-90029556 on Build GRCh38
rs76646627 Chr.16: 90029150-90029150 on Build GRCh38
rs756940648 Chr.16: 90029156-90029156 on Build GRCh38
rs746817197 Chr.16: 90029240-90029240 on Build GRCh38
rs79367624 Chr.16: 90029541-90029541 on Build GRCh38
rs930255385 Chr.16: 90029432-90029432 on Build GRCh38
rs758262118 Chr.16: 90029221-90029221 on Build GRCh38
rs774543370 Chr.16: 90029297-90029297 on Build GRCh38
rs758018372 Chr.16: 90029263-90029263 on Build GRCh38
rs774517336 Chr.16: 90029259-90029260 on Build GRCh38
rs565041606 Chr.16: 90029500-90029500 on Build GRCh38
rs974061213 Chr.16: 90029380-90029380 on Build GRCh38
rs775608049 Chr.16: 90029169-90029169 on Build GRCh38
rs528224678 Chr.16: 90029301-90029301 on Build GRCh38
rs779167450 Chr.16: 90029138-90029161 on Build GRCh38
rs75909132 Chr.16: 90029282-90029282 on Build GRCh38
rs762329520 Chr.16: 90029212-90029213 on Build GRCh38
rs759840058 Chr.16: 90029244-90029244 on Build GRCh38
rs764620466 Chr.16: 90029318-90029318 on Build GRCh38
rs777748751 Chr.16: 90029283-90029283 on Build GRCh38
rs750547360 Chr.16: 90029140-90029141 on Build GRCh38
rs774003272 Chr.16: 90029201-90029202 on Build GRCh38
rs140572603 Chr.16: 90029242-90029242 on Build GRCh38
rs184911076 Chr.16: 90029470-90029470 on Build GRCh38
rs192324871 Chr.16: 90029431-90029431 on Build GRCh38
rs749906203 Chr.16: 90029320-90029320 on Build GRCh38
rs763110146 Chr.16: 90029239-90029239 on Build GRCh38
rs61118444 Chr.16: 90029189-90029189 on Build GRCh38
rs781109039 Chr.16: 90029350-90029350 on Build GRCh38
rs781087845 Chr.16: 90029164-90029168 on Build GRCh38
rs138059586 Chr.16: 90029151-90029151 on Build GRCh38
rs201803875 Chr.16: 90029137-90029137 on Build GRCh38
rs745549957 Chr.16: 90029202-90029203 on Build GRCh38
rs768717453 Chr.16: 90029224-90029224 on Build GRCh38
rs533185852 Chr.16: 90029387-90029387 on Build GRCh38
rs754857908 Chr.16: 90029341-90029341 on Build GRCh38
rs746068936 Chr.16: 90029390-90029390 on Build GRCh38
rs62648073 Chr.16: 90029198-90029198 on Build GRCh38
rs1050346084 Chr.16: 90029151-90029174 on Build GRCh38
rs778694484 Chr.16: 90029209-90029209 on Build GRCh38
rs955959773 Chr.16: 90029289-90029289 on Build GRCh38
rs770608205 Chr.16: 90029174-90029175 on Build GRCh38
rs1021760928 Chr.16: 90029455-90029455 on Build GRCh38
rs71137702 Chr.16: 90029189-90029190 on Build GRCh38
rs776115872 Chr.16: 90029309-90029309 on Build GRCh38
rs769078996 Chr.16: 90029209-90029210 on Build GRCh38
rs1039159451 Chr.16: 90029525-90029525 on Build GRCh38
rs547505340 Chr.16: 90029316-90029316 on Build GRCh38
rs762418985 Chr.16: 90029188-90029189 on Build GRCh38
rs62640380 Chr.16: 90029174-90029174 on Build GRCh38
rs749897357 Chr.16: 90029259-90029259 on Build GRCh38
rs761096651 Chr.16: 90029187-90029188 on Build GRCh38
rs959929718 Chr.16: 90029516-90029516 on Build GRCh38
rs765506826 Chr.16: 90029184-90029184 on Build GRCh38
rs772396930 Chr.16: 90029306-90029306 on Build GRCh38
rs749327299 Chr.16: 90029285-90029285 on Build GRCh38
rs746816695 Chr.16: 90029163-90029174 on Build GRCh38
rs540458058 Chr.16: 90029423-90029423 on Build GRCh38
rs149576587 Chr.16: 90029258-90029258 on Build GRCh38
rs756031326 Chr.16: 90029191-90029212 on Build GRCh38
rs773911683 Chr.16: 90029185-90029186 on Build GRCh38
rs551360819 Chr.16: 90029424-90029424 on Build GRCh38
rs763068079 Chr.16: 90029219-90029220 on Build GRCh38
rs756011919 Chr.16: 90029360-90029360 on Build GRCh38
rs748797399 Chr.16: 90029198-90029209 on Build GRCh38
rs61740023 Chr.16: 90029212-90029212 on Build GRCh38
rs796501176 Chr.16: 90029187-90029188 on Build GRCh38
rs141303781 Chr.16: 90029190-90029191 on Build GRCh38
rs754359438 Chr.16: 90029188-90029192 on Build GRCh38
rs772231911 Chr.16: 90029394-90029394 on Build GRCh38
rs762428710 Chr.16: 90029175-90029177 on Build GRCh38
rs757365469 Chr.16: 90029134-90029135 on Build GRCh38
rs771760283 Chr.16: 90029166-90029166 on Build GRCh38
rs769226522 Chr.16: 90029171-90029189 on Build GRCh38
rs997746618 Chr.16: 90029353-90029353 on Build GRCh38
rs777204520 Chr.16: 90029385-90029385 on Build GRCh38
rs984816228 Chr.16: 90029542-90029542 on Build GRCh38
rs765770690 Chr.16: 90029213-90029214 on Build GRCh38
rs773633513 Chr.16: 90029217-90029218 on Build GRCh38
rs74424322 Chr.16: 90029246-90029246 on Build GRCh38
rs752500178 Chr.16: 90029351-90029351 on Build GRCh38
rs770995173 Chr.16: 90029367-90029367 on Build GRCh38
rs1024143845 Chr.16: 90029557-90029557 on Build GRCh38
rs762147195 Chr.16: 90029129-90029129 on Build GRCh38
rs918527332 Chr.16: 90029523-90029523 on Build GRCh38
rs150458434 Chr.16: 90029243-90029243 on Build GRCh38
rs746920655 Chr.16: 90029170-90029170 on Build GRCh38
rs57136347 Chr.16: 90029177-90029177 on Build GRCh38
rs188012743 Chr.16: 90029519-90029519 on Build GRCh38
rs372009006 Chr.16: 90029185-90029185 on Build GRCh38
rs766963957 Chr.16: 90029142-90029142 on Build GRCh38

View Full Product Details