Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1754413-1754914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINF2
Entrez Gene ID
5345
Gene Name
serpin family F member 2
Gene Aliases
A2AP, AAP, ALPHA-2-PI, API, PLI
UniGene
Hs.159509
Gene Chromosome Location
Chr.17: 1742808-1755265 on Build GRCh38
Transcripts
NM_000934.3 NM_001165920.1 NM_001165921.1 XM_005256701.4 XM_017024765.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4943027 c.1420T>C p.L474L Chr.17: 1754478-1754478 on Build GRCh38
5926980 c.1391C>T p.P464L Chr.17: 1754449-1754449 on Build GRCh38
6775669 c.1393C>A p.R465S Chr.17: 1754451-1754451 on Build GRCh38
4526701 c.1411G>A p.G471S Chr.17: 1754469-1754469 on Build GRCh38
2739603 c.1395C>T p.R465R Chr.17: 1754453-1754453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138629993 Chr.17: 1754468-1754468 on Build GRCh38
rs753477160 Chr.17: 1754533-1754533 on Build GRCh38
rs189035843 Chr.17: 1754831-1754831 on Build GRCh38
rs769869981 Chr.17: 1754511-1754511 on Build GRCh38
rs371013906 Chr.17: 1754443-1754443 on Build GRCh38
rs556923918 Chr.17: 1754524-1754524 on Build GRCh38
rs780872308 Chr.17: 1754567-1754567 on Build GRCh38
rs929363192 Chr.17: 1754846-1754846 on Build GRCh38
rs900675904 Chr.17: 1754743-1754743 on Build GRCh38
rs547103244 Chr.17: 1754781-1754781 on Build GRCh38
rs926438541 Chr.17: 1754613-1754613 on Build GRCh38
rs372733955 Chr.17: 1754472-1754472 on Build GRCh38
rs566749778 Chr.17: 1754447-1754447 on Build GRCh38
rs755809297 Chr.17: 1754451-1754451 on Build GRCh38
rs759910550 Chr.17: 1754520-1754520 on Build GRCh38
rs542730161 Chr.17: 1754572-1754572 on Build GRCh38
rs773199954 Chr.17: 1754515-1754515 on Build GRCh38
rs181358902 Chr.17: 1754733-1754733 on Build GRCh38
rs367964307 Chr.17: 1754442-1754442 on Build GRCh38
rs995748256 Chr.17: 1754585-1754585 on Build GRCh38
rs869205842 Chr.17: 1754695-1754695 on Build GRCh38
rs577213142 Chr.17: 1754455-1754455 on Build GRCh38
rs545313542 Chr.17: 1754630-1754630 on Build GRCh38
rs1045980425 Chr.17: 1754694-1754695 on Build GRCh38
rs773323308 Chr.17: 1754576-1754576 on Build GRCh38
rs144890920 Chr.17: 1754454-1754454 on Build GRCh38
rs745778434 Chr.17: 1754459-1754459 on Build GRCh38
rs775128832 Chr.17: 1754465-1754465 on Build GRCh38
rs887805240 Chr.17: 1754726-1754726 on Build GRCh38
rs184228093 Chr.17: 1754792-1754792 on Build GRCh38
rs987873982 Chr.17: 1754889-1754889 on Build GRCh38
rs756890578 Chr.17: 1754536-1754536 on Build GRCh38
rs778453597 Chr.17: 1754540-1754540 on Build GRCh38
rs936396033 Chr.17: 1754688-1754688 on Build GRCh38
rs1057355 Chr.17: 1754605-1754605 on Build GRCh38
rs780034922 Chr.17: 1754545-1754545 on Build GRCh38
rs566710940 Chr.17: 1754782-1754782 on Build GRCh38
rs538545736 Chr.17: 1754860-1754860 on Build GRCh38
rs375490272 Chr.17: 1754452-1754452 on Build GRCh38
rs746953947 Chr.17: 1754553-1754553 on Build GRCh38
rs1021021907 Chr.17: 1754804-1754804 on Build GRCh38
rs752842203 Chr.17: 1754478-1754478 on Build GRCh38
rs1047834042 Chr.17: 1754848-1754848 on Build GRCh38
rs530113512 Chr.17: 1754741-1754741 on Build GRCh38
rs918011077 Chr.17: 1754648-1754648 on Build GRCh38
rs764804209 Chr.17: 1754587-1754588 on Build GRCh38
rs1048210813 Chr.17: 1754720-1754720 on Build GRCh38
rs1056561056 Chr.17: 1754631-1754631 on Build GRCh38
rs565229249 Chr.17: 1754636-1754636 on Build GRCh38
rs986746022 Chr.17: 1754619-1754619 on Build GRCh38
rs761954931 Chr.17: 1754488-1754488 on Build GRCh38
rs929261523 Chr.17: 1754649-1754649 on Build GRCh38
rs67887323 Chr.17: 1754686-1754686 on Build GRCh38
rs748366163 Chr.17: 1754571-1754571 on Build GRCh38
rs925032463 Chr.17: 1754686-1754686 on Build GRCh38
rs117803213 Chr.17: 1754861-1754861 on Build GRCh38
rs200446774 Chr.17: 1754541-1754541 on Build GRCh38
rs1025520610 Chr.17: 1754604-1754604 on Build GRCh38
rs975078477 Chr.17: 1754640-1754640 on Build GRCh38
rs963961424 Chr.17: 1754840-1754840 on Build GRCh38
rs751965615 Chr.17: 1754519-1754519 on Build GRCh38
rs115496485 Chr.17: 1754832-1754832 on Build GRCh38
rs1025723308 Chr.17: 1754803-1754803 on Build GRCh38
rs765236572 Chr.17: 1754436-1754436 on Build GRCh38
rs959156105 Chr.17: 1754837-1754837 on Build GRCh38
rs917982415 Chr.17: 1754844-1754844 on Build GRCh38
rs1055757603 Chr.17: 1754740-1754740 on Build GRCh38
rs368540128 Chr.17: 1754453-1754453 on Build GRCh38
rs763488331 Chr.17: 1754516-1754516 on Build GRCh38
rs550661879 Chr.17: 1754728-1754728 on Build GRCh38
rs778765089 Chr.17: 1754816-1754816 on Build GRCh38
rs967768903 Chr.17: 1754811-1754811 on Build GRCh38
rs771136078 Chr.17: 1754586-1754586 on Build GRCh38
rs1039655406 Chr.17: 1754878-1754878 on Build GRCh38
rs778391884 Chr.17: 1754843-1754843 on Build GRCh38
rs886966170 Chr.17: 1754815-1754815 on Build GRCh38
rs997641746 Chr.17: 1754778-1754778 on Build GRCh38
rs865905064 Chr.17: 1754689-1754689 on Build GRCh38
rs189673650 Chr.17: 1754694-1754694 on Build GRCh38
rs768383854 Chr.17: 1754558-1754558 on Build GRCh38
rs528329599 Chr.17: 1754621-1754621 on Build GRCh38
rs779801686 Chr.17: 1754471-1754471 on Build GRCh38

View Full Product Details