Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3397355-3397876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR1E1
Entrez Gene ID
8387
Gene Name
olfactory receptor family 1 subfamily E member 1
Gene Aliases
HGM071, OR13-66, OR17-2, OR17-32, OR1E5, OR1E6, OR1E8P, OR1E9P, OST547
UniGene
Hs.278485
Gene Chromosome Location
Chr.17: 3397466-3398410 on Build GRCh38
Transcripts
NM_003553.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4510709 c.845C>T p.T282I Chr.17: 3397566-3397566 on Build GRCh38
6733915 c.874C>A p.L292M Chr.17: 3397537-3397537 on Build GRCh38
3180922 c.778T>G p.L260V Chr.17: 3397633-3397633 on Build GRCh38
4265856 c.810G>A p.K270K Chr.17: 3397601-3397601 on Build GRCh38
705544 c.730C>A p.H244N Chr.17: 3397681-3397681 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201978284 Chr.17: 3397604-3397604 on Build GRCh38
rs772998017 Chr.17: 3397658-3397658 on Build GRCh38
rs774514315 Chr.17: 3397523-3397523 on Build GRCh38
rs200854847 Chr.17: 3397554-3397554 on Build GRCh38
rs980040562 Chr.17: 3397410-3397410 on Build GRCh38
rs775588505 Chr.17: 3397823-3397823 on Build GRCh38
rs151232831 Chr.17: 3397758-3397758 on Build GRCh38
rs752172479 Chr.17: 3397745-3397745 on Build GRCh38
rs769422 Chr.17: 3397640-3397640 on Build GRCh38
rs201027671 Chr.17: 3397481-3397481 on Build GRCh38
rs918024933 Chr.17: 3397485-3397485 on Build GRCh38
rs983979805 Chr.17: 3397817-3397817 on Build GRCh38
rs959866329 Chr.17: 3397416-3397416 on Build GRCh38
rs369286312 Chr.17: 3397443-3397443 on Build GRCh38
rs1001166215 Chr.17: 3397729-3397729 on Build GRCh38
rs757074095 Chr.17: 3397427-3397427 on Build GRCh38
rs187851727 Chr.17: 3397395-3397395 on Build GRCh38
rs764241056 Chr.17: 3397655-3397655 on Build GRCh38
rs779496590 Chr.17: 3397466-3397466 on Build GRCh38
rs404983 Chr.17: 3397652-3397652 on Build GRCh38
rs144219652 Chr.17: 3397573-3397573 on Build GRCh38
rs370567870 Chr.17: 3397808-3397808 on Build GRCh38
rs779472725 Chr.17: 3397799-3397799 on Build GRCh38
rs755734934 Chr.17: 3397746-3397746 on Build GRCh38
rs758939852 Chr.17: 3397385-3397385 on Build GRCh38
rs754480072 Chr.17: 3397600-3397600 on Build GRCh38
rs899533824 Chr.17: 3397401-3397401 on Build GRCh38
rs747820921 Chr.17: 3397495-3397495 on Build GRCh38
rs758303133 Chr.17: 3397577-3397577 on Build GRCh38
rs779376848 Chr.17: 3397700-3397700 on Build GRCh38
rs771587810 Chr.17: 3397522-3397522 on Build GRCh38
rs145735312 Chr.17: 3397691-3397691 on Build GRCh38
rs771976708 Chr.17: 3397841-3397842 on Build GRCh38
rs770178976 Chr.17: 3397508-3397508 on Build GRCh38
rs1025927199 Chr.17: 3397811-3397811 on Build GRCh38
rs747811841 Chr.17: 3397606-3397606 on Build GRCh38
rs754955994 Chr.17: 3397710-3397710 on Build GRCh38
rs959919337 Chr.17: 3397801-3397801 on Build GRCh38
rs113011400 Chr.17: 3397826-3397826 on Build GRCh38
rs769364728 Chr.17: 3397610-3397610 on Build GRCh38
rs762080199 Chr.17: 3397829-3397829 on Build GRCh38
rs369810300 Chr.17: 3397390-3397390 on Build GRCh38
rs777295575 Chr.17: 3397755-3397755 on Build GRCh38
rs545030238 Chr.17: 3397777-3397777 on Build GRCh38
rs379147 Chr.17: 3397845-3397845 on Build GRCh38
rs201710869 Chr.17: 3397399-3397401 on Build GRCh38
rs763750126 Chr.17: 3397848-3397848 on Build GRCh38
rs753624782 Chr.17: 3397419-3397419 on Build GRCh38
rs377494453 Chr.17: 3397570-3397570 on Build GRCh38
rs778634585 Chr.17: 3397437-3397437 on Build GRCh38
rs58631068 Chr.17: 3397649-3397649 on Build GRCh38
rs900729801 Chr.17: 3397591-3397591 on Build GRCh38
rs373720102 Chr.17: 3397516-3397516 on Build GRCh38
rs750212682 Chr.17: 3397576-3397576 on Build GRCh38
rs779975724 Chr.17: 3397580-3397580 on Build GRCh38
rs766977107 Chr.17: 3397711-3397711 on Build GRCh38
rs751004389 Chr.17: 3397583-3397583 on Build GRCh38
rs766138437 Chr.17: 3397833-3397833 on Build GRCh38
rs758304726 Chr.17: 3397463-3397463 on Build GRCh38
rs773492322 Chr.17: 3397513-3397513 on Build GRCh38
rs767776825 Chr.17: 3397550-3397550 on Build GRCh38
rs997882053 Chr.17: 3397497-3397497 on Build GRCh38
rs379856 Chr.17: 3397627-3397627 on Build GRCh38
rs746452905 Chr.17: 3397718-3397720 on Build GRCh38
rs983015894 Chr.17: 3397743-3397743 on Build GRCh38
rs780826426 Chr.17: 3397486-3397486 on Build GRCh38
rs193164988 Chr.17: 3397670-3397670 on Build GRCh38
rs780736624 Chr.17: 3397602-3397602 on Build GRCh38
rs367907808 Chr.17: 3397813-3397813 on Build GRCh38
rs774169693 Chr.17: 3397838-3397838 on Build GRCh38
rs759697594 Chr.17: 3397527-3397527 on Build GRCh38
rs760182354 Chr.17: 3397641-3397641 on Build GRCh38
rs927186122 Chr.17: 3397412-3397412 on Build GRCh38
rs778151007 Chr.17: 3397621-3397621 on Build GRCh38
rs757667722 Chr.17: 3397528-3397529 on Build GRCh38
rs558662590 Chr.17: 3397635-3397635 on Build GRCh38
rs768239217 Chr.17: 3397482-3397482 on Build GRCh38
rs747069286 Chr.17: 3397825-3397825 on Build GRCh38
rs61735439 Chr.17: 3397676-3397676 on Build GRCh38
rs757686849 Chr.17: 3397774-3397774 on Build GRCh38

View Full Product Details