Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4900499-4900990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf107
Entrez Gene ID
100130311
Gene Name
chromosome 17 open reading frame 107
Gene Aliases
-
UniGene
Hs.579243
Gene Chromosome Location
Chr.17: 4899653-4902932 on Build GRCh38
Transcripts
NM_001145536.1 NM_000080.3 XM_017024115.1

Gene Symbol
CHRNE
Entrez Gene ID
1145
Gene Name
cholinergic receptor nicotinic epsilon subunit
Gene Aliases
ACHRE, CMS1D, CMS1E, CMS2A, CMS4A, CMS4B, CMS4C, FCCMS, SCCMS
UniGene
Hs.654535
Gene Chromosome Location
Chr.17: 4897769-4905019 on Build GRCh38
Transcripts
NM_001145536.1 NM_000080.3 XM_017024115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
980886 c.907C>T p.L303F Chr.17: 4900803-4900803 on Build GRCh38
4067523 c.904C>T p.P302S Chr.17: 4900806-4900806 on Build GRCh38
5755069 c.893C>T p.S298F Chr.17: 4900817-4900817 on Build GRCh38
1384467 c.876G>A p.Q292Q Chr.17: 4900834-4900834 on Build GRCh38
7358132 c.541G>T p.E181* Chr.17: 4900501-4900501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774291429 Chr.17: 4900751-4900751 on Build GRCh38
rs961101973 Chr.17: 4900786-4900786 on Build GRCh38
rs121909511 Chr.17: 4900845-4900845 on Build GRCh38
rs398122830 Chr.17: 4900816-4900818 on Build GRCh38
rs904469277 Chr.17: 4900696-4900696 on Build GRCh38
rs760782201 Chr.17: 4900550-4900550 on Build GRCh38
rs377633497 Chr.17: 4900702-4900702 on Build GRCh38
rs1000043550 Chr.17: 4900519-4900519 on Build GRCh38
rs374156332 Chr.17: 4900804-4900804 on Build GRCh38
rs900612569 Chr.17: 4900872-4900872 on Build GRCh38
rs1021297889 Chr.17: 4900903-4900903 on Build GRCh38
rs373958533 Chr.17: 4900564-4900564 on Build GRCh38
rs754325287 Chr.17: 4900784-4900784 on Build GRCh38
rs887770442 Chr.17: 4900891-4900891 on Build GRCh38
rs61735425 Chr.17: 4900827-4900827 on Build GRCh38
rs767922649 Chr.17: 4900911-4900912 on Build GRCh38
rs952266614 Chr.17: 4900623-4900623 on Build GRCh38
rs142735836 Chr.17: 4900814-4900814 on Build GRCh38
rs774407731 Chr.17: 4900890-4900890 on Build GRCh38
rs34293621 Chr.17: 4900581-4900581 on Build GRCh38
rs761293676 Chr.17: 4900931-4900931 on Build GRCh38
rs773929089 Chr.17: 4900863-4900863 on Build GRCh38
rs151006465 Chr.17: 4900849-4900849 on Build GRCh38
rs887616799 Chr.17: 4900692-4900692 on Build GRCh38
rs767266015 Chr.17: 4900772-4900772 on Build GRCh38
rs1022488693 Chr.17: 4900634-4900634 on Build GRCh38
rs750892511 Chr.17: 4900944-4900944 on Build GRCh38
rs960911988 Chr.17: 4900770-4900770 on Build GRCh38
rs961218388 Chr.17: 4900913-4900913 on Build GRCh38
rs762704832 Chr.17: 4900910-4900913 on Build GRCh38
rs763590279 Chr.17: 4900843-4900843 on Build GRCh38
rs1031334298 Chr.17: 4900588-4900588 on Build GRCh38
rs1027597321 Chr.17: 4900749-4900749 on Build GRCh38
rs781536510 Chr.17: 4900888-4900888 on Build GRCh38
rs996197218 Chr.17: 4900876-4900876 on Build GRCh38
rs1054386676 Chr.17: 4900737-4900760 on Build GRCh38
rs143928868 Chr.17: 4900701-4900701 on Build GRCh38
rs766217556 Chr.17: 4900750-4900750 on Build GRCh38
rs775985643 Chr.17: 4900745-4900768 on Build GRCh38
rs965749462 Chr.17: 4900548-4900548 on Build GRCh38
rs891910569 Chr.17: 4900543-4900543 on Build GRCh38
rs1019398391 Chr.17: 4900569-4900569 on Build GRCh38
rs760274806 Chr.17: 4900841-4900841 on Build GRCh38
rs908031850 Chr.17: 4900633-4900633 on Build GRCh38
rs770980262 Chr.17: 4900820-4900820 on Build GRCh38
rs536956104 Chr.17: 4900764-4900764 on Build GRCh38
rs766710383 Chr.17: 4900949-4900949 on Build GRCh38
rs775157123 Chr.17: 4900840-4900840 on Build GRCh38
rs1049088476 Chr.17: 4900880-4900880 on Build GRCh38
rs747189862 Chr.17: 4900869-4900869 on Build GRCh38
rs140023380 Chr.17: 4900809-4900809 on Build GRCh38
rs751857871 Chr.17: 4900957-4900957 on Build GRCh38
rs992228063 Chr.17: 4900795-4900795 on Build GRCh38
rs1031496136 Chr.17: 4900524-4900524 on Build GRCh38
rs570266272 Chr.17: 4900883-4900883 on Build GRCh38
rs770586668 Chr.17: 4900858-4900858 on Build GRCh38
rs933082734 Chr.17: 4900866-4900866 on Build GRCh38
rs571059090 Chr.17: 4900710-4900710 on Build GRCh38
rs979787967 Chr.17: 4900806-4900806 on Build GRCh38
rs919252917 Chr.17: 4900679-4900679 on Build GRCh38
rs540766332 Chr.17: 4900635-4900635 on Build GRCh38
rs121909510 Chr.17: 4900860-4900860 on Build GRCh38
rs775149431 Chr.17: 4900906-4900906 on Build GRCh38
rs1017271148 Chr.17: 4900545-4900545 on Build GRCh38
rs753179619 Chr.17: 4900956-4900957 on Build GRCh38
rs149568951 Chr.17: 4900768-4900769 on Build GRCh38
rs374963791 Chr.17: 4900744-4900744 on Build GRCh38
rs370199113 Chr.17: 4900699-4900699 on Build GRCh38
rs780373222 Chr.17: 4900547-4900547 on Build GRCh38
rs764752088 Chr.17: 4900937-4900937 on Build GRCh38
rs964750991 Chr.17: 4900581-4900581 on Build GRCh38
rs776407381 Chr.17: 4900929-4900929 on Build GRCh38
rs201841042 Chr.17: 4900889-4900889 on Build GRCh38
rs576292648 Chr.17: 4900554-4900554 on Build GRCh38
rs201042568 Chr.17: 4900945-4900945 on Build GRCh38
rs780429979 Chr.17: 4900864-4900864 on Build GRCh38
rs992581409 Chr.17: 4900638-4900638 on Build GRCh38
rs1040580878 Chr.17: 4900859-4900859 on Build GRCh38
rs983611047 Chr.17: 4900631-4900631 on Build GRCh38
rs750981766 Chr.17: 4900862-4900862 on Build GRCh38
rs761428919 Chr.17: 4900750-4900750 on Build GRCh38
rs532968770 Chr.17: 4900658-4900658 on Build GRCh38
rs916950933 Chr.17: 4900654-4900654 on Build GRCh38
rs1009114560 Chr.17: 4900704-4900704 on Build GRCh38
rs1015178267 Chr.17: 4900911-4900911 on Build GRCh38
rs901628873 Chr.17: 4900700-4900700 on Build GRCh38
rs776585678 Chr.17: 4900811-4900811 on Build GRCh38
rs929229289 Chr.17: 4900689-4900689 on Build GRCh38
rs969873887 Chr.17: 4900951-4900951 on Build GRCh38
rs779186524 Chr.17: 4900798-4900798 on Build GRCh38
rs747224472 Chr.17: 4900810-4900810 on Build GRCh38
rs756844933 Chr.17: 4900854-4900854 on Build GRCh38
rs1044057628 Chr.17: 4900682-4900682 on Build GRCh38
rs561299262 Chr.17: 4900575-4900575 on Build GRCh38
rs370019023 Chr.17: 4900805-4900805 on Build GRCh38
rs547550204 Chr.17: 4900708-4900708 on Build GRCh38
rs138412824 Chr.17: 4900694-4900694 on Build GRCh38
rs759372801 Chr.17: 4900822-4900822 on Build GRCh38
rs772869952 Chr.17: 4900775-4900776 on Build GRCh38
rs12942540 Chr.17: 4900778-4900778 on Build GRCh38
rs187741702 Chr.17: 4900653-4900653 on Build GRCh38
rs534347172 Chr.17: 4900703-4900703 on Build GRCh38
rs752447974 Chr.17: 4900776-4900776 on Build GRCh38
rs919271118 Chr.17: 4900796-4900796 on Build GRCh38
rs745821689 Chr.17: 4900892-4900892 on Build GRCh38
rs768177094 Chr.17: 4900923-4900923 on Build GRCh38
rs1008983151 Chr.17: 4900566-4900566 on Build GRCh38
rs1019033479 Chr.17: 4900713-4900713 on Build GRCh38
rs571849061 Chr.17: 4900595-4900595 on Build GRCh38
rs1048944477 Chr.17: 4900690-4900690 on Build GRCh38
rs759470814 Chr.17: 4900769-4900769 on Build GRCh38
rs1015081055 Chr.17: 4900753-4900753 on Build GRCh38
rs755170993 Chr.17: 4900879-4900879 on Build GRCh38
rs751670974 Chr.17: 4900597-4900597 on Build GRCh38
rs760314762 Chr.17: 4900915-4900915 on Build GRCh38
rs55735639 Chr.17: 4900721-4900721 on Build GRCh38
rs774497563 Chr.17: 4900821-4900821 on Build GRCh38
rs549915994 Chr.17: 4900676-4900676 on Build GRCh38

View Full Product Details