Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 10633508-10634004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH3
Entrez Gene ID
4621
Gene Name
myosin, heavy chain 3, skeletal muscle, embryonic
Gene Aliases
DA2A, DA2B, DA8, HEMHC, MYHC-EMB, MYHSE1, SMHCE
UniGene
Hs.440895
Gene Chromosome Location
Chr.17: 10628526-10679657 on Build GRCh38
Transcripts
NM_002470.3 XM_011523870.2 XM_011523871.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
975383 c.4543G>T p.E1515* Chr.17: 10633695-10633695 on Build GRCh38
5385665 c.4573C>T p.H1525Y Chr.17: 10633665-10633665 on Build GRCh38
1679586 c.4592G>A p.R1531K Chr.17: 10633646-10633646 on Build GRCh38
5479258 c.4595A>C p.K1532T Chr.17: 10633643-10633643 on Build GRCh38
3969812 c.4616C>G p.A1539G Chr.17: 10633622-10633622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374924521 Chr.17: 10633742-10633742 on Build GRCh38
rs771091159 Chr.17: 10633545-10633545 on Build GRCh38
rs1022552868 Chr.17: 10633975-10633975 on Build GRCh38
rs922323624 Chr.17: 10633843-10633843 on Build GRCh38
rs370151413 Chr.17: 10633695-10633695 on Build GRCh38
rs781315837 Chr.17: 10633852-10633852 on Build GRCh38
rs1057376121 Chr.17: 10633937-10633937 on Build GRCh38
rs899721074 Chr.17: 10633607-10633607 on Build GRCh38
rs549712445 Chr.17: 10633952-10633952 on Build GRCh38
rs527420177 Chr.17: 10633876-10633876 on Build GRCh38
rs1048299702 Chr.17: 10633884-10633884 on Build GRCh38
rs888336601 Chr.17: 10633623-10633623 on Build GRCh38
rs557870001 Chr.17: 10633664-10633664 on Build GRCh38
rs377200387 Chr.17: 10633549-10633549 on Build GRCh38
rs1004487948 Chr.17: 10633697-10633697 on Build GRCh38
rs753040463 Chr.17: 10633737-10633737 on Build GRCh38
rs73976873 Chr.17: 10633757-10633757 on Build GRCh38
rs577757117 Chr.17: 10633847-10633847 on Build GRCh38
rs931184141 Chr.17: 10633875-10633875 on Build GRCh38
rs77479161 Chr.17: 10633848-10633848 on Build GRCh38
rs756851580 Chr.17: 10633574-10633574 on Build GRCh38
rs374383577 Chr.17: 10633740-10633740 on Build GRCh38
rs567183603 Chr.17: 10633769-10633769 on Build GRCh38
rs957600154 Chr.17: 10633770-10633770 on Build GRCh38
rs1039700897 Chr.17: 10633853-10633854 on Build GRCh38
rs542772988 Chr.17: 10633906-10633906 on Build GRCh38
rs998948148 Chr.17: 10633760-10633760 on Build GRCh38
rs760603613 Chr.17: 10633687-10633687 on Build GRCh38
rs887062970 Chr.17: 10633901-10633901 on Build GRCh38
rs778304936 Chr.17: 10633612-10633612 on Build GRCh38
rs548988117 Chr.17: 10633529-10633529 on Build GRCh38
rs753865328 Chr.17: 10633707-10633707 on Build GRCh38
rs781349619 Chr.17: 10633602-10633602 on Build GRCh38
rs186769427 Chr.17: 10633869-10633869 on Build GRCh38
rs1033137495 Chr.17: 10633958-10633958 on Build GRCh38
rs1047032918 Chr.17: 10633608-10633608 on Build GRCh38
rs778792067 Chr.17: 10633745-10633745 on Build GRCh38
rs148603186 Chr.17: 10633951-10633951 on Build GRCh38
rs199976698 Chr.17: 10633543-10633543 on Build GRCh38
rs757985950 Chr.17: 10633733-10633733 on Build GRCh38
rs373814637 Chr.17: 10633966-10633966 on Build GRCh38
rs761871162 Chr.17: 10633710-10633710 on Build GRCh38
rs1034339254 Chr.17: 10633718-10633718 on Build GRCh38
rs560935908 Chr.17: 10633915-10633915 on Build GRCh38
rs372502153 Chr.17: 10633734-10633734 on Build GRCh38
rs758088341 Chr.17: 10633593-10633593 on Build GRCh38
rs763178595 Chr.17: 10633724-10633727 on Build GRCh38
rs377463916 Chr.17: 10633636-10633636 on Build GRCh38
rs764133412 Chr.17: 10633568-10633568 on Build GRCh38
rs775143020 Chr.17: 10633685-10633685 on Build GRCh38
rs74406856 Chr.17: 10633603-10633603 on Build GRCh38
rs1033250370 Chr.17: 10633761-10633761 on Build GRCh38
rs746767338 Chr.17: 10633756-10633756 on Build GRCh38
rs989590675 Chr.17: 10633774-10633774 on Build GRCh38
rs201922765 Chr.17: 10633746-10633746 on Build GRCh38
rs759840842 Chr.17: 10633552-10633552 on Build GRCh38
rs749336793 Chr.17: 10633629-10633629 on Build GRCh38
rs1045803427 Chr.17: 10633923-10633923 on Build GRCh38
rs368963921 Chr.17: 10633743-10633743 on Build GRCh38
rs73281077 Chr.17: 10633564-10633564 on Build GRCh38
rs886052579 Chr.17: 10633591-10633591 on Build GRCh38
rs778143713 Chr.17: 10633741-10633741 on Build GRCh38
rs770275791 Chr.17: 10633611-10633611 on Build GRCh38
rs762799099 Chr.17: 10633786-10633786 on Build GRCh38
rs774513052 Chr.17: 10633632-10633632 on Build GRCh38
rs375163919 Chr.17: 10633585-10633585 on Build GRCh38
rs567526319 Chr.17: 10633530-10633530 on Build GRCh38
rs760291583 Chr.17: 10633540-10633540 on Build GRCh38
rs776073492 Chr.17: 10633569-10633569 on Build GRCh38
rs568916955 Chr.17: 10633576-10633576 on Build GRCh38
rs761781341 Chr.17: 10633978-10633978 on Build GRCh38
rs377123796 Chr.17: 10633748-10633748 on Build GRCh38
rs941286184 Chr.17: 10633868-10633868 on Build GRCh38
rs1007816722 Chr.17: 10633972-10633972 on Build GRCh38
rs752596050 Chr.17: 10633565-10633565 on Build GRCh38
rs761672631 Chr.17: 10633766-10633766 on Build GRCh38
rs78946532 Chr.17: 10633825-10633825 on Build GRCh38
rs544927489 Chr.17: 10633804-10633804 on Build GRCh38
rs573340718 Chr.17: 10633728-10633728 on Build GRCh38
rs765208722 Chr.17: 10633749-10633749 on Build GRCh38
rs760685516 Chr.17: 10633567-10633567 on Build GRCh38
rs764600406 Chr.17: 10633720-10633720 on Build GRCh38

View Full Product Details