Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 16024925-16025435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC19
Entrez Gene ID
54902
Gene Name
tetratricopeptide repeat domain 19
Gene Aliases
2010204O13Rik, MC3DN2
UniGene
Hs.462316
Gene Chromosome Location
Chr.17: 15999380-16045428 on Build GRCh38
Transcripts
NM_001271420.1 NM_017775.3 XM_017024801.1 XM_017024802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7105222 c.1194+2T>A p.? Chr.17: 16025173-16025173 on Build GRCh38
4064142 c.1094C>T p.A365V Chr.17: 16025071-16025071 on Build GRCh38
5759322 c.1055A>T p.N352I Chr.17: 16025032-16025032 on Build GRCh38
7150088 c.1065C>T p.L355L Chr.17: 16025042-16025042 on Build GRCh38
7395561 c.1121C>T p.P374L Chr.17: 16025098-16025098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568597965 Chr.17: 16025021-16025021 on Build GRCh38
rs554437943 Chr.17: 16025199-16025199 on Build GRCh38
rs1049717331 Chr.17: 16025351-16025351 on Build GRCh38
rs761201874 Chr.17: 16025036-16025036 on Build GRCh38
rs893322511 Chr.17: 16025241-16025241 on Build GRCh38
rs889837116 Chr.17: 16025149-16025149 on Build GRCh38
rs769438082 Chr.17: 16025011-16025011 on Build GRCh38
rs768968665 Chr.17: 16025004-16025004 on Build GRCh38
rs1042653066 Chr.17: 16025313-16025313 on Build GRCh38
rs140778368 Chr.17: 16024975-16024975 on Build GRCh38
rs999066071 Chr.17: 16025213-16025213 on Build GRCh38
rs377635304 Chr.17: 16025177-16025177 on Build GRCh38
rs751667586 Chr.17: 16025080-16025080 on Build GRCh38
rs770827831 Chr.17: 16025129-16025129 on Build GRCh38
rs762239880 Chr.17: 16025064-16025064 on Build GRCh38
rs150104000 Chr.17: 16025069-16025069 on Build GRCh38
rs773512385 Chr.17: 16025193-16025193 on Build GRCh38
rs926955965 Chr.17: 16025277-16025277 on Build GRCh38
rs146462096 Chr.17: 16025096-16025096 on Build GRCh38
rs767014443 Chr.17: 16025183-16025183 on Build GRCh38
rs773851208 Chr.17: 16025045-16025045 on Build GRCh38
rs937926668 Chr.17: 16025325-16025325 on Build GRCh38
rs972608359 Chr.17: 16025197-16025197 on Build GRCh38
rs140719848 Chr.17: 16025053-16025053 on Build GRCh38
rs150945703 Chr.17: 16025032-16025032 on Build GRCh38
rs752071839 Chr.17: 16025186-16025186 on Build GRCh38
rs776438746 Chr.17: 16025271-16025271 on Build GRCh38
rs376969540 Chr.17: 16025070-16025070 on Build GRCh38
rs2301651 Chr.17: 16025286-16025286 on Build GRCh38
rs770164715 Chr.17: 16025189-16025190 on Build GRCh38
rs1008165216 Chr.17: 16025257-16025257 on Build GRCh38
rs748252883 Chr.17: 16025111-16025111 on Build GRCh38
rs771203933 Chr.17: 16025260-16025262 on Build GRCh38
rs753021454 Chr.17: 16025212-16025212 on Build GRCh38
rs794726692 Chr.17: 16025169-16025169 on Build GRCh38
rs896668351 Chr.17: 16025092-16025092 on Build GRCh38
rs965817871 Chr.17: 16025272-16025272 on Build GRCh38
rs779499875 Chr.17: 16024992-16024992 on Build GRCh38
rs149658292 Chr.17: 16025063-16025063 on Build GRCh38
rs918071691 Chr.17: 16025308-16025308 on Build GRCh38
rs758965029 Chr.17: 16025073-16025073 on Build GRCh38
rs747996216 Chr.17: 16025010-16025010 on Build GRCh38
rs141892030 Chr.17: 16025127-16025127 on Build GRCh38
rs952374266 Chr.17: 16025227-16025227 on Build GRCh38
rs1000900255 Chr.17: 16025139-16025139 on Build GRCh38
rs758010059 Chr.17: 16025196-16025196 on Build GRCh38
rs1045743259 Chr.17: 16025189-16025189 on Build GRCh38
rs754203038 Chr.17: 16024971-16024971 on Build GRCh38
rs936920185 Chr.17: 16025332-16025332 on Build GRCh38
rs755092242 Chr.17: 16025191-16025191 on Build GRCh38
rs759231114 Chr.17: 16025049-16025049 on Build GRCh38
rs147111211 Chr.17: 16025160-16025160 on Build GRCh38
rs775218326 Chr.17: 16025150-16025150 on Build GRCh38
rs756286251 Chr.17: 16025217-16025217 on Build GRCh38
rs945554055 Chr.17: 16025309-16025309 on Build GRCh38
rs771445696 Chr.17: 16025143-16025143 on Build GRCh38
rs747300891 Chr.17: 16025003-16025003 on Build GRCh38
rs760228229 Chr.17: 16025159-16025159 on Build GRCh38
rs368258821 Chr.17: 16025027-16025027 on Build GRCh38
rs1056600951 Chr.17: 16025081-16025081 on Build GRCh38
rs780976534 Chr.17: 16025382-16025382 on Build GRCh38
rs991091374 Chr.17: 16025393-16025393 on Build GRCh38
rs371498054 Chr.17: 16025074-16025074 on Build GRCh38
rs745407849 Chr.17: 16025138-16025138 on Build GRCh38
rs1042684635 Chr.17: 16025106-16025106 on Build GRCh38
rs2301652 Chr.17: 16025270-16025270 on Build GRCh38
rs8081706 Chr.17: 16025288-16025288 on Build GRCh38
rs78882347 Chr.17: 16025098-16025098 on Build GRCh38
rs950327216 Chr.17: 16025310-16025310 on Build GRCh38
rs773680046 Chr.17: 16025172-16025172 on Build GRCh38
rs192576331 Chr.17: 16025320-16025320 on Build GRCh38
rs766798071 Chr.17: 16025043-16025043 on Build GRCh38
rs773904382 Chr.17: 16025133-16025133 on Build GRCh38
rs781506238 Chr.17: 16025099-16025099 on Build GRCh38
rs188950109 Chr.17: 16024954-16024954 on Build GRCh38
rs755952117 Chr.17: 16025121-16025121 on Build GRCh38
rs780896858 Chr.17: 16025006-16025006 on Build GRCh38
rs761945008 Chr.17: 16025176-16025176 on Build GRCh38
rs542249659 Chr.17: 16025367-16025367 on Build GRCh38
rs373260669 Chr.17: 16025165-16025165 on Build GRCh38
rs762819874 Chr.17: 16025037-16025037 on Build GRCh38
rs750890125 Chr.17: 16024993-16024993 on Build GRCh38
rs780264767 Chr.17: 16024997-16024997 on Build GRCh38
rs777418331 Chr.17: 16025123-16025123 on Build GRCh38
rs1028821942 Chr.17: 16025148-16025148 on Build GRCh38
rs1026888102 Chr.17: 16025215-16025215 on Build GRCh38

View Full Product Details