Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28232633-28233144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC8
Entrez Gene ID
22826
Gene Name
DnaJ heat shock protein family (Hsp40) member C8
Gene Aliases
HSPC331, SPF31
UniGene
Hs.433540
Gene Chromosome Location
Chr.1: 28200278-28233031 on Build GRCh38
Transcripts
NM_014280.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7175586 c.38G>T p.G13V Chr.1: 28232961-28232961 on Build GRCh38
2236649 c.21C>G p.S7R Chr.1: 28232978-28232978 on Build GRCh38
1341571 c.20G>A p.S7N Chr.1: 28232979-28232979 on Build GRCh38
5977932 c.18G>C p.E6D Chr.1: 28232981-28232981 on Build GRCh38
4969279 c.21C>T p.S7S Chr.1: 28232978-28232978 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764436390 Chr.1: 28232957-28232957 on Build GRCh38
rs989021835 Chr.1: 28233089-28233089 on Build GRCh38
rs1029855092 Chr.1: 28232660-28232660 on Build GRCh38
rs773473828 Chr.1: 28233008-28233008 on Build GRCh38
rs373798993 Chr.1: 28232896-28232896 on Build GRCh38
rs200720254 Chr.1: 28233037-28233037 on Build GRCh38
rs772287428 Chr.1: 28232938-28232938 on Build GRCh38
rs754354594 Chr.1: 28233093-28233093 on Build GRCh38
rs946602917 Chr.1: 28232688-28232688 on Build GRCh38
rs568098029 Chr.1: 28232727-28232727 on Build GRCh38
rs987911698 Chr.1: 28232666-28232666 on Build GRCh38
rs913674804 Chr.1: 28232837-28232837 on Build GRCh38
rs950163652 Chr.1: 28232766-28232766 on Build GRCh38
rs182710353 Chr.1: 28232736-28232736 on Build GRCh38
rs751501441 Chr.1: 28232965-28232965 on Build GRCh38
rs996674593 Chr.1: 28232710-28232710 on Build GRCh38
rs1040767505 Chr.1: 28232875-28232875 on Build GRCh38
rs955260123 Chr.1: 28232927-28232927 on Build GRCh38
rs534588684 Chr.1: 28232754-28232754 on Build GRCh38
rs1053962151 Chr.1: 28232836-28232836 on Build GRCh38
rs199527099 Chr.1: 28232967-28232967 on Build GRCh38
rs1055872937 Chr.1: 28232723-28232723 on Build GRCh38
rs760060976 Chr.1: 28232934-28232934 on Build GRCh38
rs776938007 Chr.1: 28232994-28232994 on Build GRCh38
rs756946326 Chr.1: 28233035-28233035 on Build GRCh38
rs551820261 Chr.1: 28232807-28232807 on Build GRCh38
rs752577422 Chr.1: 28233018-28233018 on Build GRCh38
rs367568646 Chr.1: 28233039-28233039 on Build GRCh38
rs537267717 Chr.1: 28232858-28232858 on Build GRCh38
rs537898253 Chr.1: 28232744-28232744 on Build GRCh38
rs370236951 Chr.1: 28233006-28233006 on Build GRCh38
rs372981004 Chr.1: 28232954-28232954 on Build GRCh38
rs1051168002 Chr.1: 28233066-28233066 on Build GRCh38
rs549914312 Chr.1: 28232717-28232717 on Build GRCh38
rs913741046 Chr.1: 28232676-28232676 on Build GRCh38
rs890831614 Chr.1: 28233087-28233087 on Build GRCh38
rs573802098 Chr.1: 28232884-28232884 on Build GRCh38
rs559509 Chr.1: 28232721-28232721 on Build GRCh38
rs777115118 Chr.1: 28232790-28232790 on Build GRCh38
rs758827379 Chr.1: 28233041-28233041 on Build GRCh38
rs955163889 Chr.1: 28232664-28232664 on Build GRCh38
rs777415384 Chr.1: 28232969-28232969 on Build GRCh38
rs753962397 Chr.1: 28232958-28232958 on Build GRCh38
rs564385762 Chr.1: 28232680-28232680 on Build GRCh38
rs1038548632 Chr.1: 28233050-28233050 on Build GRCh38
rs571874240 Chr.1: 28232834-28232834 on Build GRCh38
rs868583439 Chr.1: 28232783-28232783 on Build GRCh38
rs138050264 Chr.1: 28232925-28232925 on Build GRCh38
rs778428016 Chr.1: 28232978-28232978 on Build GRCh38
rs557783227 Chr.1: 28232981-28232981 on Build GRCh38
rs780543903 Chr.1: 28233047-28233047 on Build GRCh38
rs980198146 Chr.1: 28232792-28232792 on Build GRCh38
rs763853497 Chr.1: 28232797-28232797 on Build GRCh38
rs926120639 Chr.1: 28232793-28232793 on Build GRCh38
rs979301390 Chr.1: 28232973-28232973 on Build GRCh38
rs763815168 Chr.1: 28233025-28233025 on Build GRCh38
rs879175948 Chr.1: 28232658-28232658 on Build GRCh38
rs899483905 Chr.1: 28232865-28232865 on Build GRCh38
rs367617170 Chr.1: 28232882-28232882 on Build GRCh38
rs772688253 Chr.1: 28232980-28232980 on Build GRCh38
rs755878774 Chr.1: 28233024-28233024 on Build GRCh38
rs768908939 Chr.1: 28232987-28232987 on Build GRCh38
rs533015720 Chr.1: 28232716-28232716 on Build GRCh38
rs747312265 Chr.1: 28232650-28232650 on Build GRCh38
rs563394856 Chr.1: 28232720-28232720 on Build GRCh38
rs759100471 Chr.1: 28232806-28232806 on Build GRCh38
rs748949399 Chr.1: 28232974-28232974 on Build GRCh38
rs510803 Chr.1: 28233063-28233063 on Build GRCh38
rs1015572538 Chr.1: 28232731-28232731 on Build GRCh38
rs759377790 Chr.1: 28233011-28233011 on Build GRCh38
rs900058672 Chr.1: 28233062-28233062 on Build GRCh38
rs775966995 Chr.1: 28232948-28232948 on Build GRCh38
rs376755697 Chr.1: 28232912-28232912 on Build GRCh38
rs780827303 Chr.1: 28232966-28232966 on Build GRCh38
rs772144987 Chr.1: 28232974-28232975 on Build GRCh38
rs908690972 Chr.1: 28233021-28233021 on Build GRCh38
rs770382174 Chr.1: 28232975-28232975 on Build GRCh38
rs185422705 Chr.1: 28233049-28233049 on Build GRCh38
rs765422686 Chr.1: 28232964-28232964 on Build GRCh38
rs1049842731 Chr.1: 28232826-28232826 on Build GRCh38
rs201460078 Chr.1: 28232968-28232968 on Build GRCh38
rs536391147 Chr.1: 28232893-28232893 on Build GRCh38
rs1006063190 Chr.1: 28232726-28232726 on Build GRCh38
rs373769279 Chr.1: 28233014-28233014 on Build GRCh38
rs762157336 Chr.1: 28233002-28233002 on Build GRCh38
rs926564834 Chr.1: 28232976-28232976 on Build GRCh38
rs901117508 Chr.1: 28232706-28232706 on Build GRCh38
rs761918659 Chr.1: 28232963-28232963 on Build GRCh38
rs745810562 Chr.1: 28232908-28232908 on Build GRCh38
rs945010398 Chr.1: 28232874-28232874 on Build GRCh38
rs1038655919 Chr.1: 28232864-28232864 on Build GRCh38
rs769524295 Chr.1: 28232703-28232703 on Build GRCh38
rs961658127 Chr.1: 28232762-28232762 on Build GRCh38

View Full Product Details