Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21001297-21001813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP22
Entrez Gene ID
23326
Gene Name
ubiquitin specific peptidase 22
Gene Aliases
USP3L
UniGene
Hs.462492
Gene Chromosome Location
Chr.17: 20999593-21043409 on Build GRCh38
Transcripts
NM_015276.1 XM_005256575.2

SNP Detail

SNP ID Chromosome Location
rs1042600914 Chr.17: 21001781-21001781 on Build GRCh38
rs957535963 Chr.17: 21001734-21001734 on Build GRCh38
rs754739108 Chr.17: 21001733-21001733 on Build GRCh38
rs1033172417 Chr.17: 21001399-21001399 on Build GRCh38
rs367691583 Chr.17: 21001382-21001382 on Build GRCh38
rs11541318 Chr.17: 21001571-21001571 on Build GRCh38
rs929026976 Chr.17: 21001633-21001633 on Build GRCh38
rs1034965662 Chr.17: 21001558-21001558 on Build GRCh38
rs893608514 Chr.17: 21001482-21001482 on Build GRCh38
rs943277120 Chr.17: 21001689-21001689 on Build GRCh38
rs943246082 Chr.17: 21001464-21001464 on Build GRCh38
rs200572625 Chr.17: 21001634-21001634 on Build GRCh38
rs1021079492 Chr.17: 21001511-21001511 on Build GRCh38
rs950168716 Chr.17: 21001485-21001485 on Build GRCh38
rs911534481 Chr.17: 21001635-21001635 on Build GRCh38
rs888816770 Chr.17: 21001642-21001642 on Build GRCh38
rs929019207 Chr.17: 21001455-21001456 on Build GRCh38
rs767518710 Chr.17: 21001397-21001397 on Build GRCh38
rs1048975415 Chr.17: 21001528-21001528 on Build GRCh38
rs772929165 Chr.17: 21001372-21001372 on Build GRCh38
rs750366952 Chr.17: 21001516-21001516 on Build GRCh38
rs3612 Chr.17: 21001771-21001771 on Build GRCh38
rs927338775 Chr.17: 21001494-21001494 on Build GRCh38
rs149307123 Chr.17: 21001549-21001549 on Build GRCh38
rs1051413811 Chr.17: 21001456-21001456 on Build GRCh38
rs564192942 Chr.17: 21001593-21001593 on Build GRCh38
rs1038900595 Chr.17: 21001480-21001480 on Build GRCh38
rs918997377 Chr.17: 21001432-21001432 on Build GRCh38
rs372156656 Chr.17: 21001586-21001586 on Build GRCh38
rs779986466 Chr.17: 21001711-21001711 on Build GRCh38
rs973302889 Chr.17: 21001627-21001628 on Build GRCh38
rs11541315 Chr.17: 21001716-21001716 on Build GRCh38
rs73301793 Chr.17: 21001787-21001787 on Build GRCh38
rs375357743 Chr.17: 21001671-21001671 on Build GRCh38
rs182025734 Chr.17: 21001661-21001661 on Build GRCh38
rs902620873 Chr.17: 21001783-21001783 on Build GRCh38
rs957777210 Chr.17: 21001407-21001407 on Build GRCh38
rs997289153 Chr.17: 21001595-21001595 on Build GRCh38
rs1044410 Chr.17: 21001356-21001356 on Build GRCh38
rs12452757 Chr.17: 21001374-21001374 on Build GRCh38
rs778557855 Chr.17: 21001761-21001761 on Build GRCh38
rs890399469 Chr.17: 21001456-21001458 on Build GRCh38
rs766334723 Chr.17: 21001570-21001570 on Build GRCh38
rs1038932014 Chr.17: 21001719-21001719 on Build GRCh38
rs543662219 Chr.17: 21001628-21001628 on Build GRCh38
rs981391690 Chr.17: 21001493-21001493 on Build GRCh38
rs189344461 Chr.17: 21001512-21001512 on Build GRCh38
rs553528705 Chr.17: 21001517-21001517 on Build GRCh38
rs1036082634 Chr.17: 21001572-21001572 on Build GRCh38
rs573999731 Chr.17: 21001442-21001442 on Build GRCh38
rs529212279 Chr.17: 21001748-21001748 on Build GRCh38
rs893713002 Chr.17: 21001764-21001764 on Build GRCh38
rs988870751 Chr.17: 21001429-21001429 on Build GRCh38
rs547657995 Chr.17: 21001752-21001752 on Build GRCh38
rs555131762 Chr.17: 21001729-21001729 on Build GRCh38
rs1007112929 Chr.17: 21001657-21001657 on Build GRCh38
rs531398656 Chr.17: 21001625-21001625 on Build GRCh38
rs184925642 Chr.17: 21001373-21001373 on Build GRCh38
rs753740378 Chr.17: 21001732-21001732 on Build GRCh38

View Full Product Details