Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28353616-28354120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLDIP2
Entrez Gene ID
26073
Gene Name
DNA polymerase delta interacting protein 2
Gene Aliases
PDIP38, POLD4, p38
UniGene
Hs.241543
Gene Chromosome Location
Chr.17: 28346628-28357589 on Build GRCh38
Transcripts
NM_001290145.1 NM_015584.4

SNP Detail

SNP ID Chromosome Location
rs782424258 Chr.17: 28353652-28353652 on Build GRCh38
rs1053670252 Chr.17: 28353812-28353812 on Build GRCh38
rs79354185 Chr.17: 28353923-28353923 on Build GRCh38
rs149683227 Chr.17: 28353795-28353795 on Build GRCh38
rs781631764 Chr.17: 28353696-28353696 on Build GRCh38
rs932312564 Chr.17: 28353782-28353782 on Build GRCh38
rs576945354 Chr.17: 28353957-28353957 on Build GRCh38
rs1027684784 Chr.17: 28354010-28354010 on Build GRCh38
rs139525737 Chr.17: 28353781-28353781 on Build GRCh38
rs532435280 Chr.17: 28353958-28353958 on Build GRCh38
rs376731715 Chr.17: 28353701-28353701 on Build GRCh38
rs782396384 Chr.17: 28353759-28353759 on Build GRCh38
rs75275681 Chr.17: 28353907-28353907 on Build GRCh38
rs769813603 Chr.17: 28353949-28353949 on Build GRCh38
rs574606608 Chr.17: 28353976-28353976 on Build GRCh38
rs782023901 Chr.17: 28353656-28353656 on Build GRCh38
rs79183819 Chr.17: 28353803-28353803 on Build GRCh38
rs1044488784 Chr.17: 28353885-28353885 on Build GRCh38
rs782601762 Chr.17: 28353732-28353732 on Build GRCh38
rs376470070 Chr.17: 28353666-28353666 on Build GRCh38
rs566519993 Chr.17: 28353914-28353914 on Build GRCh38
rs201741010 Chr.17: 28353758-28353758 on Build GRCh38
rs895987185 Chr.17: 28353834-28353834 on Build GRCh38
rs114122431 Chr.17: 28354011-28354011 on Build GRCh38
rs782237926 Chr.17: 28353790-28353790 on Build GRCh38
rs905984161 Chr.17: 28353890-28353890 on Build GRCh38
rs782194233 Chr.17: 28353734-28353734 on Build GRCh38
rs782485145 Chr.17: 28353720-28353720 on Build GRCh38
rs782153561 Chr.17: 28353811-28353811 on Build GRCh38
rs1014313036 Chr.17: 28353843-28353843 on Build GRCh38
rs782692450 Chr.17: 28353742-28353742 on Build GRCh38
rs558400009 Chr.17: 28353930-28353930 on Build GRCh38
rs371729042 Chr.17: 28353808-28353808 on Build GRCh38
rs782734726 Chr.17: 28353685-28353685 on Build GRCh38
rs371044724 Chr.17: 28353675-28353675 on Build GRCh38
rs372448666 Chr.17: 28353658-28353658 on Build GRCh38
rs182299035 Chr.17: 28353971-28353971 on Build GRCh38
rs537613872 Chr.17: 28353963-28353963 on Build GRCh38
rs375110077 Chr.17: 28354090-28354090 on Build GRCh38
rs564958680 Chr.17: 28353713-28353713 on Build GRCh38
rs782433068 Chr.17: 28353740-28353740 on Build GRCh38
rs781974815 Chr.17: 28353669-28353669 on Build GRCh38
rs781893839 Chr.17: 28353690-28353690 on Build GRCh38
rs782179375 Chr.17: 28353650-28353650 on Build GRCh38
rs554543661 Chr.17: 28354017-28354017 on Build GRCh38
rs773293480 Chr.17: 28354037-28354037 on Build GRCh38
rs532631156 Chr.17: 28353640-28353640 on Build GRCh38

View Full Product Details