Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28527163-28527661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXN1
Entrez Gene ID
8456
Gene Name
forkhead box N1
Gene Aliases
FKHL20, RONU, WHN
UniGene
Hs.58611
Gene Chromosome Location
Chr.17: 28506211-28538900 on Build GRCh38
Transcripts
NM_003593.2 XM_005258046.3 XM_011525358.1 XM_011525359.1 XM_011525362.1 XM_011525367.1 XM_011525368.2 XM_011525369.2 XM_011525370.2 XM_017025228.1 XM_017025229.1 XM_017025230.1 XM_017025231.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5531410 c.681C>T p.S227S Chr.17: 28527343-28527343 on Build GRCh38
4774209 c.593C>A p.S198* Chr.17: 28527255-28527255 on Build GRCh38
4960863 c.632C>T p.A211V Chr.17: 28527294-28527294 on Build GRCh38
1207437 c.655C>T p.P219S Chr.17: 28527317-28527317 on Build GRCh38
6666415 c.683A>G p.Q228R Chr.17: 28527345-28527345 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896211050 Chr.17: 28527436-28527436 on Build GRCh38
rs951567061 Chr.17: 28527518-28527518 on Build GRCh38
rs539716042 Chr.17: 28527412-28527413 on Build GRCh38
rs576257217 Chr.17: 28527389-28527389 on Build GRCh38
rs780747628 Chr.17: 28527392-28527392 on Build GRCh38
rs576519250 Chr.17: 28527525-28527525 on Build GRCh38
rs189412912 Chr.17: 28527413-28527413 on Build GRCh38
rs912645688 Chr.17: 28527561-28527561 on Build GRCh38
rs376187163 Chr.17: 28527248-28527248 on Build GRCh38
rs746253550 Chr.17: 28527230-28527230 on Build GRCh38
rs756501832 Chr.17: 28527231-28527231 on Build GRCh38
rs1002428746 Chr.17: 28527289-28527289 on Build GRCh38
rs568219000 Chr.17: 28527330-28527330 on Build GRCh38
rs541918281 Chr.17: 28527632-28527632 on Build GRCh38
rs764151407 Chr.17: 28527204-28527204 on Build GRCh38
rs756591659 Chr.17: 28527369-28527369 on Build GRCh38
rs921844080 Chr.17: 28527408-28527408 on Build GRCh38
rs531929794 Chr.17: 28527194-28527194 on Build GRCh38
rs905182420 Chr.17: 28527445-28527445 on Build GRCh38
rs779114720 Chr.17: 28527408-28527409 on Build GRCh38
rs995750235 Chr.17: 28527504-28527504 on Build GRCh38
rs144560330 Chr.17: 28527545-28527545 on Build GRCh38
rs35009868 Chr.17: 28527395-28527395 on Build GRCh38
rs985994799 Chr.17: 28527409-28527410 on Build GRCh38
rs781411937 Chr.17: 28527223-28527223 on Build GRCh38
rs535522153 Chr.17: 28527306-28527306 on Build GRCh38
rs962960245 Chr.17: 28527273-28527273 on Build GRCh38
rs1043655804 Chr.17: 28527410-28527410 on Build GRCh38
rs369200003 Chr.17: 28527219-28527219 on Build GRCh38
rs1014095205 Chr.17: 28527193-28527193 on Build GRCh38
rs975899241 Chr.17: 28527583-28527583 on Build GRCh38
rs779412826 Chr.17: 28527387-28527387 on Build GRCh38
rs377176881 Chr.17: 28527388-28527388 on Build GRCh38
rs1046531148 Chr.17: 28527428-28527428 on Build GRCh38
rs185748978 Chr.17: 28527239-28527239 on Build GRCh38
rs780561388 Chr.17: 28527379-28527379 on Build GRCh38
rs962690381 Chr.17: 28527480-28527480 on Build GRCh38
rs759297920 Chr.17: 28527311-28527311 on Build GRCh38
rs368663316 Chr.17: 28527228-28527228 on Build GRCh38
rs777135993 Chr.17: 28527259-28527259 on Build GRCh38
rs757328321 Chr.17: 28527400-28527401 on Build GRCh38
rs1011894564 Chr.17: 28527256-28527256 on Build GRCh38
rs368285745 Chr.17: 28527351-28527351 on Build GRCh38
rs951542551 Chr.17: 28527322-28527322 on Build GRCh38
rs139245723 Chr.17: 28527517-28527517 on Build GRCh38
rs746471609 Chr.17: 28527272-28527272 on Build GRCh38
rs1056246235 Chr.17: 28527426-28527426 on Build GRCh38
rs1012333990 Chr.17: 28527446-28527446 on Build GRCh38
rs763958930 Chr.17: 28527347-28527347 on Build GRCh38
rs749372001 Chr.17: 28527271-28527272 on Build GRCh38
rs771775724 Chr.17: 28527250-28527250 on Build GRCh38
rs758508712 Chr.17: 28527200-28527200 on Build GRCh38
rs547559375 Chr.17: 28527337-28527337 on Build GRCh38
rs1023261921 Chr.17: 28527460-28527460 on Build GRCh38
rs775469489 Chr.17: 28527336-28527336 on Build GRCh38
rs567994435 Chr.17: 28527292-28527292 on Build GRCh38
rs886052757 Chr.17: 28527273-28527273 on Build GRCh38
rs565781236 Chr.17: 28527358-28527358 on Build GRCh38
rs79861097 Chr.17: 28527383-28527383 on Build GRCh38
rs754455211 Chr.17: 28527380-28527380 on Build GRCh38
rs769545823 Chr.17: 28527315-28527315 on Build GRCh38
rs774969583 Chr.17: 28527461-28527461 on Build GRCh38
rs555037603 Chr.17: 28527190-28527190 on Build GRCh38
rs749782134 Chr.17: 28527241-28527241 on Build GRCh38
rs751862988 Chr.17: 28527218-28527218 on Build GRCh38
rs776297762 Chr.17: 28527296-28527296 on Build GRCh38
rs960677282 Chr.17: 28527252-28527252 on Build GRCh38
rs771311912 Chr.17: 28527417-28527417 on Build GRCh38
rs1034756587 Chr.17: 28527242-28527242 on Build GRCh38
rs749618117 Chr.17: 28527409-28527409 on Build GRCh38
rs964119085 Chr.17: 28527565-28527565 on Build GRCh38
rs975821179 Chr.17: 28527374-28527374 on Build GRCh38

View Full Product Details