Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28573981-28574486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDOC
Entrez Gene ID
230
Gene Name
aldolase, fructose-bisphosphate C
Gene Aliases
ALDC
UniGene
Hs.155247
Gene Chromosome Location
Chr.17: 28573115-28576963 on Build GRCh38
Transcripts
NM_005165.2 XM_005257949.2 XM_011524556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1559461 c.769C>G p.L257V Chr.17: 28574097-28574097 on Build GRCh38
4064938 c.785C>A p.P262H Chr.17: 28574081-28574081 on Build GRCh38
1479368 c.626T>C p.V209A Chr.17: 28574240-28574240 on Build GRCh38
6008248 c.754G>A p.A252T Chr.17: 28574112-28574112 on Build GRCh38
3515406 c.673G>A p.E225K Chr.17: 28574193-28574193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190125715 Chr.17: 28574431-28574431 on Build GRCh38
rs765845189 Chr.17: 28574090-28574090 on Build GRCh38
rs754551689 Chr.17: 28574396-28574396 on Build GRCh38
rs746954627 Chr.17: 28574141-28574141 on Build GRCh38
rs747733387 Chr.17: 28574109-28574109 on Build GRCh38
rs376602783 Chr.17: 28574272-28574272 on Build GRCh38
rs749512701 Chr.17: 28574281-28574281 on Build GRCh38
rs894458011 Chr.17: 28574203-28574203 on Build GRCh38
rs367769875 Chr.17: 28574187-28574187 on Build GRCh38
rs200453652 Chr.17: 28574151-28574151 on Build GRCh38
rs372905293 Chr.17: 28574036-28574036 on Build GRCh38
rs886888484 Chr.17: 28574278-28574278 on Build GRCh38
rs1014335834 Chr.17: 28574107-28574107 on Build GRCh38
rs757201490 Chr.17: 28574248-28574248 on Build GRCh38
rs970044322 Chr.17: 28574161-28574161 on Build GRCh38
rs905872703 Chr.17: 28574372-28574372 on Build GRCh38
rs975307946 Chr.17: 28574462-28574462 on Build GRCh38
rs773023993 Chr.17: 28574034-28574034 on Build GRCh38
rs145665688 Chr.17: 28574093-28574093 on Build GRCh38
rs762273619 Chr.17: 28574186-28574186 on Build GRCh38
rs932310944 Chr.17: 28574299-28574299 on Build GRCh38
rs772110552 Chr.17: 28574017-28574017 on Build GRCh38
rs34236636 Chr.17: 28574038-28574038 on Build GRCh38
rs138076634 Chr.17: 28574179-28574179 on Build GRCh38
rs754322337 Chr.17: 28574197-28574197 on Build GRCh38
rs768900131 Chr.17: 28574283-28574283 on Build GRCh38
rs2228419 Chr.17: 28574176-28574176 on Build GRCh38
rs371286350 Chr.17: 28574221-28574221 on Build GRCh38
rs775892539 Chr.17: 28574053-28574053 on Build GRCh38
rs575831680 Chr.17: 28574200-28574200 on Build GRCh38
rs751068177 Chr.17: 28574349-28574349 on Build GRCh38
rs74902288 Chr.17: 28574086-28574086 on Build GRCh38
rs770873495 Chr.17: 28574150-28574150 on Build GRCh38
rs141206659 Chr.17: 28574170-28574170 on Build GRCh38
rs372027129 Chr.17: 28574249-28574249 on Build GRCh38
rs550685757 Chr.17: 28574284-28574284 on Build GRCh38
rs975275293 Chr.17: 28574153-28574153 on Build GRCh38
rs1044493464 Chr.17: 28574356-28574356 on Build GRCh38
rs1014388281 Chr.17: 28574350-28574350 on Build GRCh38
rs753375307 Chr.17: 28574091-28574091 on Build GRCh38
rs369770154 Chr.17: 28574286-28574286 on Build GRCh38
rs759797077 Chr.17: 28574043-28574043 on Build GRCh38
rs778701127 Chr.17: 28574094-28574094 on Build GRCh38
rs763223986 Chr.17: 28574058-28574058 on Build GRCh38
rs542170619 Chr.17: 28574395-28574395 on Build GRCh38
rs372331591 Chr.17: 28574460-28574460 on Build GRCh38
rs995264430 Chr.17: 28574042-28574042 on Build GRCh38
rs138221835 Chr.17: 28574159-28574159 on Build GRCh38
rs148066786 Chr.17: 28574435-28574436 on Build GRCh38
rs763465941 Chr.17: 28574091-28574091 on Build GRCh38
rs555229057 Chr.17: 28574311-28574311 on Build GRCh38
rs149519856 Chr.17: 28574224-28574224 on Build GRCh38
rs376126698 Chr.17: 28574015-28574015 on Build GRCh38
rs764777700 Chr.17: 28574219-28574219 on Build GRCh38
rs370804682 Chr.17: 28574120-28574120 on Build GRCh38
rs947187130 Chr.17: 28574167-28574167 on Build GRCh38
rs904722340 Chr.17: 28574085-28574085 on Build GRCh38
rs574770978 Chr.17: 28574318-28574318 on Build GRCh38
rs567996353 Chr.17: 28574064-28574064 on Build GRCh38
rs746800381 Chr.17: 28574235-28574235 on Build GRCh38
rs745936751 Chr.17: 28574158-28574158 on Build GRCh38
rs766399034 Chr.17: 28574221-28574222 on Build GRCh38
rs777472706 Chr.17: 28574229-28574229 on Build GRCh38
rs926186641 Chr.17: 28574321-28574321 on Build GRCh38
rs181289784 Chr.17: 28574184-28574184 on Build GRCh38
rs765425713 Chr.17: 28574044-28574044 on Build GRCh38
rs761211184 Chr.17: 28574449-28574449 on Build GRCh38
rs553608756 Chr.17: 28574134-28574134 on Build GRCh38
rs377287523 Chr.17: 28574063-28574063 on Build GRCh38
rs760607303 Chr.17: 28574035-28574035 on Build GRCh38
rs764875596 Chr.17: 28574450-28574450 on Build GRCh38
rs1053319765 Chr.17: 28574325-28574325 on Build GRCh38
rs1043310864 Chr.17: 28574082-28574082 on Build GRCh38
rs934838217 Chr.17: 28574324-28574324 on Build GRCh38
rs780067738 Chr.17: 28574275-28574275 on Build GRCh38
rs745740470 Chr.17: 28574260-28574260 on Build GRCh38
rs920915069 Chr.17: 28574020-28574020 on Build GRCh38

View Full Product Details