Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44683793-44684245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC43
Entrez Gene ID
124808
Gene Name
coiled-coil domain containing 43
Gene Aliases
-
UniGene
Hs.579115
Gene Chromosome Location
Chr.17: 44677437-44689797 on Build GRCh38
Transcripts
NM_001099225.1 NM_144609.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1147778 c.217C>T p.L73F Chr.17: 44683947-44683947 on Build GRCh38
4484027 c.275C>T p.T92I Chr.17: 44683889-44683889 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768900106 Chr.17: 44683912-44683912 on Build GRCh38
rs950649314 Chr.17: 44684137-44684137 on Build GRCh38
rs979039680 Chr.17: 44684070-44684070 on Build GRCh38
rs1004708322 Chr.17: 44684185-44684185 on Build GRCh38
rs773287547 Chr.17: 44683923-44683923 on Build GRCh38
rs1009400553 Chr.17: 44684029-44684029 on Build GRCh38
rs780044722 Chr.17: 44683995-44683995 on Build GRCh38
rs754628618 Chr.17: 44683937-44683937 on Build GRCh38
rs369330972 Chr.17: 44683917-44683917 on Build GRCh38
rs768577833 Chr.17: 44684005-44684005 on Build GRCh38
rs147870893 Chr.17: 44684171-44684172 on Build GRCh38
rs11549083 Chr.17: 44683902-44683902 on Build GRCh38
rs771249020 Chr.17: 44683830-44683830 on Build GRCh38
rs553980550 Chr.17: 44684117-44684117 on Build GRCh38
rs746312553 Chr.17: 44683895-44683895 on Build GRCh38
rs1038749331 Chr.17: 44684129-44684129 on Build GRCh38
rs1039010201 Chr.17: 44684063-44684063 on Build GRCh38
rs749355152 Chr.17: 44684002-44684002 on Build GRCh38
rs371343848 Chr.17: 44683824-44683824 on Build GRCh38
rs886325588 Chr.17: 44684201-44684201 on Build GRCh38
rs560666908 Chr.17: 44683843-44683843 on Build GRCh38
rs781465718 Chr.17: 44683975-44683975 on Build GRCh38
rs933108875 Chr.17: 44684172-44684172 on Build GRCh38
rs890508198 Chr.17: 44684028-44684028 on Build GRCh38
rs201785763 Chr.17: 44683926-44683926 on Build GRCh38
rs765865138 Chr.17: 44683929-44683929 on Build GRCh38
rs769864465 Chr.17: 44683896-44683896 on Build GRCh38
rs763140517 Chr.17: 44683904-44683904 on Build GRCh38
rs191820391 Chr.17: 44684011-44684011 on Build GRCh38
rs1027964295 Chr.17: 44684088-44684088 on Build GRCh38
rs753368790 Chr.17: 44683930-44683930 on Build GRCh38
rs35493229 Chr.17: 44683958-44683959 on Build GRCh38
rs774656031 Chr.17: 44684009-44684009 on Build GRCh38
rs900513885 Chr.17: 44684065-44684065 on Build GRCh38
rs1047645946 Chr.17: 44684175-44684175 on Build GRCh38
rs752247687 Chr.17: 44683861-44683861 on Build GRCh38
rs375171007 Chr.17: 44683950-44683950 on Build GRCh38
rs915519999 Chr.17: 44683882-44683882 on Build GRCh38
rs747371194 Chr.17: 44683828-44683828 on Build GRCh38
rs533725023 Chr.17: 44684075-44684075 on Build GRCh38
rs746437351 Chr.17: 44683988-44683988 on Build GRCh38
rs369517118 Chr.17: 44683990-44683990 on Build GRCh38
rs764975172 Chr.17: 44683940-44683940 on Build GRCh38
rs757635439 Chr.17: 44683971-44683971 on Build GRCh38
rs114682066 Chr.17: 44684071-44684071 on Build GRCh38
rs148980983 Chr.17: 44683916-44683916 on Build GRCh38
rs774052626 Chr.17: 44683913-44683913 on Build GRCh38
rs551781416 Chr.17: 44684135-44684135 on Build GRCh38

View Full Product Details