Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32862190-32862699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC5
Entrez Gene ID
252995
Gene Name
fibronectin type III domain containing 5
Gene Aliases
FRCP2, irisin
UniGene
Hs.524234
Gene Chromosome Location
Chr.1: 32862268-32872492 on Build GRCh38
Transcripts
NM_001171940.1 NM_001171941.2 NM_153756.2

SNP Detail

SNP ID Chromosome Location
rs950501698 Chr.1: 32862347-32862347 on Build GRCh38
rs911905051 Chr.1: 32862667-32862667 on Build GRCh38
rs3480 Chr.1: 32862564-32862564 on Build GRCh38
rs900670632 Chr.1: 32862296-32862296 on Build GRCh38
rs902229701 Chr.1: 32862251-32862251 on Build GRCh38
rs999173327 Chr.1: 32862627-32862627 on Build GRCh38
rs1039576980 Chr.1: 32862268-32862268 on Build GRCh38
rs748053787 Chr.1: 32862523-32862523 on Build GRCh38
rs974063411 Chr.1: 32862467-32862467 on Build GRCh38
rs962955488 Chr.1: 32862437-32862437 on Build GRCh38
rs781228918 Chr.1: 32862263-32862263 on Build GRCh38
rs1018163236 Chr.1: 32862394-32862394 on Build GRCh38
rs1004929656 Chr.1: 32862544-32862544 on Build GRCh38
rs372054809 Chr.1: 32862562-32862562 on Build GRCh38
rs1043791267 Chr.1: 32862246-32862246 on Build GRCh38
rs947627018 Chr.1: 32862591-32862591 on Build GRCh38
rs544110773 Chr.1: 32862345-32862345 on Build GRCh38
rs532694562 Chr.1: 32862547-32862547 on Build GRCh38
rs1029397448 Chr.1: 32862331-32862331 on Build GRCh38
rs768507080 Chr.1: 32862218-32862218 on Build GRCh38
rs578201136 Chr.1: 32862507-32862507 on Build GRCh38
rs1007156731 Chr.1: 32862349-32862349 on Build GRCh38
rs141940806 Chr.1: 32862557-32862557 on Build GRCh38
rs548697855 Chr.1: 32862602-32862602 on Build GRCh38
rs1034711863 Chr.1: 32862545-32862545 on Build GRCh38
rs1010540802 Chr.1: 32862235-32862235 on Build GRCh38
rs769349233 Chr.1: 32862556-32862556 on Build GRCh38
rs531247443 Chr.1: 32862666-32862666 on Build GRCh38
rs1042008156 Chr.1: 32862603-32862603 on Build GRCh38
rs183460978 Chr.1: 32862585-32862585 on Build GRCh38
rs955192326 Chr.1: 32862340-32862340 on Build GRCh38
rs925998051 Chr.1: 32862607-32862609 on Build GRCh38
rs918590144 Chr.1: 32862473-32862473 on Build GRCh38
rs981511921 Chr.1: 32862555-32862555 on Build GRCh38
rs565488826 Chr.1: 32862531-32862531 on Build GRCh38
rs146223915 Chr.1: 32862613-32862613 on Build GRCh38

View Full Product Details